Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48597529-48597776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB-AS3
Entrez Gene ID
404266
Gene Name
HOXB cluster antisense RNA 3
Gene Aliases
-
UniGene
Hs.660088
Gene Chromosome Location
Chr.17: 48590420-48606414 on Build GRCh38
Transcripts
NR_033201.2 NR_033202.2 NR_033203.1 NR_033204.2 NR_033205.1 NR_110329.1 NR_110330.1 NR_110331.1 NM_018952.4 XM_005257283.3 XM_005257284.3 XM_011524727.2

Gene Symbol
HOXB6
Entrez Gene ID
3216
Gene Name
homeobox B6
Gene Aliases
HOX2, HOX2B, HU-2, Hox-2.2
UniGene
Hs.98428
Gene Chromosome Location
Chr.17: 48595736-48604992 on Build GRCh38
Transcripts
NR_033201.2 NR_033202.2 NR_033203.1 NR_033204.2 NR_033205.1 NR_110329.1 NR_110330.1 NR_110331.1 NM_018952.4 XM_005257283.3 XM_005257284.3 XM_011524727.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6681713 c.397C>T p.R133W Chr.17: 48597754-48597754 on Build GRCh38
5594683 c.407C>T p.S136L Chr.17: 48597744-48597744 on Build GRCh38
6575032 c.381C>G p.V127V Chr.17: 48597770-48597770 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755606607 Chr.17: 48597717-48597717 on Build GRCh38
rs751202512 Chr.17: 48597706-48597706 on Build GRCh38
rs746873581 Chr.17: 48597715-48597716 on Build GRCh38
rs910719729 Chr.17: 48597708-48597708 on Build GRCh38
rs926285455 Chr.17: 48597567-48597567 on Build GRCh38
rs767182042 Chr.17: 48597710-48597710 on Build GRCh38
rs762667665 Chr.17: 48597687-48597687 on Build GRCh38
rs763879868 Chr.17: 48597697-48597697 on Build GRCh38
rs780344627 Chr.17: 48597705-48597706 on Build GRCh38
rs765823392 Chr.17: 48597733-48597733 on Build GRCh38
rs775414840 Chr.17: 48597684-48597684 on Build GRCh38
rs778154907 Chr.17: 48597749-48597749 on Build GRCh38
rs1032015745 Chr.17: 48597591-48597591 on Build GRCh38
rs1017528079 Chr.17: 48597719-48597719 on Build GRCh38
rs955705839 Chr.17: 48597592-48597592 on Build GRCh38
rs1021324651 Chr.17: 48597663-48597663 on Build GRCh38
rs990241076 Chr.17: 48597626-48597626 on Build GRCh38
rs976325526 Chr.17: 48597730-48597730 on Build GRCh38
rs753184788 Chr.17: 48597742-48597742 on Build GRCh38
rs1056416212 Chr.17: 48597615-48597618 on Build GRCh38
rs758723592 Chr.17: 48597746-48597746 on Build GRCh38

View Full Product Details