Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44510963-44511232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEACAM20
Entrez Gene ID
125931
Gene Name
carcinoembryonic antigen related cell adhesion molecule 20
Gene Aliases
UNQ9366
UniGene
Hs.689632
Gene Chromosome Location
Chr.19: 44503838-44530100 on Build GRCh38
Transcripts
NM_001102597.2 NM_001102598.2 NM_001102599.2 NM_001102600.2 XM_011526429.2 XM_011526430.2 XM_011526431.2 XM_011526432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3892818 c.1701C>T p.S567S Chr.19: 44511063-44511063 on Build GRCh38
3535609 c.1675C>T p.L559L Chr.19: 44511089-44511089 on Build GRCh38
7318299 c.1647C>T p.F549F Chr.19: 44511117-44511117 on Build GRCh38
354010 c.1708C>A p.P570T Chr.19: 44511056-44511056 on Build GRCh38
3535610 c.1609-1G>A p.? Chr.19: 44511156-44511156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762179860 Chr.19: 44511153-44511153 on Build GRCh38
rs781222936 Chr.19: 44511081-44511081 on Build GRCh38
rs376936436 Chr.19: 44511170-44511170 on Build GRCh38
rs998170414 Chr.19: 44511082-44511082 on Build GRCh38
rs758283920 Chr.19: 44510994-44510994 on Build GRCh38
rs750571699 Chr.19: 44511162-44511162 on Build GRCh38
rs115699318 Chr.19: 44511011-44511011 on Build GRCh38
rs769255358 Chr.19: 44511027-44511027 on Build GRCh38
rs756031325 Chr.19: 44511201-44511201 on Build GRCh38
rs754912537 Chr.19: 44511080-44511080 on Build GRCh38
rs368576958 Chr.19: 44511077-44511077 on Build GRCh38
rs534487383 Chr.19: 44511132-44511132 on Build GRCh38
rs770595487 Chr.19: 44511105-44511105 on Build GRCh38
rs116775350 Chr.19: 44511048-44511048 on Build GRCh38
rs558551827 Chr.19: 44511106-44511106 on Build GRCh38
rs754060965 Chr.19: 44510986-44510986 on Build GRCh38
rs760007098 Chr.19: 44511047-44511047 on Build GRCh38
rs764215242 Chr.19: 44511053-44511053 on Build GRCh38
rs183097658 Chr.19: 44511163-44511163 on Build GRCh38
rs748031361 Chr.19: 44511089-44511089 on Build GRCh38
rs570594938 Chr.19: 44511113-44511113 on Build GRCh38
rs372851437 Chr.19: 44511133-44511133 on Build GRCh38
rs772862763 Chr.19: 44511033-44511033 on Build GRCh38
rs1029689212 Chr.19: 44511083-44511083 on Build GRCh38
rs113140885 Chr.19: 44511184-44511184 on Build GRCh38
rs748998231 Chr.19: 44511096-44511096 on Build GRCh38
rs539857619 Chr.19: 44511092-44511092 on Build GRCh38
rs150406547 Chr.19: 44511145-44511145 on Build GRCh38
rs772273465 Chr.19: 44511124-44511124 on Build GRCh38
rs776151648 Chr.19: 44511127-44511127 on Build GRCh38
rs779774719 Chr.19: 44511075-44511075 on Build GRCh38
rs372035768 Chr.19: 44511161-44511161 on Build GRCh38
rs951414714 Chr.19: 44511100-44511100 on Build GRCh38
rs754827540 Chr.19: 44511168-44511168 on Build GRCh38
rs774764245 Chr.19: 44511043-44511043 on Build GRCh38
rs904355901 Chr.19: 44511028-44511028 on Build GRCh38
rs762524360 Chr.19: 44511034-44511034 on Build GRCh38
rs551142245 Chr.19: 44511025-44511025 on Build GRCh38
rs765849949 Chr.19: 44511027-44511028 on Build GRCh38
rs757552125 Chr.19: 44511072-44511072 on Build GRCh38
rs777258350 Chr.19: 44511095-44511095 on Build GRCh38
rs760857021 Chr.19: 44511050-44511050 on Build GRCh38
rs778509499 Chr.19: 44511114-44511114 on Build GRCh38
rs754119967 Chr.19: 44511065-44511065 on Build GRCh38
rs192847663 Chr.19: 44511154-44511154 on Build GRCh38
rs779833734 Chr.19: 44511000-44511000 on Build GRCh38
rs536744477 Chr.19: 44511176-44511176 on Build GRCh38
rs771267427 Chr.19: 44511042-44511042 on Build GRCh38
rs768628431 Chr.19: 44511007-44511007 on Build GRCh38
rs369521717 Chr.19: 44511194-44511194 on Build GRCh38
rs753102567 Chr.19: 44511049-44511049 on Build GRCh38
rs200684821 Chr.19: 44511134-44511134 on Build GRCh38
rs868809231 Chr.19: 44511191-44511191 on Build GRCh38
rs866957364 Chr.19: 44511066-44511066 on Build GRCh38

View Full Product Details