Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57764962-57765223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP27B1
Entrez Gene ID
1594
Gene Name
cytochrome P450 family 27 subfamily B member 1
Gene Aliases
CP2B, CYP1, CYP1alpha, CYP27B, P450c1, PDDR, VDD1, VDDR, VDDRI, VDR
UniGene
Hs.524528
Gene Chromosome Location
Chr.12: 57762334-57767193 on Build GRCh38
Transcripts
NM_000785.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
42726 c.754C>T p.R252C Chr.12: 57765047-57765047 on Build GRCh38
4043952 c.631G>T p.E211* Chr.12: 57765170-57765170 on Build GRCh38
5515491 c.670_686del17 p.A224fs*103 Chr.12: 57765115-57765131 on Build GRCh38
549517 c.591C>A p.G197G Chr.12: 57765210-57765210 on Build GRCh38
4405115 c.721T>A p.W241R Chr.12: 57765080-57765080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs387906259 Chr.12: 57765109-57765109 on Build GRCh38
rs780269626 Chr.12: 57765070-57765070 on Build GRCh38
rs775114472 Chr.12: 57765097-57765097 on Build GRCh38
rs772898470 Chr.12: 57765074-57765074 on Build GRCh38
rs999392694 Chr.12: 57764984-57764984 on Build GRCh38
rs1028387024 Chr.12: 57765047-57765047 on Build GRCh38
rs752696414 Chr.12: 57765022-57765022 on Build GRCh38
rs778534360 Chr.12: 57765145-57765145 on Build GRCh38
rs746021709 Chr.12: 57765066-57765066 on Build GRCh38
rs774153859 Chr.12: 57765084-57765084 on Build GRCh38
rs769013170 Chr.12: 57765073-57765073 on Build GRCh38
rs892730807 Chr.12: 57764988-57764988 on Build GRCh38
rs767152707 Chr.12: 57765008-57765008 on Build GRCh38
rs1009781420 Chr.12: 57765093-57765093 on Build GRCh38
rs886049727 Chr.12: 57765013-57765013 on Build GRCh38
rs753733417 Chr.12: 57765123-57765123 on Build GRCh38
rs764038309 Chr.12: 57765120-57765120 on Build GRCh38
rs757277620 Chr.12: 57765048-57765048 on Build GRCh38
rs770528985 Chr.12: 57765081-57765081 on Build GRCh38
rs952793854 Chr.12: 57765200-57765200 on Build GRCh38
rs751848951 Chr.12: 57765179-57765179 on Build GRCh38
rs769953744 Chr.12: 57765150-57765150 on Build GRCh38
rs957570758 Chr.12: 57765033-57765033 on Build GRCh38
rs779137478 Chr.12: 57765059-57765059 on Build GRCh38
rs1054858813 Chr.12: 57765173-57765173 on Build GRCh38
rs1033180599 Chr.12: 57765003-57765003 on Build GRCh38
rs1033790241 Chr.12: 57765185-57765185 on Build GRCh38
rs387906258 Chr.12: 57765171-57765171 on Build GRCh38
rs758802757 Chr.12: 57765068-57765068 on Build GRCh38
rs758546382 Chr.12: 57765174-57765174 on Build GRCh38
rs149518825 Chr.12: 57765134-57765134 on Build GRCh38
rs770581711 Chr.12: 57765191-57765191 on Build GRCh38
rs974435159 Chr.12: 57765142-57765142 on Build GRCh38
rs745571910 Chr.12: 57765197-57765197 on Build GRCh38
rs771598984 Chr.12: 57765200-57765200 on Build GRCh38
rs748602968 Chr.12: 57765187-57765187 on Build GRCh38
rs190470241 Chr.12: 57765037-57765037 on Build GRCh38
rs1025413409 Chr.12: 57765104-57765104 on Build GRCh38
rs184727850 Chr.12: 57765004-57765004 on Build GRCh38
rs61734540 Chr.12: 57765111-57765111 on Build GRCh38
rs1002601613 Chr.12: 57765127-57765127 on Build GRCh38
rs373542360 Chr.12: 57765095-57765095 on Build GRCh38
rs748833715 Chr.12: 57765079-57765079 on Build GRCh38
rs745723541 Chr.12: 57765148-57765160 on Build GRCh38
rs200174985 Chr.12: 57765035-57765035 on Build GRCh38
rs745908985 Chr.12: 57765057-57765057 on Build GRCh38
rs572765133 Chr.12: 57765186-57765186 on Build GRCh38
rs757283928 Chr.12: 57765143-57765152 on Build GRCh38
rs772171082 Chr.12: 57765155-57765159 on Build GRCh38
rs774100842 Chr.12: 57764996-57764996 on Build GRCh38
rs753888767 Chr.12: 57765041-57765041 on Build GRCh38
rs937852515 Chr.12: 57765156-57765156 on Build GRCh38
rs367743385 Chr.12: 57765131-57765131 on Build GRCh38
rs201503366 Chr.12: 57765175-57765175 on Build GRCh38
rs374481894 Chr.12: 57765038-57765038 on Build GRCh38
rs765241743 Chr.12: 57765132-57765132 on Build GRCh38
rs370743468 Chr.12: 57765183-57765183 on Build GRCh38
rs759410154 Chr.12: 57765005-57765005 on Build GRCh38
rs759056278 Chr.12: 57765090-57765090 on Build GRCh38
rs961320678 Chr.12: 57765135-57765135 on Build GRCh38

View Full Product Details