Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176123111-176123354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD9
Entrez Gene ID
3235
Gene Name
homeobox D9
Gene Aliases
HOX4, HOX4C, Hox-4.3, Hox-5.2
UniGene
Hs.236646
Gene Chromosome Location
Chr.2: 176122685-176124917 on Build GRCh38
Transcripts
NM_014213.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5416681 c.396C>A p.G132G Chr.2: 176123194-176123194 on Build GRCh38
1659335 c.398G>A p.R133H Chr.2: 176123196-176123196 on Build GRCh38
5474366 c.541T>A p.S181T Chr.2: 176123309-176123309 on Build GRCh38
6010405 c.434C>T p.P145L Chr.2: 176123202-176123202 on Build GRCh38
5474367 c.511T>A p.S171T Chr.2: 176123309-176123309 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747468230 Chr.2: 176123202-176123202 on Build GRCh38
rs34862292 Chr.2: 176123331-176123331 on Build GRCh38
rs769173326 Chr.2: 176123226-176123226 on Build GRCh38
rs915064158 Chr.2: 176123171-176123171 on Build GRCh38
rs779099311 Chr.2: 176123139-176123139 on Build GRCh38
rs745324831 Chr.2: 176123277-176123277 on Build GRCh38
rs1043702848 Chr.2: 176123197-176123197 on Build GRCh38
rs1036732609 Chr.2: 176123258-176123258 on Build GRCh38
rs762371628 Chr.2: 176123278-176123278 on Build GRCh38
rs898140490 Chr.2: 176123273-176123273 on Build GRCh38
rs767343328 Chr.2: 176123299-176123299 on Build GRCh38
rs539591023 Chr.2: 176123138-176123138 on Build GRCh38
rs745923004 Chr.2: 176123160-176123160 on Build GRCh38
rs780718652 Chr.2: 176123193-176123193 on Build GRCh38
rs1015721717 Chr.2: 176123288-176123288 on Build GRCh38
rs545576144 Chr.2: 176123337-176123337 on Build GRCh38
rs759433381 Chr.2: 176123289-176123289 on Build GRCh38
rs752413649 Chr.2: 176123304-176123304 on Build GRCh38
rs1011115309 Chr.2: 176123174-176123174 on Build GRCh38
rs956848008 Chr.2: 176123152-176123152 on Build GRCh38
rs927929743 Chr.2: 176123140-176123140 on Build GRCh38
rs973974359 Chr.2: 176123300-176123300 on Build GRCh38
rs576187632 Chr.2: 176123262-176123262 on Build GRCh38
rs944378796 Chr.2: 176123207-176123207 on Build GRCh38
rs746053967 Chr.2: 176123321-176123323 on Build GRCh38
rs778980335 Chr.2: 176123298-176123300 on Build GRCh38
rs765820936 Chr.2: 176123283-176123283 on Build GRCh38
rs946500426 Chr.2: 176123186-176123186 on Build GRCh38
rs370816034 Chr.2: 176123328-176123328 on Build GRCh38
rs759012736 Chr.2: 176123166-176123166 on Build GRCh38
rs191379716 Chr.2: 176123231-176123231 on Build GRCh38
rs769313756 Chr.2: 176123269-176123269 on Build GRCh38
rs757557164 Chr.2: 176123318-176123318 on Build GRCh38
rs899742500 Chr.2: 176123198-176123198 on Build GRCh38
rs757538742 Chr.2: 176123267-176123278 on Build GRCh38
rs988051029 Chr.2: 176123163-176123163 on Build GRCh38
rs962277643 Chr.2: 176123293-176123293 on Build GRCh38
rs1052920007 Chr.2: 176123130-176123130 on Build GRCh38
rs772843535 Chr.2: 176123276-176123276 on Build GRCh38
rs878866381 Chr.2: 176123158-176123159 on Build GRCh38
rs711821 Chr.2: 176123287-176123287 on Build GRCh38

View Full Product Details