Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 67018094-67018359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG4
Entrez Gene ID
27092
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 4
Gene Aliases
-
UniGene
Hs.514423
Gene Chromosome Location
Chr.17: 66964864-67033402 on Build GRCh38
Transcripts
NM_014405.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
332303 c.292G>T p.E98* Chr.17: 67018260-67018260 on Build GRCh38
5923195 c.259C>T p.P87S Chr.17: 67018227-67018227 on Build GRCh38
1710670 c.292G>A p.E98K Chr.17: 67018260-67018260 on Build GRCh38
1710669 c.253C>T p.H85Y Chr.17: 67018221-67018221 on Build GRCh38
294209 c.277G>A p.D93N Chr.17: 67018245-67018245 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753019358 Chr.17: 67018178-67018178 on Build GRCh38
rs113216164 Chr.17: 67018331-67018331 on Build GRCh38
rs186352782 Chr.17: 67018250-67018250 on Build GRCh38
rs939401352 Chr.17: 67018335-67018335 on Build GRCh38
rs544483021 Chr.17: 67018128-67018128 on Build GRCh38
rs774987460 Chr.17: 67018273-67018273 on Build GRCh38
rs751824454 Chr.17: 67018160-67018160 on Build GRCh38
rs866944756 Chr.17: 67018209-67018209 on Build GRCh38
rs34743848 Chr.17: 67018244-67018244 on Build GRCh38
rs772670298 Chr.17: 67018258-67018258 on Build GRCh38
rs758357413 Chr.17: 67018212-67018212 on Build GRCh38
rs539209686 Chr.17: 67018314-67018314 on Build GRCh38
rs756360843 Chr.17: 67018303-67018303 on Build GRCh38
rs138208538 Chr.17: 67018184-67018184 on Build GRCh38
rs200886756 Chr.17: 67018243-67018243 on Build GRCh38
rs367886594 Chr.17: 67018134-67018134 on Build GRCh38
rs370769158 Chr.17: 67018167-67018167 on Build GRCh38
rs964813318 Chr.17: 67018269-67018269 on Build GRCh38
rs987415514 Chr.17: 67018328-67018328 on Build GRCh38
rs759804398 Chr.17: 67018284-67018284 on Build GRCh38
rs112211002 Chr.17: 67018214-67018214 on Build GRCh38
rs78727776 Chr.17: 67018138-67018138 on Build GRCh38
rs999312930 Chr.17: 67018202-67018202 on Build GRCh38
rs1013055437 Chr.17: 67018196-67018196 on Build GRCh38
rs763385383 Chr.17: 67018145-67018145 on Build GRCh38
rs753114643 Chr.17: 67018288-67018288 on Build GRCh38
rs113600789 Chr.17: 67018313-67018313 on Build GRCh38
rs767865711 Chr.17: 67018285-67018285 on Build GRCh38
rs772725679 Chr.17: 67018144-67018144 on Build GRCh38
rs753944048 Chr.17: 67018316-67018316 on Build GRCh38
rs920649210 Chr.17: 67018317-67018317 on Build GRCh38
rs955916924 Chr.17: 67018305-67018305 on Build GRCh38
rs1031545966 Chr.17: 67018265-67018265 on Build GRCh38
rs768565187 Chr.17: 67018233-67018233 on Build GRCh38
rs372687382 Chr.17: 67018190-67018190 on Build GRCh38
rs955108285 Chr.17: 67018220-67018220 on Build GRCh38
rs757449342 Chr.17: 67018322-67018322 on Build GRCh38
rs776136999 Chr.17: 67018235-67018235 on Build GRCh38
rs201531279 Chr.17: 67018213-67018213 on Build GRCh38
rs920568341 Chr.17: 67018119-67018119 on Build GRCh38
rs753612032 Chr.17: 67018332-67018332 on Build GRCh38
rs56107700 Chr.17: 67018333-67018333 on Build GRCh38
rs376140158 Chr.17: 67018177-67018177 on Build GRCh38
rs756348036 Chr.17: 67018113-67018113 on Build GRCh38
rs373201649 Chr.17: 67018152-67018152 on Build GRCh38

View Full Product Details