Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86531100-86531333 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPNE3
Entrez Gene ID
8895
Gene Name
copine 3
Gene Aliases
CPN3, PRO1071
UniGene
Hs.191219
Gene Chromosome Location
Chr.8: 86514423-86561498 on Build GRCh38
Transcripts
NM_003909.4 XM_005251093.4 XM_017013945.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3901877 c.386C>T p.T129M Chr.8: 86531228-86531228 on Build GRCh38
7426785 c.359G>A p.G120D Chr.8: 86531201-86531201 on Build GRCh38
1102192 c.339A>G p.R113R Chr.8: 86531181-86531181 on Build GRCh38
4445707 c.313-3C>A p.? Chr.8: 86531152-86531152 on Build GRCh38
1661261 c.338G>A p.R113Q Chr.8: 86531180-86531180 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779754739 Chr.8: 86531271-86531271 on Build GRCh38
rs768582756 Chr.8: 86531197-86531197 on Build GRCh38
rs749400012 Chr.8: 86531175-86531175 on Build GRCh38
rs775396018 Chr.8: 86531185-86531185 on Build GRCh38
rs531293592 Chr.8: 86531148-86531148 on Build GRCh38
rs746149003 Chr.8: 86531264-86531264 on Build GRCh38
rs761404417 Chr.8: 86531212-86531212 on Build GRCh38
rs1009358591 Chr.8: 86531276-86531276 on Build GRCh38
rs76328401 Chr.8: 86531228-86531228 on Build GRCh38
rs374650748 Chr.8: 86531247-86531247 on Build GRCh38
rs374191824 Chr.8: 86531286-86531286 on Build GRCh38
rs753304318 Chr.8: 86531134-86531134 on Build GRCh38
rs982430940 Chr.8: 86531136-86531136 on Build GRCh38
rs753118449 Chr.8: 86531230-86531230 on Build GRCh38
rs367956394 Chr.8: 86531229-86531229 on Build GRCh38
rs182868985 Chr.8: 86531161-86531161 on Build GRCh38
rs749690809 Chr.8: 86531150-86531150 on Build GRCh38
rs562348920 Chr.8: 86531131-86531131 on Build GRCh38
rs374711387 Chr.8: 86531152-86531152 on Build GRCh38
rs764755747 Chr.8: 86531223-86531223 on Build GRCh38
rs1042363687 Chr.8: 86531270-86531270 on Build GRCh38
rs1027936909 Chr.8: 86531304-86531304 on Build GRCh38
rs370279023 Chr.8: 86531285-86531285 on Build GRCh38
rs377733818 Chr.8: 86531204-86531204 on Build GRCh38
rs986204950 Chr.8: 86531241-86531241 on Build GRCh38
rs76425775 Chr.8: 86531279-86531279 on Build GRCh38
rs755559079 Chr.8: 86531171-86531171 on Build GRCh38
rs753822116 Chr.8: 86531249-86531249 on Build GRCh38
rs760670043 Chr.8: 86531190-86531190 on Build GRCh38
rs949154766 Chr.8: 86531188-86531188 on Build GRCh38
rs944449299 Chr.8: 86531281-86531281 on Build GRCh38
rs771847586 Chr.8: 86531265-86531265 on Build GRCh38
rs778984085 Chr.8: 86531259-86531259 on Build GRCh38
rs769219061 Chr.8: 86531173-86531173 on Build GRCh38
rs756632790 Chr.8: 86531244-86531244 on Build GRCh38
rs746815558 Chr.8: 86531273-86531273 on Build GRCh38
rs745622693 Chr.8: 86531180-86531180 on Build GRCh38
rs779231067 Chr.8: 86531179-86531179 on Build GRCh38
rs149300782 Chr.8: 86531254-86531254 on Build GRCh38
rs567930517 Chr.8: 86531178-86531178 on Build GRCh38

View Full Product Details