Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47839854-47840079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRX
Entrez Gene ID
1406
Gene Name
cone-rod homeobox
Gene Aliases
CORD2, CRD, LCA7, OTX3
UniGene
Hs.617342
Gene Chromosome Location
Chr.19: 47821842-47843330 on Build GRCh38
Transcripts
NM_000554.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3990136 c.877G>T p.A293S Chr.19: 47839944-47839944 on Build GRCh38
5441077 c.791T>C p.V264A Chr.19: 47839858-47839858 on Build GRCh38
712133 c.856C>T p.L286L Chr.19: 47839923-47839923 on Build GRCh38
6085229 c.815C>A p.P272H Chr.19: 47839882-47839882 on Build GRCh38
6003979 c.839A>G p.Y280C Chr.19: 47839906-47839906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770494420 Chr.19: 47839901-47839901 on Build GRCh38
rs764137500 Chr.19: 47839961-47839961 on Build GRCh38
rs371493617 Chr.19: 47839972-47839972 on Build GRCh38
rs756636410 Chr.19: 47839886-47839886 on Build GRCh38
rs527236063 Chr.19: 47839964-47839964 on Build GRCh38
rs201875881 Chr.19: 47839904-47839904 on Build GRCh38
rs79186398 Chr.19: 47839986-47839986 on Build GRCh38
rs755208952 Chr.19: 47839878-47839878 on Build GRCh38
rs779790279 Chr.19: 47840011-47840011 on Build GRCh38
rs751630154 Chr.19: 47839974-47839974 on Build GRCh38
rs781453718 Chr.19: 47839881-47839881 on Build GRCh38
rs978128869 Chr.19: 47839995-47839995 on Build GRCh38
rs570266728 Chr.19: 47839892-47839892 on Build GRCh38
rs373098327 Chr.19: 47840001-47840001 on Build GRCh38
rs375770558 Chr.19: 47839973-47839973 on Build GRCh38
rs281865203 Chr.19: 47839885-47839885 on Build GRCh38
rs771721958 Chr.19: 47839919-47839919 on Build GRCh38
rs369336775 Chr.19: 47839982-47839982 on Build GRCh38
rs534975253 Chr.19: 47840014-47840014 on Build GRCh38
rs775175937 Chr.19: 47839925-47839925 on Build GRCh38
rs554988994 Chr.19: 47840050-47840050 on Build GRCh38
rs769100863 Chr.19: 47840046-47840046 on Build GRCh38
rs886054547 Chr.19: 47839924-47839924 on Build GRCh38
rs375168206 Chr.19: 47839994-47839994 on Build GRCh38
rs572089877 Chr.19: 47840047-47840047 on Build GRCh38
rs745553387 Chr.19: 47839911-47839911 on Build GRCh38
rs760493282 Chr.19: 47839956-47839956 on Build GRCh38
rs745339682 Chr.19: 47840002-47840002 on Build GRCh38
rs959402881 Chr.19: 47840025-47840025 on Build GRCh38
rs146699567 Chr.19: 47839999-47839999 on Build GRCh38
rs995204794 Chr.19: 47839934-47839934 on Build GRCh38
rs1025075301 Chr.19: 47840051-47840051 on Build GRCh38
rs766870005 Chr.19: 47839998-47839998 on Build GRCh38
rs371964860 Chr.19: 47839983-47839983 on Build GRCh38
rs773897663 Chr.19: 47839903-47839903 on Build GRCh38
rs755187618 Chr.19: 47839977-47839977 on Build GRCh38
rs778106655 Chr.19: 47839992-47839992 on Build GRCh38

View Full Product Details