Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110919019-110919292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYL2
Entrez Gene ID
4633
Gene Name
myosin light chain 2
Gene Aliases
CMH10, MLC-2s/v, MLC2
UniGene
Hs.75535
Gene Chromosome Location
Chr.12: 110910819-110920600 on Build GRCh38
Transcripts
NM_000432.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3456033 c.63C>T p.F21F Chr.12: 110919134-110919134 on Build GRCh38
1639034 c.4-8delT p.? Chr.12: 110919201-110919201 on Build GRCh38
3980761 c.29C>A p.A10D Chr.12: 110919168-110919168 on Build GRCh38
5827117 c.4-3delC p.? Chr.12: 110919196-110919196 on Build GRCh38
6719623 c.30C>T p.A10A Chr.12: 110919167-110919167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770204363 Chr.12: 110919229-110919229 on Build GRCh38
rs730880942 Chr.12: 110919169-110919169 on Build GRCh38
rs758688050 Chr.12: 110919202-110919202 on Build GRCh38
rs375567712 Chr.12: 110919056-110919056 on Build GRCh38
rs201580029 Chr.12: 110919095-110919095 on Build GRCh38
rs199474806 Chr.12: 110919206-110919206 on Build GRCh38
rs10849917 Chr.12: 110919270-110919270 on Build GRCh38
rs781578488 Chr.12: 110919223-110919223 on Build GRCh38
rs750730703 Chr.12: 110919201-110919201 on Build GRCh38
rs768776836 Chr.12: 110919198-110919201 on Build GRCh38
rs374870836 Chr.12: 110919198-110919198 on Build GRCh38
rs756245911 Chr.12: 110919201-110919201 on Build GRCh38
rs773565112 Chr.12: 110919233-110919233 on Build GRCh38
rs1041603211 Chr.12: 110919082-110919082 on Build GRCh38
rs104894370 Chr.12: 110919145-110919145 on Build GRCh38
rs12301951 Chr.12: 110919207-110919207 on Build GRCh38
rs753749960 Chr.12: 110919087-110919087 on Build GRCh38
rs112595556 Chr.12: 110919196-110919196 on Build GRCh38
rs868306878 Chr.12: 110919128-110919128 on Build GRCh38
rs104894363 Chr.12: 110919160-110919160 on Build GRCh38
rs955921220 Chr.12: 110919222-110919222 on Build GRCh38
rs727504300 Chr.12: 110919151-110919152 on Build GRCh38
rs749260543 Chr.12: 110919216-110919217 on Build GRCh38
rs762185101 Chr.12: 110919152-110919152 on Build GRCh38
rs730880943 Chr.12: 110919148-110919148 on Build GRCh38
rs747082468 Chr.12: 110919182-110919186 on Build GRCh38
rs765315783 Chr.12: 110919226-110919226 on Build GRCh38
rs762479350 Chr.12: 110919072-110919072 on Build GRCh38
rs111443790 Chr.12: 110919102-110919102 on Build GRCh38
rs752641375 Chr.12: 110919167-110919167 on Build GRCh38
rs878853980 Chr.12: 110919155-110919155 on Build GRCh38
rs730880933 Chr.12: 110919211-110919211 on Build GRCh38
rs397516410 Chr.12: 110919113-110919113 on Build GRCh38
rs867996559 Chr.12: 110919112-110919112 on Build GRCh38
rs113465073 Chr.12: 110919101-110919101 on Build GRCh38
rs762584624 Chr.12: 110919162-110919162 on Build GRCh38
rs199474816 Chr.12: 110919139-110919139 on Build GRCh38
rs1021632743 Chr.12: 110919272-110919272 on Build GRCh38
rs781431079 Chr.12: 110919150-110919150 on Build GRCh38
rs886039195 Chr.12: 110919174-110919174 on Build GRCh38
rs397516408 Chr.12: 110919117-110919117 on Build GRCh38
rs372595991 Chr.12: 110919215-110919215 on Build GRCh38
rs751576646 Chr.12: 110919081-110919081 on Build GRCh38
rs766186218 Chr.12: 110919080-110919080 on Build GRCh38
rs780255697 Chr.12: 110919203-110919203 on Build GRCh38
rs104894368 Chr.12: 110919133-110919133 on Build GRCh38
rs1041275916 Chr.12: 110919236-110919236 on Build GRCh38
rs755926255 Chr.12: 110919119-110919119 on Build GRCh38
rs113167834 Chr.12: 110919138-110919138 on Build GRCh38
rs730880941 Chr.12: 110919183-110919183 on Build GRCh38
rs200619503 Chr.12: 110919080-110919080 on Build GRCh38
rs781118169 Chr.12: 110919091-110919091 on Build GRCh38
rs886048961 Chr.12: 110919196-110919196 on Build GRCh38
rs550302003 Chr.12: 110919050-110919050 on Build GRCh38
rs199742269 Chr.12: 110919164-110919164 on Build GRCh38
rs552004172 Chr.12: 110919149-110919149 on Build GRCh38
rs779983470 Chr.12: 110919134-110919134 on Build GRCh38
rs139794370 Chr.12: 110919161-110919161 on Build GRCh38
rs905741278 Chr.12: 110919181-110919181 on Build GRCh38
rs762166340 Chr.12: 110919054-110919054 on Build GRCh38
rs531661702 Chr.12: 110919207-110919207 on Build GRCh38
rs763957786 Chr.12: 110919190-110919190 on Build GRCh38
rs538286849 Chr.12: 110919255-110919255 on Build GRCh38
rs397516402 Chr.12: 110919166-110919166 on Build GRCh38
rs754871718 Chr.12: 110919089-110919089 on Build GRCh38
rs750026287 Chr.12: 110919057-110919057 on Build GRCh38
rs730880937 Chr.12: 110919163-110919163 on Build GRCh38
rs730880944 Chr.12: 110919144-110919144 on Build GRCh38
rs754744244 Chr.12: 110919136-110919146 on Build GRCh38
rs397516409 Chr.12: 110919115-110919115 on Build GRCh38
rs751392310 Chr.12: 110919103-110919105 on Build GRCh38

View Full Product Details