Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 38727542-38727803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC32A1
Entrez Gene ID
140679
Gene Name
solute carrier family 32 member 1
Gene Aliases
VGAT, VIAAT
UniGene
Hs.179080
Gene Chromosome Location
Chr.20: 38724462-38729372 on Build GRCh38
Transcripts
NM_080552.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6781514 c.548G>A p.R183Q Chr.20: 38727609-38727609 on Build GRCh38
1026691 c.503C>T p.A168V Chr.20: 38727564-38727564 on Build GRCh38
6046912 c.573C>T p.N191N Chr.20: 38727634-38727634 on Build GRCh38
4429637 c.529G>A p.G177S Chr.20: 38727590-38727590 on Build GRCh38
3911288 c.585C>T p.A195A Chr.20: 38727646-38727646 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772284025 Chr.20: 38727719-38727719 on Build GRCh38
rs968205956 Chr.20: 38727564-38727564 on Build GRCh38
rs201506462 Chr.20: 38727682-38727682 on Build GRCh38
rs777297939 Chr.20: 38727767-38727767 on Build GRCh38
rs922868461 Chr.20: 38727630-38727630 on Build GRCh38
rs751200572 Chr.20: 38727667-38727667 on Build GRCh38
rs749366553 Chr.20: 38727636-38727636 on Build GRCh38
rs770350121 Chr.20: 38727772-38727772 on Build GRCh38
rs761280843 Chr.20: 38727660-38727660 on Build GRCh38
rs779013587 Chr.20: 38727715-38727715 on Build GRCh38
rs375927191 Chr.20: 38727694-38727694 on Build GRCh38
rs143574180 Chr.20: 38727574-38727574 on Build GRCh38
rs764847103 Chr.20: 38727575-38727575 on Build GRCh38
rs759168820 Chr.20: 38727781-38727781 on Build GRCh38
rs780628581 Chr.20: 38727727-38727727 on Build GRCh38
rs772884195 Chr.20: 38727661-38727661 on Build GRCh38
rs772357025 Chr.20: 38727648-38727648 on Build GRCh38
rs368724585 Chr.20: 38727701-38727701 on Build GRCh38
rs886696860 Chr.20: 38727605-38727605 on Build GRCh38
rs780997867 Chr.20: 38727589-38727589 on Build GRCh38
rs567071470 Chr.20: 38727738-38727738 on Build GRCh38
rs150170740 Chr.20: 38727718-38727718 on Build GRCh38
rs752635600 Chr.20: 38727590-38727590 on Build GRCh38
rs746056178 Chr.20: 38727646-38727646 on Build GRCh38
rs370827944 Chr.20: 38727697-38727697 on Build GRCh38
rs756110955 Chr.20: 38727684-38727684 on Build GRCh38
rs377010570 Chr.20: 38727578-38727578 on Build GRCh38
rs765312308 Chr.20: 38727643-38727643 on Build GRCh38
rs1051897820 Chr.20: 38727591-38727591 on Build GRCh38
rs140808070 Chr.20: 38727639-38727639 on Build GRCh38
rs777888313 Chr.20: 38727619-38727619 on Build GRCh38
rs755986414 Chr.20: 38727616-38727616 on Build GRCh38
rs778982202 Chr.20: 38727644-38727644 on Build GRCh38
rs769332696 Chr.20: 38727749-38727749 on Build GRCh38
rs765861934 Chr.20: 38727664-38727664 on Build GRCh38
rs758187434 Chr.20: 38727576-38727576 on Build GRCh38
rs759565716 Chr.20: 38727583-38727583 on Build GRCh38
rs767228334 Chr.20: 38727674-38727674 on Build GRCh38
rs761227662 Chr.20: 38727563-38727563 on Build GRCh38
rs775935038 Chr.20: 38727657-38727657 on Build GRCh38

View Full Product Details