Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70276858-70277111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCYOX1
Entrez Gene ID
51449
Gene Name
prenylcysteine oxidase 1
Gene Aliases
PCL1
UniGene
Hs.567502
Gene Chromosome Location
Chr.2: 70258099-70281185 on Build GRCh38
Transcripts
NM_016297.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4770248 c.1103A>G p.N368S Chr.2: 70276977-70276977 on Build GRCh38
3839901 c.1168A>T p.R390* Chr.2: 70277042-70277042 on Build GRCh38
4838371 c.1012C>T p.Q338* Chr.2: 70276886-70276886 on Build GRCh38
3582794 c.991C>T p.P331S Chr.2: 70276865-70276865 on Build GRCh38
1752620 c.1024C>T p.H342Y Chr.2: 70276898-70276898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146213046 Chr.2: 70277030-70277030 on Build GRCh38
rs773692111 Chr.2: 70277083-70277083 on Build GRCh38
rs78117294 Chr.2: 70277037-70277037 on Build GRCh38
rs756726852 Chr.2: 70277067-70277067 on Build GRCh38
rs756127044 Chr.2: 70276961-70276961 on Build GRCh38
rs776226532 Chr.2: 70276914-70276914 on Build GRCh38
rs769418136 Chr.2: 70277044-70277044 on Build GRCh38
rs780923578 Chr.2: 70277035-70277035 on Build GRCh38
rs148301071 Chr.2: 70276972-70276972 on Build GRCh38
rs144903509 Chr.2: 70277011-70277011 on Build GRCh38
rs369034697 Chr.2: 70277048-70277048 on Build GRCh38
rs747599132 Chr.2: 70277046-70277047 on Build GRCh38
rs902114505 Chr.2: 70276935-70276937 on Build GRCh38
rs748919442 Chr.2: 70277063-70277063 on Build GRCh38
rs762046573 Chr.2: 70277069-70277069 on Build GRCh38
rs535546126 Chr.2: 70277084-70277084 on Build GRCh38
rs372245978 Chr.2: 70276890-70276890 on Build GRCh38
rs774760948 Chr.2: 70276950-70276950 on Build GRCh38
rs771837637 Chr.2: 70276888-70276888 on Build GRCh38
rs377054846 Chr.2: 70276937-70276937 on Build GRCh38
rs745329841 Chr.2: 70276916-70276916 on Build GRCh38
rs762364389 Chr.2: 70276955-70276955 on Build GRCh38
rs983238750 Chr.2: 70276957-70276957 on Build GRCh38
rs764516298 Chr.2: 70276944-70276944 on Build GRCh38
rs200321499 Chr.2: 70276953-70276953 on Build GRCh38
rs547773653 Chr.2: 70276958-70276958 on Build GRCh38
rs776458022 Chr.2: 70277041-70277071 on Build GRCh38
rs746929394 Chr.2: 70276900-70276900 on Build GRCh38
rs888818592 Chr.2: 70276894-70276894 on Build GRCh38
rs372017809 Chr.2: 70277021-70277021 on Build GRCh38
rs570286043 Chr.2: 70277080-70277080 on Build GRCh38
rs865890023 Chr.2: 70276971-70276971 on Build GRCh38
rs769572406 Chr.2: 70276929-70276929 on Build GRCh38
rs747947804 Chr.2: 70276886-70276886 on Build GRCh38
rs757247920 Chr.2: 70277022-70277022 on Build GRCh38
rs1038568126 Chr.2: 70276974-70276974 on Build GRCh38
rs754051133 Chr.2: 70276996-70276996 on Build GRCh38
rs898788427 Chr.2: 70276995-70276995 on Build GRCh38
rs369390896 Chr.2: 70276997-70276997 on Build GRCh38
rs775227432 Chr.2: 70276936-70276936 on Build GRCh38
rs149137513 Chr.2: 70276902-70276902 on Build GRCh38

View Full Product Details