Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17225736-17225987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PADI1
Entrez Gene ID
29943
Gene Name
peptidyl arginine deiminase 1
Gene Aliases
HPAD10, PAD1, PDI, PDI1
UniGene
Hs.412941
Gene Chromosome Location
Chr.1: 17205123-17246006 on Build GRCh38
Transcripts
NM_013358.2 XM_011541307.1 XM_017001101.1 XM_017001102.1 XM_017001103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6736831 c.483C>T p.S161S Chr.1: 17225885-17225885 on Build GRCh38
6406994 c.418C>A p.R140S Chr.1: 17225820-17225820 on Build GRCh38
5672460 c.466C>A p.R156R Chr.1: 17225868-17225868 on Build GRCh38
3789124 c.513G>A p.L171L Chr.1: 17225915-17225915 on Build GRCh38
6059430 c.433G>T p.G145C Chr.1: 17225835-17225835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939319431 Chr.1: 17225949-17225949 on Build GRCh38
rs747857996 Chr.1: 17225948-17225948 on Build GRCh38
rs865903812 Chr.1: 17225880-17225880 on Build GRCh38
rs759762014 Chr.1: 17225882-17225882 on Build GRCh38
rs769571573 Chr.1: 17225855-17225855 on Build GRCh38
rs962302918 Chr.1: 17225772-17225772 on Build GRCh38
rs756434267 Chr.1: 17225798-17225816 on Build GRCh38
rs1006865514 Chr.1: 17225762-17225762 on Build GRCh38
rs374890157 Chr.1: 17225886-17225886 on Build GRCh38
rs767988224 Chr.1: 17225809-17225809 on Build GRCh38
rs753913972 Chr.1: 17225965-17225965 on Build GRCh38
rs985934556 Chr.1: 17225828-17225828 on Build GRCh38
rs756516910 Chr.1: 17225820-17225820 on Build GRCh38
rs746655728 Chr.1: 17225836-17225836 on Build GRCh38
rs367908904 Chr.1: 17225869-17225869 on Build GRCh38
rs546470322 Chr.1: 17225839-17225839 on Build GRCh38
rs924496090 Chr.1: 17225898-17225898 on Build GRCh38
rs753107043 Chr.1: 17225810-17225810 on Build GRCh38
rs141463441 Chr.1: 17225792-17225792 on Build GRCh38
rs200170680 Chr.1: 17225941-17225941 on Build GRCh38
rs749232891 Chr.1: 17225965-17225965 on Build GRCh38
rs780290399 Chr.1: 17225867-17225868 on Build GRCh38
rs144406233 Chr.1: 17225928-17225928 on Build GRCh38
rs376832336 Chr.1: 17225960-17225960 on Build GRCh38
rs878932730 Chr.1: 17225891-17225891 on Build GRCh38
rs757876298 Chr.1: 17225827-17225827 on Build GRCh38
rs372248039 Chr.1: 17225773-17225773 on Build GRCh38
rs747989856 Chr.1: 17225853-17225853 on Build GRCh38
rs934579433 Chr.1: 17225914-17225914 on Build GRCh38
rs772610303 Chr.1: 17225795-17225795 on Build GRCh38
rs144288276 Chr.1: 17225821-17225821 on Build GRCh38
rs754350072 Chr.1: 17225931-17225931 on Build GRCh38
rs768422769 Chr.1: 17225763-17225763 on Build GRCh38
rs755308786 Chr.1: 17225798-17225798 on Build GRCh38
rs745326779 Chr.1: 17225824-17225824 on Build GRCh38
rs779590846 Chr.1: 17225835-17225835 on Build GRCh38
rs369250178 Chr.1: 17225936-17225936 on Build GRCh38
rs765226406 Chr.1: 17225783-17225783 on Build GRCh38
rs757823246 Chr.1: 17225934-17225934 on Build GRCh38
rs773110007 Chr.1: 17225785-17225785 on Build GRCh38
rs776223277 Chr.1: 17225844-17225844 on Build GRCh38
rs763031946 Chr.1: 17225868-17225868 on Build GRCh38
rs761128662 Chr.1: 17225920-17225920 on Build GRCh38
rs776551290 Chr.1: 17225771-17225771 on Build GRCh38
rs373426172 Chr.1: 17225943-17225943 on Build GRCh38
rs528441261 Chr.1: 17225957-17225957 on Build GRCh38
rs372107973 Chr.1: 17225877-17225877 on Build GRCh38
rs752953244 Chr.1: 17225901-17225901 on Build GRCh38
rs773234819 Chr.1: 17225858-17225858 on Build GRCh38

View Full Product Details