Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 5221172-5221441 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100130950
Entrez Gene ID
100130950
Gene Name
uncharacterized LOC100130950
Gene Aliases
-
UniGene
Hs.668646
Gene Chromosome Location
Chr.17: 5192084-5235636 on Build GRCh38
Transcripts
NR_034082.1 NM_001271842.1 NM_001319190.1 NM_207103.3 XM_017024615.1

Gene Symbol
SCIMP
Entrez Gene ID
388325
Gene Name
SLP adaptor and CSK interacting membrane protein
Gene Aliases
C17orf87, UNQ5783
UniGene
Hs.462080
Gene Chromosome Location
Chr.17: 5208920-5234860 on Build GRCh38
Transcripts
NR_034082.1 NM_001271842.1 NM_001319190.1 NM_207103.3 XM_017024615.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6772690 c.129G>A p.K43K Chr.17: 5221346-5221346 on Build GRCh38
6772689 c.150G>A p.K50K Chr.17: 5221346-5221346 on Build GRCh38
7003195 c.197delA p.K67fs*25 Chr.17: 5221299-5221299 on Build GRCh38
7003196 c.176delA p.K60fs*25 Chr.17: 5221299-5221299 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370629176 Chr.17: 5221384-5221384 on Build GRCh38
rs748147788 Chr.17: 5221400-5221401 on Build GRCh38
rs761381411 Chr.17: 5221360-5221360 on Build GRCh38
rs749187772 Chr.17: 5221221-5221221 on Build GRCh38
rs765775837 Chr.17: 5221254-5221254 on Build GRCh38
rs1031675740 Chr.17: 5221372-5221372 on Build GRCh38
rs775183995 Chr.17: 5221304-5221304 on Build GRCh38
rs377191874 Chr.17: 5221323-5221323 on Build GRCh38
rs768714355 Chr.17: 5221216-5221216 on Build GRCh38
rs757741375 Chr.17: 5221253-5221253 on Build GRCh38
rs775154711 Chr.17: 5221235-5221235 on Build GRCh38
rs753371950 Chr.17: 5221246-5221246 on Build GRCh38
rs759438575 Chr.17: 5221230-5221230 on Build GRCh38
rs192594848 Chr.17: 5221276-5221276 on Build GRCh38
rs747213992 Chr.17: 5221268-5221268 on Build GRCh38
rs372732996 Chr.17: 5221295-5221295 on Build GRCh38
rs766714843 Chr.17: 5221368-5221368 on Build GRCh38
rs552635465 Chr.17: 5221307-5221307 on Build GRCh38
rs936667297 Chr.17: 5221413-5221413 on Build GRCh38
rs750748726 Chr.17: 5221255-5221255 on Build GRCh38
rs1016938266 Chr.17: 5221218-5221218 on Build GRCh38
rs747208413 Chr.17: 5221210-5221210 on Build GRCh38
rs760254370 Chr.17: 5221239-5221239 on Build GRCh38
rs774254532 Chr.17: 5221229-5221229 on Build GRCh38
rs750874732 Chr.17: 5221364-5221364 on Build GRCh38
rs776880853 Chr.17: 5221219-5221219 on Build GRCh38
rs774483522 Chr.17: 5221278-5221278 on Build GRCh38
rs760658915 Chr.17: 5221231-5221231 on Build GRCh38
rs74400579 Chr.17: 5221277-5221277 on Build GRCh38
rs771037880 Chr.17: 5221223-5221223 on Build GRCh38
rs765720585 Chr.17: 5221361-5221361 on Build GRCh38
rs776089064 Chr.17: 5221350-5221350 on Build GRCh38
rs930440030 Chr.17: 5221211-5221211 on Build GRCh38
rs781037048 Chr.17: 5221314-5221314 on Build GRCh38
rs745802677 Chr.17: 5221286-5221286 on Build GRCh38
rs560025272 Chr.17: 5221377-5221377 on Build GRCh38
rs1011981110 Chr.17: 5221291-5221291 on Build GRCh38
rs374572309 Chr.17: 5221270-5221270 on Build GRCh38
rs187604605 Chr.17: 5221265-5221265 on Build GRCh38
rs369901318 Chr.17: 5221264-5221264 on Build GRCh38
rs764029497 Chr.17: 5221272-5221272 on Build GRCh38
rs530080808 Chr.17: 5221243-5221243 on Build GRCh38
rs780285760 Chr.17: 5221266-5221266 on Build GRCh38

View Full Product Details