Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140856241-140856494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCDHA1
Entrez Gene ID
56147
Gene Name
protocadherin alpha 1
Gene Aliases
PCDH-ALPHA1
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140786136-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA10
Entrez Gene ID
56139
Gene Name
protocadherin alpha 10
Gene Aliases
CNR8, CNRN8, CNRS8, CRNR8, PCDH-ALPHA10
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140855883-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA2
Entrez Gene ID
56146
Gene Name
protocadherin alpha 2
Gene Aliases
PCDH-ALPHA2
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140794859-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA3
Entrez Gene ID
56145
Gene Name
protocadherin alpha 3
Gene Aliases
PCDH-ALPHA3
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140801198-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA4
Entrez Gene ID
56144
Gene Name
protocadherin alpha 4
Gene Aliases
CNR1, CNRN1, CRNR1, PCDH-ALPHA4
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140807074-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA5
Entrez Gene ID
56143
Gene Name
protocadherin alpha 5
Gene Aliases
CNR6, CNRN6, CNRS6, CRNR6, PCDH-ALPHA5
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140821776-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA6
Entrez Gene ID
56142
Gene Name
protocadherin alpha 6
Gene Aliases
CNR2, CNRN2, CNRS2, CRNR2, PCDH-ALPHA6
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140827922-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA7
Entrez Gene ID
56141
Gene Name
protocadherin alpha 7
Gene Aliases
CNR4, CNRN4, CNRS4, CRNR4, PCDH-ALPHA7
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140834384-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA8
Entrez Gene ID
56140
Gene Name
protocadherin alpha 8
Gene Aliases
PCDH-ALPHA8
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140841322-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Gene Symbol
PCDHA9
Entrez Gene ID
9752
Gene Name
protocadherin alpha 9
Gene Aliases
PCDH-ALPHA9
UniGene
Hs.199343
Gene Chromosome Location
Chr.5: 140847772-141012344 on Build GRCh38
Transcripts
NM_018900.3 NM_031411.2 NM_018901.3 NM_031859.2 NM_031860.2 NM_018905.2 NM_018906.2 NM_018907.3 NM_018908.2 NM_018909.3 NM_031849.2 NM_018910.2 NM_018911.2 NM_031857.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5545549 c.395T>C p.F132S Chr.5: 140856443-140856443 on Build GRCh38
125179 c.310C>A p.H104N Chr.5: 140856358-140856358 on Build GRCh38
5966476 c.357C>G p.D119E Chr.5: 140856405-140856405 on Build GRCh38
4425680 c.291G>C p.R97R Chr.5: 140856339-140856339 on Build GRCh38
3240752 c.285C>T p.C95C Chr.5: 140856333-140856333 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782173275 Chr.5: 140856431-140856431 on Build GRCh38
rs782670614 Chr.5: 140856428-140856428 on Build GRCh38
rs544932528 Chr.5: 140856321-140856321 on Build GRCh38
rs782323132 Chr.5: 140856305-140856305 on Build GRCh38
rs782430101 Chr.5: 140856372-140856372 on Build GRCh38
rs782635462 Chr.5: 140856281-140856281 on Build GRCh38
rs782325790 Chr.5: 140856295-140856295 on Build GRCh38
rs782418117 Chr.5: 140856432-140856432 on Build GRCh38
rs200204071 Chr.5: 140856318-140856318 on Build GRCh38
rs782584293 Chr.5: 140856341-140856341 on Build GRCh38
rs782447462 Chr.5: 140856403-140856403 on Build GRCh38
rs200722492 Chr.5: 140856425-140856425 on Build GRCh38
rs782568790 Chr.5: 140856315-140856315 on Build GRCh38
rs782335067 Chr.5: 140856344-140856344 on Build GRCh38
rs782392908 Chr.5: 140856448-140856448 on Build GRCh38
rs202075661 Chr.5: 140856384-140856384 on Build GRCh38
rs1053203263 Chr.5: 140856383-140856383 on Build GRCh38
rs782782390 Chr.5: 140856337-140856337 on Build GRCh38
rs190288099 Chr.5: 140856385-140856385 on Build GRCh38
rs199611545 Chr.5: 140856347-140856347 on Build GRCh38
rs781846770 Chr.5: 140856323-140856323 on Build GRCh38
rs782472888 Chr.5: 140856271-140856271 on Build GRCh38
rs782279735 Chr.5: 140856463-140856463 on Build GRCh38
rs782180138 Chr.5: 140856292-140856292 on Build GRCh38
rs782109045 Chr.5: 140856427-140856427 on Build GRCh38
rs782271561 Chr.5: 140856369-140856369 on Build GRCh38
rs367902357 Chr.5: 140856269-140856269 on Build GRCh38
rs782124565 Chr.5: 140856394-140856394 on Build GRCh38
rs138367057 Chr.5: 140856289-140856289 on Build GRCh38
rs781914523 Chr.5: 140856373-140856373 on Build GRCh38
rs782470045 Chr.5: 140856418-140856418 on Build GRCh38
rs782195215 Chr.5: 140856375-140856375 on Build GRCh38
rs782023286 Chr.5: 140856399-140856399 on Build GRCh38
rs782092511 Chr.5: 140856319-140856319 on Build GRCh38
rs781808272 Chr.5: 140856411-140856411 on Build GRCh38
rs782497272 Chr.5: 140856356-140856356 on Build GRCh38
rs782661319 Chr.5: 140856364-140856364 on Build GRCh38
rs782114957 Chr.5: 140856304-140856304 on Build GRCh38
rs1030293372 Chr.5: 140856468-140856468 on Build GRCh38
rs782709201 Chr.5: 140856406-140856406 on Build GRCh38
rs782595746 Chr.5: 140856286-140856286 on Build GRCh38
rs782776522 Chr.5: 140856397-140856397 on Build GRCh38
rs782438318 Chr.5: 140856471-140856471 on Build GRCh38
rs782445424 Chr.5: 140856260-140856260 on Build GRCh38
rs377709063 Chr.5: 140856426-140856426 on Build GRCh38
rs782387382 Chr.5: 140856310-140856310 on Build GRCh38
rs199720508 Chr.5: 140856413-140856413 on Build GRCh38
rs377602570 Chr.5: 140856297-140856297 on Build GRCh38
rs551569829 Chr.5: 140856434-140856434 on Build GRCh38
rs782383115 Chr.5: 140856448-140856448 on Build GRCh38
rs568319142 Chr.5: 140856374-140856374 on Build GRCh38
rs782523397 Chr.5: 140856336-140856336 on Build GRCh38
rs782164641 Chr.5: 140856401-140856401 on Build GRCh38
rs566691333 Chr.5: 140856459-140856459 on Build GRCh38
rs199529084 Chr.5: 140856259-140856259 on Build GRCh38
rs782582964 Chr.5: 140856261-140856261 on Build GRCh38
rs782213392 Chr.5: 140856451-140856451 on Build GRCh38
rs781903292 Chr.5: 140856338-140856338 on Build GRCh38

View Full Product Details