Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43761337-43761610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDIA3
Entrez Gene ID
2923
Gene Name
protein disulfide isomerase family A member 3
Gene Aliases
ER60, ERp57, ERp60, ERp61, GRP57, GRP58, HEL-S-269, HEL-S-93n, HsT17083, P58, PI-PLC
UniGene
Hs.591095
Gene Chromosome Location
Chr.15: 43746392-43772606 on Build GRCh38
Transcripts
NM_005313.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2188703 c.417C>G p.L139L Chr.15: 43761476-43761476 on Build GRCh38
5425282 c.365-5C>T p.? Chr.15: 43761419-43761419 on Build GRCh38
3501515 c.393G>A p.Q131Q Chr.15: 43761452-43761452 on Build GRCh38
3501516 c.435T>C p.F145F Chr.15: 43761494-43761494 on Build GRCh38
1678420 c.472G>A p.G158S Chr.15: 43761531-43761531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759045484 Chr.15: 43761505-43761505 on Build GRCh38
rs956488291 Chr.15: 43761541-43761541 on Build GRCh38
rs542882802 Chr.15: 43761582-43761582 on Build GRCh38
rs754189358 Chr.15: 43761538-43761538 on Build GRCh38
rs375445252 Chr.15: 43761561-43761561 on Build GRCh38
rs996234681 Chr.15: 43761554-43761554 on Build GRCh38
rs769559002 Chr.15: 43761472-43761472 on Build GRCh38
rs768073813 Chr.15: 43761368-43761368 on Build GRCh38
rs543412569 Chr.15: 43761449-43761449 on Build GRCh38
rs542820973 Chr.15: 43761522-43761522 on Build GRCh38
rs758475479 Chr.15: 43761551-43761551 on Build GRCh38
rs8040336 Chr.15: 43761419-43761419 on Build GRCh38
rs748068214 Chr.15: 43761558-43761558 on Build GRCh38
rs887811660 Chr.15: 43761574-43761574 on Build GRCh38
rs750646071 Chr.15: 43761547-43761547 on Build GRCh38
rs55968494 Chr.15: 43761504-43761504 on Build GRCh38
rs557451225 Chr.15: 43761390-43761390 on Build GRCh38
rs139576892 Chr.15: 43761542-43761542 on Build GRCh38
rs777697034 Chr.15: 43761563-43761563 on Build GRCh38
rs144040692 Chr.15: 43761585-43761585 on Build GRCh38
rs746541222 Chr.15: 43761549-43761549 on Build GRCh38
rs201421563 Chr.15: 43761464-43761464 on Build GRCh38
rs765707220 Chr.15: 43761404-43761404 on Build GRCh38
rs1054408341 Chr.15: 43761514-43761514 on Build GRCh38
rs147342240 Chr.15: 43761500-43761500 on Build GRCh38
rs761321604 Chr.15: 43761527-43761527 on Build GRCh38
rs757621327 Chr.15: 43761432-43761432 on Build GRCh38
rs749083049 Chr.15: 43761486-43761486 on Build GRCh38
rs760004755 Chr.15: 43761553-43761553 on Build GRCh38
rs765451171 Chr.15: 43761546-43761546 on Build GRCh38
rs773053423 Chr.15: 43761474-43761474 on Build GRCh38
rs780050794 Chr.15: 43761552-43761552 on Build GRCh38
rs760789393 Chr.15: 43761436-43761436 on Build GRCh38
rs540688699 Chr.15: 43761377-43761377 on Build GRCh38
rs757678736 Chr.15: 43761374-43761375 on Build GRCh38
rs543485322 Chr.15: 43761525-43761525 on Build GRCh38
rs367690999 Chr.15: 43761382-43761382 on Build GRCh38
rs745905098 Chr.15: 43761461-43761461 on Build GRCh38
rs766974612 Chr.15: 43761521-43761521 on Build GRCh38
rs764550382 Chr.15: 43761534-43761534 on Build GRCh38
rs372871422 Chr.15: 43761468-43761468 on Build GRCh38
rs185878833 Chr.15: 43761467-43761467 on Build GRCh38

View Full Product Details