Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65538311-65538572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LTBP3
Entrez Gene ID
4054
Gene Name
latent transforming growth factor beta binding protein 3
Gene Aliases
DASS, LTBP-3, LTBP2, STHAG6, pp6425
UniGene
Hs.289019
Gene Chromosome Location
Chr.11: 65538559-65558388 on Build GRCh38
Transcripts
NM_001130144.2 NM_001164266.1 NM_021070.4 XM_011545032.2 XM_011545033.2 NM_001048218.1 NM_020680.3 XM_005274118.3 XM_005274120.3 XM_005274121.3 XM_017018029.1 XM_017018030.1 XM_017018031.1 XM_017018032.1 XM_017018033.1

Gene Symbol
SCYL1
Entrez Gene ID
57410
Gene Name
SCY1 like pseudokinase 1
Gene Aliases
GKLP, HT019, NKTL, NTKL, P105, SCAR21, TAPK, TEIF, TRAP
UniGene
Hs.238839
Gene Chromosome Location
Chr.11: 65525065-65538711 on Build GRCh38
Transcripts
NM_001130144.2 NM_001164266.1 NM_021070.4 XM_011545032.2 XM_011545033.2 NM_001048218.1 NM_020680.3 XM_005274118.3 XM_005274120.3 XM_005274121.3 XM_017018029.1 XM_017018030.1 XM_017018031.1 XM_017018032.1 XM_017018033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3451817 c.2397C>T p.P799P Chr.11: 65538536-65538536 on Build GRCh38
5352085 c.2400G>T p.M800I Chr.11: 65538539-65538539 on Build GRCh38
6773578 c.2407G>T p.G803* Chr.11: 65538546-65538546 on Build GRCh38
4645792 c.2337C>T p.R779R Chr.11: 65538476-65538476 on Build GRCh38
6566857 c.2302+5G>A p.? Chr.11: 65538329-65538329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764451963 Chr.11: 65538504-65538504 on Build GRCh38
rs200704028 Chr.11: 65538497-65538497 on Build GRCh38
rs749493894 Chr.11: 65538465-65538465 on Build GRCh38
rs1025230766 Chr.11: 65538539-65538539 on Build GRCh38
rs769900982 Chr.11: 65538448-65538448 on Build GRCh38
rs773092805 Chr.11: 65538474-65538474 on Build GRCh38
rs1784221 Chr.11: 65538397-65538397 on Build GRCh38
rs753340940 Chr.11: 65538358-65538358 on Build GRCh38
rs780141252 Chr.11: 65538530-65538530 on Build GRCh38
rs899960373 Chr.11: 65538454-65538454 on Build GRCh38
rs768942150 Chr.11: 65538537-65538537 on Build GRCh38
rs752383759 Chr.11: 65538387-65538387 on Build GRCh38
rs757698806 Chr.11: 65538409-65538409 on Build GRCh38
rs764785199 Chr.11: 65538494-65538494 on Build GRCh38
rs899628558 Chr.11: 65538492-65538492 on Build GRCh38
rs754666116 Chr.11: 65538374-65538374 on Build GRCh38
rs556081911 Chr.11: 65538469-65538469 on Build GRCh38
rs908835036 Chr.11: 65538392-65538392 on Build GRCh38
rs766354334 Chr.11: 65538484-65538484 on Build GRCh38
rs373512455 Chr.11: 65538375-65538375 on Build GRCh38
rs1013420547 Chr.11: 65538536-65538536 on Build GRCh38
rs376500168 Chr.11: 65538521-65538521 on Build GRCh38
rs370701660 Chr.11: 65538466-65538466 on Build GRCh38
rs1050154207 Chr.11: 65538490-65538490 on Build GRCh38
rs781250002 Chr.11: 65538419-65538419 on Build GRCh38
rs764128856 Chr.11: 65538356-65538357 on Build GRCh38
rs200041750 Chr.11: 65538493-65538493 on Build GRCh38
rs746011430 Chr.11: 65538424-65538424 on Build GRCh38
rs751582948 Chr.11: 65538515-65538522 on Build GRCh38
rs573462705 Chr.11: 65538462-65538462 on Build GRCh38
rs759029574 Chr.11: 65538489-65538489 on Build GRCh38
rs773146247 Chr.11: 65538549-65538549 on Build GRCh38
rs533603138 Chr.11: 65538391-65538391 on Build GRCh38
rs1037738916 Chr.11: 65538382-65538382 on Build GRCh38
rs369741245 Chr.11: 65538372-65538372 on Build GRCh38
rs564277938 Chr.11: 65538509-65538509 on Build GRCh38
rs370774721 Chr.11: 65538527-65538527 on Build GRCh38
rs766010966 Chr.11: 65538340-65538340 on Build GRCh38
rs781704984 Chr.11: 65538512-65538512 on Build GRCh38
rs373594681 Chr.11: 65538533-65538533 on Build GRCh38
rs775621894 Chr.11: 65538461-65538461 on Build GRCh38
rs758033326 Chr.11: 65538500-65538500 on Build GRCh38
rs757610725 Chr.11: 65538510-65538510 on Build GRCh38
rs575977540 Chr.11: 65538477-65538477 on Build GRCh38
rs746869284 Chr.11: 65538553-65538553 on Build GRCh38
rs776706991 Chr.11: 65538488-65538488 on Build GRCh38
rs747549292 Chr.11: 65538407-65538407 on Build GRCh38

View Full Product Details