Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50278110-50278379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLXNB2
Entrez Gene ID
23654
Gene Name
plexin B2
Gene Aliases
MM1, Nbla00445, PLEXB2, dJ402G11.3
UniGene
Hs.3989
Gene Chromosome Location
Chr.22: 50274979-50307637 on Build GRCh38
Transcripts
NM_012401.3 XM_005261909.1 XM_005261910.2 XM_005261911.4 XM_006724413.2 XM_011530682.2 XM_017028703.1 XM_017028704.1

SNP Detail

SNP ID Chromosome Location
rs754300840 Chr.22: 50278150-50278150 on Build GRCh38
rs1051626606 Chr.22: 50278307-50278307 on Build GRCh38
rs780823777 Chr.22: 50278296-50278296 on Build GRCh38
rs752759021 Chr.22: 50278296-50278296 on Build GRCh38
rs541570546 Chr.22: 50278163-50278163 on Build GRCh38
rs766865347 Chr.22: 50278283-50278283 on Build GRCh38
rs370400977 Chr.22: 50278207-50278207 on Build GRCh38
rs935955410 Chr.22: 50278261-50278261 on Build GRCh38
rs374439102 Chr.22: 50278282-50278282 on Build GRCh38
rs200430810 Chr.22: 50278219-50278219 on Build GRCh38
rs759231441 Chr.22: 50278269-50278269 on Build GRCh38
rs768203410 Chr.22: 50278226-50278226 on Build GRCh38
rs371906643 Chr.22: 50278297-50278297 on Build GRCh38
rs775062503 Chr.22: 50278222-50278222 on Build GRCh38
rs771784244 Chr.22: 50278313-50278313 on Build GRCh38
rs763521580 Chr.22: 50278147-50278147 on Build GRCh38
rs201644992 Chr.22: 50278183-50278183 on Build GRCh38
rs776067354 Chr.22: 50278244-50278244 on Build GRCh38
rs199645325 Chr.22: 50278177-50278177 on Build GRCh38
rs986307397 Chr.22: 50278253-50278253 on Build GRCh38
rs34446050 Chr.22: 50278300-50278301 on Build GRCh38
rs1004686537 Chr.22: 50278342-50278342 on Build GRCh38
rs368875408 Chr.22: 50278290-50278290 on Build GRCh38
rs751765896 Chr.22: 50278285-50278285 on Build GRCh38
rs770516365 Chr.22: 50278166-50278166 on Build GRCh38
rs760320759 Chr.22: 50278223-50278223 on Build GRCh38
rs779887492 Chr.22: 50278315-50278315 on Build GRCh38
rs765707742 Chr.22: 50278260-50278260 on Build GRCh38
rs778771087 Chr.22: 50278299-50278299 on Build GRCh38
rs375683936 Chr.22: 50278201-50278201 on Build GRCh38
rs781136241 Chr.22: 50278190-50278190 on Build GRCh38
rs765625516 Chr.22: 50278162-50278162 on Build GRCh38
rs762362721 Chr.22: 50278245-50278245 on Build GRCh38
rs745816119 Chr.22: 50278306-50278306 on Build GRCh38
rs762298147 Chr.22: 50278158-50278158 on Build GRCh38
rs574952389 Chr.22: 50278182-50278182 on Build GRCh38
rs34660958 Chr.22: 50278331-50278332 on Build GRCh38
rs991246172 Chr.22: 50278203-50278203 on Build GRCh38
rs966470315 Chr.22: 50278187-50278187 on Build GRCh38
rs570065916 Chr.22: 50278135-50278135 on Build GRCh38
rs756845977 Chr.22: 50278286-50278287 on Build GRCh38
rs933510618 Chr.22: 50278243-50278243 on Build GRCh38
rs564490648 Chr.22: 50278321-50278321 on Build GRCh38
rs775152335 Chr.22: 50278131-50278131 on Build GRCh38
rs749235325 Chr.22: 50278192-50278192 on Build GRCh38
rs746663764 Chr.22: 50278319-50278319 on Build GRCh38
rs778646635 Chr.22: 50278216-50278216 on Build GRCh38
rs879151685 Chr.22: 50278221-50278221 on Build GRCh38
rs528812002 Chr.22: 50278337-50278337 on Build GRCh38
rs942311329 Chr.22: 50278292-50278292 on Build GRCh38

View Full Product Details