Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39313995-39314240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK17
Entrez Gene ID
89822
Gene Name
potassium two pore domain channel subfamily K member 17
Gene Aliases
K2p17.1, TALK-2, TALK2, TASK-4, TASK4
UniGene
Hs.162282
Gene Chromosome Location
Chr.6: 39299001-39314461 on Build GRCh38
Transcripts
NM_001135111.1 NM_031460.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6359690 c.233T>A p.I78N Chr.6: 39314088-39314088 on Build GRCh38
6691307 c.215C>T p.P72L Chr.6: 39314106-39314106 on Build GRCh38
6391759 c.220C>A p.L74M Chr.6: 39314101-39314101 on Build GRCh38
5787070 c.81C>T p.L27L Chr.6: 39314240-39314240 on Build GRCh38
6359691 c.233T>A p.I78N Chr.6: 39314088-39314088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769190726 Chr.6: 39314078-39314078 on Build GRCh38
rs1021970270 Chr.6: 39314059-39314059 on Build GRCh38
rs753996296 Chr.6: 39314042-39314042 on Build GRCh38
rs575249030 Chr.6: 39314209-39314209 on Build GRCh38
rs774012245 Chr.6: 39314183-39314183 on Build GRCh38
rs906031976 Chr.6: 39314029-39314029 on Build GRCh38
rs762943020 Chr.6: 39314145-39314145 on Build GRCh38
rs201418585 Chr.6: 39314131-39314131 on Build GRCh38
rs956253428 Chr.6: 39314045-39314045 on Build GRCh38
rs746939056 Chr.6: 39314072-39314072 on Build GRCh38
rs745806536 Chr.6: 39314151-39314151 on Build GRCh38
rs540036217 Chr.6: 39314076-39314076 on Build GRCh38
rs760990793 Chr.6: 39314186-39314186 on Build GRCh38
rs1056913670 Chr.6: 39314026-39314026 on Build GRCh38
rs373663901 Chr.6: 39314127-39314127 on Build GRCh38
rs748326556 Chr.6: 39314055-39314055 on Build GRCh38
rs765250531 Chr.6: 39314121-39314121 on Build GRCh38
rs897918400 Chr.6: 39314041-39314041 on Build GRCh38
rs375910334 Chr.6: 39314079-39314079 on Build GRCh38
rs192466468 Chr.6: 39314173-39314173 on Build GRCh38
rs755021136 Chr.6: 39314110-39314110 on Build GRCh38
rs1002973113 Chr.6: 39314206-39314206 on Build GRCh38
rs1040806698 Chr.6: 39314160-39314173 on Build GRCh38
rs779494063 Chr.6: 39314163-39314163 on Build GRCh38
rs771293542 Chr.6: 39314187-39314187 on Build GRCh38
rs761146240 Chr.6: 39314085-39314085 on Build GRCh38
rs28422024 Chr.6: 39314016-39314016 on Build GRCh38
rs368194171 Chr.6: 39314165-39314165 on Build GRCh38
rs766819933 Chr.6: 39314098-39314098 on Build GRCh38
rs755038296 Chr.6: 39314047-39314047 on Build GRCh38
rs201819169 Chr.6: 39314146-39314146 on Build GRCh38
rs61748648 Chr.6: 39314140-39314140 on Build GRCh38
rs771033826 Chr.6: 39314074-39314074 on Build GRCh38
rs909535328 Chr.6: 39314147-39314147 on Build GRCh38
rs766412671 Chr.6: 39314035-39314035 on Build GRCh38
rs765231421 Chr.6: 39314086-39314086 on Build GRCh38
rs762570124 Chr.6: 39314080-39314080 on Build GRCh38
rs781115980 Chr.6: 39314142-39314142 on Build GRCh38
rs1002941876 Chr.6: 39314037-39314037 on Build GRCh38
rs372753787 Chr.6: 39314052-39314052 on Build GRCh38
rs542153673 Chr.6: 39314158-39314158 on Build GRCh38
rs773603729 Chr.6: 39314081-39314081 on Build GRCh38
rs370525658 Chr.6: 39314115-39314115 on Build GRCh38
rs757147464 Chr.6: 39314141-39314141 on Build GRCh38
rs754336373 Chr.6: 39314106-39314106 on Build GRCh38
rs370961739 Chr.6: 39314138-39314138 on Build GRCh38
rs201082190 Chr.6: 39314216-39314216 on Build GRCh38

View Full Product Details