Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 38851424-38851695 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRAGC
Entrez Gene ID
64121
Gene Name
Ras related GTP binding C
Gene Aliases
GTR2, RAGC, TIB929
UniGene
Hs.532461
Gene Chromosome Location
Chr.1: 38838197-38859823 on Build GRCh38
Transcripts
NM_001271851.1 NM_022157.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6769752 c.868G>A p.D290N Chr.1: 38851646-38851646 on Build GRCh38
291355 c.867C>T p.I289I Chr.1: 38851647-38851647 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754620928 Chr.1: 38851603-38851603 on Build GRCh38
rs754595121 Chr.1: 38851636-38851647 on Build GRCh38
rs907606412 Chr.1: 38851460-38851460 on Build GRCh38
rs201785970 Chr.1: 38851575-38851575 on Build GRCh38
rs891862421 Chr.1: 38851550-38851550 on Build GRCh38
rs1055429876 Chr.1: 38851474-38851474 on Build GRCh38
rs374780565 Chr.1: 38851519-38851519 on Build GRCh38
rs766939467 Chr.1: 38851466-38851466 on Build GRCh38
rs200140491 Chr.1: 38851576-38851576 on Build GRCh38
rs1011435182 Chr.1: 38851555-38851555 on Build GRCh38
rs746683580 Chr.1: 38851596-38851596 on Build GRCh38
rs752888855 Chr.1: 38851568-38851568 on Build GRCh38
rs376828760 Chr.1: 38851605-38851605 on Build GRCh38
rs957990348 Chr.1: 38851559-38851559 on Build GRCh38
rs750858130 Chr.1: 38851580-38851580 on Build GRCh38
rs983258547 Chr.1: 38851454-38851454 on Build GRCh38
rs1000113969 Chr.1: 38851542-38851542 on Build GRCh38
rs764907659 Chr.1: 38851647-38851647 on Build GRCh38
rs191578481 Chr.1: 38851482-38851482 on Build GRCh38
rs184487976 Chr.1: 38851495-38851495 on Build GRCh38
rs918762975 Chr.1: 38851628-38851628 on Build GRCh38
rs760849741 Chr.1: 38851570-38851570 on Build GRCh38
rs747607019 Chr.1: 38851620-38851620 on Build GRCh38
rs376349580 Chr.1: 38851564-38851564 on Build GRCh38
rs72925115 Chr.1: 38851491-38851491 on Build GRCh38
rs1030249362 Chr.1: 38851584-38851584 on Build GRCh38
rs1053242696 Chr.1: 38851516-38851516 on Build GRCh38
rs776174880 Chr.1: 38851501-38851501 on Build GRCh38
rs769831491 Chr.1: 38851641-38851641 on Build GRCh38
rs764254581 Chr.1: 38851571-38851571 on Build GRCh38
rs779890450 Chr.1: 38851590-38851590 on Build GRCh38
rs974629330 Chr.1: 38851471-38851471 on Build GRCh38
rs552206172 Chr.1: 38851510-38851510 on Build GRCh38
rs758812628 Chr.1: 38851588-38851588 on Build GRCh38
rs142273262 Chr.1: 38851674-38851674 on Build GRCh38

View Full Product Details