Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 68709904-68710149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUP107
Entrez Gene ID
57122
Gene Name
nucleoporin 107
Gene Aliases
NPHS11, NUP84
UniGene
Hs.524574
Gene Chromosome Location
Chr.12: 68686951-68742693 on Build GRCh38
Transcripts
NM_020401.2 XM_005269037.3 XM_011538576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6531589 c.847G>C p.E283Q Chr.12: 68710050-68710050 on Build GRCh38
297602 c.839A>G p.E280G Chr.12: 68710042-68710042 on Build GRCh38
6872102 c.820G>T p.E274* Chr.12: 68710023-68710023 on Build GRCh38
4708682 c.835G>T p.D279Y Chr.12: 68710038-68710038 on Build GRCh38
431743 c.847G>A p.E283K Chr.12: 68710050-68710050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771125885 Chr.12: 68710046-68710046 on Build GRCh38
rs774598891 Chr.12: 68709985-68709985 on Build GRCh38
rs767954561 Chr.12: 68710076-68710076 on Build GRCh38
rs377008894 Chr.12: 68710023-68710023 on Build GRCh38
rs764485425 Chr.12: 68710107-68710107 on Build GRCh38
rs376134696 Chr.12: 68709962-68709962 on Build GRCh38
rs371158543 Chr.12: 68710051-68710051 on Build GRCh38
rs369203710 Chr.12: 68709976-68709976 on Build GRCh38
rs750133198 Chr.12: 68710048-68710048 on Build GRCh38
rs773458740 Chr.12: 68710008-68710008 on Build GRCh38
rs142546393 Chr.12: 68710084-68710084 on Build GRCh38
rs750113158 Chr.12: 68710117-68710117 on Build GRCh38
rs1040596390 Chr.12: 68709940-68709940 on Build GRCh38
rs745961066 Chr.12: 68709989-68709992 on Build GRCh38
rs375091871 Chr.12: 68710088-68710088 on Build GRCh38
rs1033126311 Chr.12: 68710115-68710115 on Build GRCh38
rs772138741 Chr.12: 68710006-68710016 on Build GRCh38
rs776157918 Chr.12: 68710078-68710079 on Build GRCh38
rs769585054 Chr.12: 68709985-68709985 on Build GRCh38
rs781155155 Chr.12: 68709972-68709972 on Build GRCh38
rs201316282 Chr.12: 68709959-68709959 on Build GRCh38

View Full Product Details