Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62579941-62580214 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO11
Entrez Gene ID
51194
Gene Name
importin 11
Gene Aliases
RanBP11
UniGene
Hs.482269
Gene Chromosome Location
Chr.5: 62412746-62628589 on Build GRCh38
Transcripts
NM_001134779.1 NM_016338.4 NR_073584.1 NM_181506.4

Gene Symbol
IPO11-LRRC70
Entrez Gene ID
101180901
Gene Name
IPO11-LRRC70 readthrough
Gene Aliases
-
UniGene
Hs.482269
Gene Chromosome Location
Chr.5: 62578735-62628589 on Build GRCh38
Transcripts
NM_001134779.1 NM_016338.4 NR_073584.1 NM_181506.4

Gene Symbol
LRRC70
Entrez Gene ID
100130733
Gene Name
leucine rich repeat containing 70
Gene Aliases
SLRN
UniGene
Hs.721348
Gene Chromosome Location
Chr.5: 62578735-62581448 on Build GRCh38
Transcripts
NM_001134779.1 NM_016338.4 NR_073584.1 NM_181506.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5975647 c.774C>T p.L258L Chr.5: 62580212-62580212 on Build GRCh38
3828268 c.607C>T p.L203F Chr.5: 62580045-62580045 on Build GRCh38
5946738 c.676G>A p.V226I Chr.5: 62580114-62580114 on Build GRCh38
4647113 c.678A>G p.V226V Chr.5: 62580116-62580116 on Build GRCh38
5946739 c.676G>A p.V226I Chr.5: 62580114-62580114 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756244618 Chr.5: 62580170-62580170 on Build GRCh38
rs903690690 Chr.5: 62580062-62580062 on Build GRCh38
rs762249819 Chr.5: 62579991-62579991 on Build GRCh38
rs766030459 Chr.5: 62580100-62580101 on Build GRCh38
rs946129717 Chr.5: 62580026-62580026 on Build GRCh38
rs774910601 Chr.5: 62579986-62579986 on Build GRCh38
rs140074399 Chr.5: 62580058-62580058 on Build GRCh38
rs767704690 Chr.5: 62579992-62579992 on Build GRCh38
rs781725917 Chr.5: 62580116-62580116 on Build GRCh38
rs748549308 Chr.5: 62580147-62580147 on Build GRCh38
rs753560236 Chr.5: 62580007-62580007 on Build GRCh38
rs752175755 Chr.5: 62580088-62580088 on Build GRCh38
rs769473235 Chr.5: 62579974-62579975 on Build GRCh38
rs777674785 Chr.5: 62580181-62580181 on Build GRCh38
rs772975524 Chr.5: 62580085-62580086 on Build GRCh38
rs769277422 Chr.5: 62579975-62579975 on Build GRCh38
rs184754326 Chr.5: 62580016-62580016 on Build GRCh38
rs762492173 Chr.5: 62580093-62580097 on Build GRCh38
rs561159484 Chr.5: 62580123-62580123 on Build GRCh38
rs981914426 Chr.5: 62580169-62580169 on Build GRCh38
rs199809934 Chr.5: 62580076-62580076 on Build GRCh38
rs972460729 Chr.5: 62579977-62579977 on Build GRCh38
rs751909556 Chr.5: 62580012-62580012 on Build GRCh38

View Full Product Details