Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 30876928-30877173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDC3
Entrez Gene ID
9672
Gene Name
syndecan 3
Gene Aliases
SDCN, SYND3
UniGene
Hs.158287
Gene Chromosome Location
Chr.1: 30869466-30909735 on Build GRCh38
Transcripts
NM_014654.3 XM_011542463.1 XM_011542464.2 XM_011542466.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4031333 c.416C>T p.A139V Chr.1: 30877006-30877006 on Build GRCh38
1341705 c.430G>A p.V144M Chr.1: 30876992-30876992 on Build GRCh38
41081 c.307G>A p.V103I Chr.1: 30877115-30877115 on Build GRCh38
7295455 c.399C>T p.R133R Chr.1: 30877023-30877023 on Build GRCh38
4785976 c.256G>A p.V86M Chr.1: 30876992-30876992 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762632276 Chr.1: 30877056-30877056 on Build GRCh38
rs150946427 Chr.1: 30876979-30876979 on Build GRCh38
rs775349919 Chr.1: 30876973-30876973 on Build GRCh38
rs930365684 Chr.1: 30877001-30877001 on Build GRCh38
rs748470737 Chr.1: 30876951-30876951 on Build GRCh38
rs777924866 Chr.1: 30877021-30877021 on Build GRCh38
rs970997426 Chr.1: 30877058-30877058 on Build GRCh38
rs767961193 Chr.1: 30877011-30877011 on Build GRCh38
rs777652983 Chr.1: 30877113-30877113 on Build GRCh38
rs377686596 Chr.1: 30877121-30877121 on Build GRCh38
rs898808921 Chr.1: 30876990-30876990 on Build GRCh38
rs756403371 Chr.1: 30877017-30877017 on Build GRCh38
rs373189918 Chr.1: 30877104-30877104 on Build GRCh38
rs777993332 Chr.1: 30876955-30876955 on Build GRCh38
rs377201191 Chr.1: 30877034-30877034 on Build GRCh38
rs772909989 Chr.1: 30876991-30876991 on Build GRCh38
rs753197860 Chr.1: 30877084-30877084 on Build GRCh38
rs772785365 Chr.1: 30877048-30877048 on Build GRCh38
rs901325573 Chr.1: 30877103-30877103 on Build GRCh38
rs765564861 Chr.1: 30877149-30877149 on Build GRCh38
rs1050005699 Chr.1: 30877117-30877117 on Build GRCh38
rs753072963 Chr.1: 30877014-30877014 on Build GRCh38
rs748984700 Chr.1: 30877115-30877115 on Build GRCh38
rs747965137 Chr.1: 30877035-30877035 on Build GRCh38
rs141463201 Chr.1: 30877047-30877047 on Build GRCh38
rs143651606 Chr.1: 30877086-30877086 on Build GRCh38
rs764498159 Chr.1: 30877087-30877087 on Build GRCh38
rs760065032 Chr.1: 30877003-30877003 on Build GRCh38
rs778957598 Chr.1: 30877097-30877097 on Build GRCh38
rs776025850 Chr.1: 30877129-30877129 on Build GRCh38
rs764469938 Chr.1: 30877132-30877132 on Build GRCh38
rs570645571 Chr.1: 30876986-30876986 on Build GRCh38
rs757534300 Chr.1: 30877095-30877095 on Build GRCh38
rs769513925 Chr.1: 30877112-30877112 on Build GRCh38
rs201563377 Chr.1: 30877024-30877024 on Build GRCh38
rs17851080 Chr.1: 30877089-30877089 on Build GRCh38
rs752098151 Chr.1: 30876999-30876999 on Build GRCh38
rs372685746 Chr.1: 30876976-30876976 on Build GRCh38
rs367599166 Chr.1: 30877083-30877083 on Build GRCh38
rs770448216 Chr.1: 30877062-30877062 on Build GRCh38
rs771039175 Chr.1: 30876963-30876963 on Build GRCh38
rs148395976 Chr.1: 30876954-30876954 on Build GRCh38
rs149166920 Chr.1: 30877130-30877130 on Build GRCh38
rs556133593 Chr.1: 30877105-30877105 on Build GRCh38
rs749333883 Chr.1: 30876956-30876956 on Build GRCh38
rs754049937 Chr.1: 30877092-30877092 on Build GRCh38
rs761656557 Chr.1: 30876982-30876982 on Build GRCh38
rs978127227 Chr.1: 30877124-30877124 on Build GRCh38
rs771557426 Chr.1: 30877125-30877125 on Build GRCh38
rs746922572 Chr.1: 30876975-30876975 on Build GRCh38
rs142879690 Chr.1: 30876948-30876948 on Build GRCh38
rs762424792 Chr.1: 30876994-30876994 on Build GRCh38
rs953054046 Chr.1: 30876989-30876989 on Build GRCh38
rs745905618 Chr.1: 30877030-30877030 on Build GRCh38
rs759112640 Chr.1: 30877122-30877122 on Build GRCh38
rs762028465 Chr.1: 30877138-30877138 on Build GRCh38
rs76411652 Chr.1: 30877114-30877114 on Build GRCh38
rs146793522 Chr.1: 30877072-30877072 on Build GRCh38
rs201140173 Chr.1: 30877150-30877150 on Build GRCh38
rs377656744 Chr.1: 30877025-30877025 on Build GRCh38
rs768739693 Chr.1: 30877033-30877033 on Build GRCh38
rs41269523 Chr.1: 30877136-30877136 on Build GRCh38
rs770628492 Chr.1: 30877118-30877118 on Build GRCh38
rs139509747 Chr.1: 30876998-30876998 on Build GRCh38

View Full Product Details