Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60432254-60432519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A5
Entrez Gene ID
64232
Gene Name
membrane spanning 4-domains A5
Gene Aliases
CD20-L2, CD20L2, TETM4
UniGene
Hs.178066
Gene Chromosome Location
Chr.11: 60429589-60447792 on Build GRCh38
Transcripts
NM_023945.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5804355 c.283-2A>T p.? Chr.11: 60432409-60432409 on Build GRCh38
5644619 c.339G>A p.L113L Chr.11: 60432467-60432467 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs953816483 Chr.11: 60432421-60432421 on Build GRCh38
rs777423185 Chr.11: 60432383-60432383 on Build GRCh38
rs753407365 Chr.11: 60432480-60432480 on Build GRCh38
rs372731011 Chr.11: 60432367-60432367 on Build GRCh38
rs748374772 Chr.11: 60432420-60432420 on Build GRCh38
rs115468185 Chr.11: 60432318-60432318 on Build GRCh38
rs370162997 Chr.11: 60432439-60432439 on Build GRCh38
rs751431872 Chr.11: 60432365-60432365 on Build GRCh38
rs1035194636 Chr.11: 60432355-60432355 on Build GRCh38
rs781486324 Chr.11: 60432419-60432419 on Build GRCh38
rs763143458 Chr.11: 60432435-60432435 on Build GRCh38
rs200264025 Chr.11: 60432363-60432363 on Build GRCh38
rs763863048 Chr.11: 60432472-60432472 on Build GRCh38
rs1036800197 Chr.11: 60432310-60432310 on Build GRCh38
rs1002819637 Chr.11: 60432334-60432334 on Build GRCh38
rs118146335 Chr.11: 60432369-60432369 on Build GRCh38
rs977763599 Chr.11: 60432352-60432352 on Build GRCh38
rs142868297 Chr.11: 60432434-60432434 on Build GRCh38
rs756706663 Chr.11: 60432390-60432390 on Build GRCh38
rs78731820 Chr.11: 60432436-60432436 on Build GRCh38
rs569049729 Chr.11: 60432316-60432316 on Build GRCh38
rs767437800 Chr.11: 60432441-60432441 on Build GRCh38
rs780658252 Chr.11: 60432391-60432391 on Build GRCh38
rs752425411 Chr.11: 60432376-60432376 on Build GRCh38
rs554471877 Chr.11: 60432456-60432456 on Build GRCh38
rs760538367 Chr.11: 60432457-60432457 on Build GRCh38
rs905244797 Chr.11: 60432330-60432330 on Build GRCh38
rs117418499 Chr.11: 60432387-60432387 on Build GRCh38
rs549048250 Chr.11: 60432311-60432311 on Build GRCh38
rs776361292 Chr.11: 60432427-60432427 on Build GRCh38
rs939934099 Chr.11: 60432303-60432303 on Build GRCh38
rs994815158 Chr.11: 60432317-60432317 on Build GRCh38
rs770069338 Chr.11: 60432426-60432426 on Build GRCh38
rs963388299 Chr.11: 60432290-60432290 on Build GRCh38
rs191061257 Chr.11: 60432398-60432398 on Build GRCh38
rs994789586 Chr.11: 60432308-60432308 on Build GRCh38
rs921044389 Chr.11: 60432392-60432392 on Build GRCh38
rs369566123 Chr.11: 60432377-60432377 on Build GRCh38
rs769038818 Chr.11: 60432400-60432400 on Build GRCh38
rs967225492 Chr.11: 60432344-60432344 on Build GRCh38

View Full Product Details