Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 108176215-108176474 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRCAM
Entrez Gene ID
4897
Gene Name
neuronal cell adhesion molecule
Gene Aliases
-
UniGene
Hs.21422
Gene Chromosome Location
Chr.7: 108147626-108456442 on Build GRCh38
Transcripts
NM_001037132.2 NM_001193582.1 NM_001193583.1 NM_001193584.1 NM_005010.4 XM_005250373.2 XM_005250383.2 XM_005250385.2 XM_006716003.1 XM_006716007.1 XM_006716012.1 XM_006716014.1 XM_011516253.1 XM_011516255.1 XM_011516256.1 XM_011516257.1 XM_011516258.1 XM_011516259.1 XM_011516261.1 XM_011516262.1 XM_011516265.2 XM_011516266.2 XM_011516267.1 XM_011516268.2 XM_011516269.1 XM_011516270.1 XM_011516271.1 XM_017012236.1 XM_017012237.1 XM_017012238.1 XM_017012239.1 XM_017012240.1 XM_017012241.1 XM_017012242.1 XM_017012243.1 XM_017012244.1 XM_017012245.1 XM_017012246.1 XM_017012247.1 XM_017012248.1 XM_017012249.1 XM_017012250.1 XM_017012251.1 XM_017012252.1 XM_017012253.1 XM_017012254.1 XM_017012255.1 XM_017012256.1 XM_017012257.1 XM_017012258.1 XM_017012259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3674795 c.3145G>T p.D1049Y Chr.7: 108176436-108176436 on Build GRCh38
6175644 c.3131C>A p.A1044E Chr.7: 108176450-108176450 on Build GRCh38
7211333 c.3126G>A p.E1042E Chr.7: 108176455-108176455 on Build GRCh38
4708088 c.3108A>C p.S1036S Chr.7: 108176473-108176473 on Build GRCh38
7211331 c.3126G>A p.E1042E Chr.7: 108176455-108176455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767334085 Chr.7: 108176423-108176423 on Build GRCh38
rs774062367 Chr.7: 108176420-108176420 on Build GRCh38
rs777312097 Chr.7: 108176393-108176393 on Build GRCh38
rs773043323 Chr.7: 108176428-108176428 on Build GRCh38
rs978058216 Chr.7: 108176340-108176340 on Build GRCh38
rs889945624 Chr.7: 108176389-108176389 on Build GRCh38
rs996166133 Chr.7: 108176345-108176345 on Build GRCh38
rs570860144 Chr.7: 108176293-108176293 on Build GRCh38
rs768142182 Chr.7: 108176408-108176408 on Build GRCh38
rs901900648 Chr.7: 108176339-108176339 on Build GRCh38
rs771661129 Chr.7: 108176378-108176378 on Build GRCh38
rs761443725 Chr.7: 108176421-108176421 on Build GRCh38
rs748858702 Chr.7: 108176405-108176405 on Build GRCh38
rs551050761 Chr.7: 108176271-108176271 on Build GRCh38
rs752689383 Chr.7: 108176388-108176388 on Build GRCh38
rs539988988 Chr.7: 108176311-108176311 on Build GRCh38
rs17155125 Chr.7: 108176346-108176346 on Build GRCh38
rs771516752 Chr.7: 108176363-108176363 on Build GRCh38
rs765045400 Chr.7: 108176270-108176270 on Build GRCh38
rs146955970 Chr.7: 108176320-108176320 on Build GRCh38
rs917967802 Chr.7: 108176317-108176317 on Build GRCh38
rs760577886 Chr.7: 108176429-108176429 on Build GRCh38
rs927105643 Chr.7: 108176262-108176262 on Build GRCh38
rs1053986643 Chr.7: 108176347-108176347 on Build GRCh38
rs924235846 Chr.7: 108176344-108176344 on Build GRCh38
rs878969359 Chr.7: 108176248-108176248 on Build GRCh38

View Full Product Details