Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90906339-90906604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2A2
Entrez Gene ID
4122
Gene Name
mannosidase alpha class 2A member 2
Gene Aliases
MANA2X
UniGene
Hs.116459
Gene Chromosome Location
Chr.15: 90902218-90922585 on Build GRCh38
Transcripts
NM_001320977.1 NM_006122.3 NR_135502.1 NR_135503.1 NR_135504.1 XM_005254910.3 XM_011521565.2 XM_011521566.2 XM_017022184.1 XM_017022185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5759399 c.725A>G p.Q242R Chr.15: 90906387-90906387 on Build GRCh38
116710 c.732G>T p.E244D Chr.15: 90906394-90906394 on Build GRCh38
4418674 c.810A>T p.G270G Chr.15: 90906472-90906472 on Build GRCh38
4567478 c.729_730GG>AA p.E244K Chr.15: 90906391-90906392 on Build GRCh38
1286071 c.784G>T p.A262S Chr.15: 90906446-90906446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047691627 Chr.15: 90906509-90906509 on Build GRCh38
rs142426842 Chr.15: 90906383-90906383 on Build GRCh38
rs761190345 Chr.15: 90906453-90906453 on Build GRCh38
rs755915919 Chr.15: 90906532-90906533 on Build GRCh38
rs112038092 Chr.15: 90906500-90906500 on Build GRCh38
rs374045438 Chr.15: 90906531-90906531 on Build GRCh38
rs78252368 Chr.15: 90906571-90906571 on Build GRCh38
rs775827411 Chr.15: 90906432-90906432 on Build GRCh38
rs368402304 Chr.15: 90906427-90906427 on Build GRCh38
rs12594307 Chr.15: 90906472-90906472 on Build GRCh38
rs749986284 Chr.15: 90906466-90906466 on Build GRCh38
rs985500650 Chr.15: 90906487-90906487 on Build GRCh38
rs775915081 Chr.15: 90906359-90906359 on Build GRCh38
rs12591833 Chr.15: 90906530-90906530 on Build GRCh38
rs149181282 Chr.15: 90906382-90906382 on Build GRCh38
rs995992027 Chr.15: 90906558-90906558 on Build GRCh38
rs370746457 Chr.15: 90906367-90906367 on Build GRCh38
rs767280276 Chr.15: 90906527-90906527 on Build GRCh38
rs753676841 Chr.15: 90906479-90906479 on Build GRCh38
rs763926806 Chr.15: 90906548-90906573 on Build GRCh38
rs774876491 Chr.15: 90906450-90906450 on Build GRCh38
rs763365099 Chr.15: 90906435-90906435 on Build GRCh38
rs759056815 Chr.15: 90906502-90906507 on Build GRCh38
rs138836756 Chr.15: 90906574-90906574 on Build GRCh38
rs772775939 Chr.15: 90906546-90906546 on Build GRCh38
rs778965055 Chr.15: 90906488-90906488 on Build GRCh38
rs757602409 Chr.15: 90906478-90906478 on Build GRCh38
rs2072077 Chr.15: 90906424-90906424 on Build GRCh38
rs754872289 Chr.15: 90906486-90906486 on Build GRCh38
rs748703649 Chr.15: 90906538-90906538 on Build GRCh38
rs779055040 Chr.15: 90906405-90906405 on Build GRCh38
rs568139641 Chr.15: 90906550-90906550 on Build GRCh38
rs11630752 Chr.15: 90906580-90906580 on Build GRCh38
rs756315188 Chr.15: 90906510-90906510 on Build GRCh38
rs747067505 Chr.15: 90906412-90906412 on Build GRCh38
rs750856238 Chr.15: 90906505-90906505 on Build GRCh38
rs747813406 Chr.15: 90906562-90906562 on Build GRCh38
rs947677312 Chr.15: 90906579-90906579 on Build GRCh38
rs994077671 Chr.15: 90906419-90906419 on Build GRCh38
rs374359568 Chr.15: 90906399-90906399 on Build GRCh38
rs140631146 Chr.15: 90906421-90906421 on Build GRCh38
rs151326082 Chr.15: 90906400-90906400 on Build GRCh38
rs780303210 Chr.15: 90906522-90906522 on Build GRCh38
rs764652236 Chr.15: 90906379-90906379 on Build GRCh38
rs550047197 Chr.15: 90906544-90906544 on Build GRCh38
rs757223875 Chr.15: 90906504-90906504 on Build GRCh38
rs771086157 Chr.15: 90906416-90906416 on Build GRCh38
rs934856729 Chr.15: 90906565-90906565 on Build GRCh38
rs965460398 Chr.15: 90906516-90906516 on Build GRCh38
rs752390684 Chr.15: 90906533-90906552 on Build GRCh38
rs752698698 Chr.15: 90906498-90906498 on Build GRCh38
rs763450647 Chr.15: 90906375-90906375 on Build GRCh38
rs767078385 Chr.15: 90906457-90906457 on Build GRCh38
rs905867460 Chr.15: 90906581-90906581 on Build GRCh38

View Full Product Details