Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112086553-112086808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TIMM8B
Entrez Gene ID
26521
Gene Name
translocase of inner mitochondrial membrane 8 homolog B
Gene Aliases
DDP2, TIM8B
UniGene
Hs.279915
Gene Chromosome Location
Chr.11: 112084800-112086798 on Build GRCh38
Transcripts
NM_012459.2 NR_028383.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1229303 c.58G>T p.G20C Chr.11: 112086711-112086711 on Build GRCh38
6130951 c.61G>A p.E21K Chr.11: 112086708-112086708 on Build GRCh38
1475037 c.10C>A p.L4M Chr.11: 112086714-112086714 on Build GRCh38
923086 c.81A>G p.A27A Chr.11: 112086643-112086643 on Build GRCh38
1585381 c.126A>G p.A42A Chr.11: 112086643-112086643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372830468 Chr.11: 112086729-112086729 on Build GRCh38
rs753439722 Chr.11: 112086764-112086764 on Build GRCh38
rs370549054 Chr.11: 112086606-112086606 on Build GRCh38
rs763107283 Chr.11: 112086740-112086740 on Build GRCh38
rs778329771 Chr.11: 112086724-112086724 on Build GRCh38
rs112730013 Chr.11: 112086638-112086638 on Build GRCh38
rs1003347831 Chr.11: 112086776-112086776 on Build GRCh38
rs756852334 Chr.11: 112086766-112086766 on Build GRCh38
rs368094693 Chr.11: 112086622-112086622 on Build GRCh38
rs780502006 Chr.11: 112086663-112086663 on Build GRCh38
rs770471196 Chr.11: 112086675-112086675 on Build GRCh38
rs769285186 Chr.11: 112086667-112086667 on Build GRCh38
rs375164166 Chr.11: 112086612-112086612 on Build GRCh38
rs776373345 Chr.11: 112086693-112086693 on Build GRCh38
rs766875580 Chr.11: 112086711-112086711 on Build GRCh38
rs371410717 Chr.11: 112086737-112086737 on Build GRCh38
rs536857236 Chr.11: 112086704-112086704 on Build GRCh38
rs542296302 Chr.11: 112086775-112086775 on Build GRCh38
rs141007756 Chr.11: 112086681-112086681 on Build GRCh38
rs555586706 Chr.11: 112086784-112086784 on Build GRCh38
rs367779813 Chr.11: 112086731-112086731 on Build GRCh38
rs61734353 Chr.11: 112086703-112086703 on Build GRCh38
rs933258191 Chr.11: 112086575-112086575 on Build GRCh38
rs779626319 Chr.11: 112086651-112086651 on Build GRCh38
rs776642808 Chr.11: 112086688-112086711 on Build GRCh38
rs752103426 Chr.11: 112086750-112086750 on Build GRCh38
rs758845119 Chr.11: 112086661-112086661 on Build GRCh38
rs757898976 Chr.11: 112086778-112086778 on Build GRCh38
rs759538442 Chr.11: 112086695-112086695 on Build GRCh38
rs940596396 Chr.11: 112086759-112086759 on Build GRCh38
rs184311742 Chr.11: 112086623-112086623 on Build GRCh38
rs11553148 Chr.11: 112086694-112086694 on Build GRCh38
rs368516346 Chr.11: 112086719-112086719 on Build GRCh38
rs760495018 Chr.11: 112086701-112086701 on Build GRCh38
rs758936251 Chr.11: 112086709-112086709 on Build GRCh38
rs776728751 Chr.11: 112086734-112086734 on Build GRCh38
rs374827058 Chr.11: 112086730-112086730 on Build GRCh38
rs540010123 Chr.11: 112086626-112086626 on Build GRCh38
rs375367254 Chr.11: 112086715-112086715 on Build GRCh38
rs753913889 Chr.11: 112086645-112086645 on Build GRCh38
rs773379728 Chr.11: 112086739-112086739 on Build GRCh38
rs745826399 Chr.11: 112086725-112086725 on Build GRCh38
rs777463291 Chr.11: 112086668-112086668 on Build GRCh38
rs956040377 Chr.11: 112086639-112086639 on Build GRCh38
rs374830098 Chr.11: 112086768-112086768 on Build GRCh38
rs577227947 Chr.11: 112086706-112086706 on Build GRCh38
rs756774737 Chr.11: 112086722-112086722 on Build GRCh38
rs777356583 Chr.11: 112086718-112086718 on Build GRCh38
rs748790165 Chr.11: 112086673-112086673 on Build GRCh38
rs369355134 Chr.11: 112086753-112086753 on Build GRCh38
rs548020854 Chr.11: 112086670-112086670 on Build GRCh38
rs374914791 Chr.11: 112086738-112086738 on Build GRCh38
rs750950577 Chr.11: 112086659-112086659 on Build GRCh38
rs1052074957 Chr.11: 112086581-112086581 on Build GRCh38
rs757325354 Chr.11: 112086647-112086647 on Build GRCh38
rs914610565 Chr.11: 112086687-112086687 on Build GRCh38
rs148364534 Chr.11: 112086789-112086789 on Build GRCh38
rs557265620 Chr.11: 112086705-112086705 on Build GRCh38
rs368903806 Chr.11: 112086772-112086772 on Build GRCh38
rs767037177 Chr.11: 112086745-112086745 on Build GRCh38
rs767866062 Chr.11: 112086761-112086761 on Build GRCh38
rs764254817 Chr.11: 112086636-112086636 on Build GRCh38
rs747868911 Chr.11: 112086666-112086666 on Build GRCh38
rs765500251 Chr.11: 112086692-112086693 on Build GRCh38
rs774241163 Chr.11: 112086611-112086611 on Build GRCh38

View Full Product Details