Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 4900833-4901104 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf107
Entrez Gene ID
100130311
Gene Name
chromosome 17 open reading frame 107
Gene Aliases
-
UniGene
Hs.579243
Gene Chromosome Location
Chr.17: 4899653-4902932 on Build GRCh38
Transcripts
NM_001145536.1 NM_000080.3 XM_017024115.1

Gene Symbol
CHRNE
Entrez Gene ID
1145
Gene Name
cholinergic receptor nicotinic epsilon subunit
Gene Aliases
ACHRE, CMS1D, CMS1E, CMS2A, CMS4A, CMS4B, CMS4C, FCCMS, SCCMS
UniGene
Hs.654535
Gene Chromosome Location
Chr.17: 4897769-4905019 on Build GRCh38
Transcripts
NM_001145536.1 NM_000080.3 XM_017024115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6081535 c.688G>T p.V230F Chr.17: 4901104-4901104 on Build GRCh38
5936282 c.713G>A p.R238Q Chr.17: 4901079-4901079 on Build GRCh38
1384467 c.876G>A p.Q292Q Chr.17: 4900834-4900834 on Build GRCh38
1384468 c.699G>A p.S233S Chr.17: 4901093-4901093 on Build GRCh38
4579822 c.818G>C p.C273S Chr.17: 4900892-4900892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772723622 Chr.17: 4901009-4901009 on Build GRCh38
rs374113313 Chr.17: 4900975-4900975 on Build GRCh38
rs756407780 Chr.17: 4901036-4901036 on Build GRCh38
rs201434993 Chr.17: 4901082-4901082 on Build GRCh38
rs121909510 Chr.17: 4900860-4900860 on Build GRCh38
rs775149431 Chr.17: 4900906-4900906 on Build GRCh38
rs755437862 Chr.17: 4900972-4900972 on Build GRCh38
rs753179619 Chr.17: 4900956-4900957 on Build GRCh38
rs900612569 Chr.17: 4900872-4900872 on Build GRCh38
rs1021297889 Chr.17: 4900903-4900903 on Build GRCh38
rs887770442 Chr.17: 4900891-4900891 on Build GRCh38
rs767922649 Chr.17: 4900911-4900912 on Build GRCh38
rs757421529 Chr.17: 4901047-4901047 on Build GRCh38
rs774407731 Chr.17: 4900890-4900890 on Build GRCh38
rs764752088 Chr.17: 4900937-4900937 on Build GRCh38
rs752926340 Chr.17: 4901030-4901030 on Build GRCh38
rs773929089 Chr.17: 4900863-4900863 on Build GRCh38
rs761293676 Chr.17: 4900931-4900931 on Build GRCh38
rs776407381 Chr.17: 4900929-4900929 on Build GRCh38
rs201841042 Chr.17: 4900889-4900889 on Build GRCh38
rs745731778 Chr.17: 4900987-4900987 on Build GRCh38
rs201463598 Chr.17: 4900995-4900995 on Build GRCh38
rs780429979 Chr.17: 4900864-4900864 on Build GRCh38
rs201042568 Chr.17: 4900945-4900945 on Build GRCh38
rs1040580878 Chr.17: 4900859-4900859 on Build GRCh38
rs772145808 Chr.17: 4900990-4900990 on Build GRCh38
rs756675414 Chr.17: 4900998-4900998 on Build GRCh38
rs756213406 Chr.17: 4900983-4900983 on Build GRCh38
rs750981766 Chr.17: 4900862-4900862 on Build GRCh38
rs750892511 Chr.17: 4900944-4900944 on Build GRCh38
rs961218388 Chr.17: 4900913-4900913 on Build GRCh38
rs762704832 Chr.17: 4900910-4900913 on Build GRCh38
rs780094155 Chr.17: 4900992-4900992 on Build GRCh38
rs1015178267 Chr.17: 4900911-4900911 on Build GRCh38
rs781536510 Chr.17: 4900888-4900888 on Build GRCh38
rs996197218 Chr.17: 4900876-4900876 on Build GRCh38
rs932101535 Chr.17: 4901057-4901057 on Build GRCh38
rs994082504 Chr.17: 4901075-4901076 on Build GRCh38
rs969873887 Chr.17: 4900951-4900951 on Build GRCh38
rs762509943 Chr.17: 4901011-4901011 on Build GRCh38
rs766710383 Chr.17: 4900949-4900949 on Build GRCh38
rs753951775 Chr.17: 4901043-4901043 on Build GRCh38
rs28999110 Chr.17: 4901071-4901071 on Build GRCh38
rs1049088476 Chr.17: 4900880-4900880 on Build GRCh38
rs886053126 Chr.17: 4901069-4901069 on Build GRCh38
rs778028755 Chr.17: 4900984-4900984 on Build GRCh38
rs747189862 Chr.17: 4900869-4900869 on Build GRCh38
rs759226183 Chr.17: 4900998-4900998 on Build GRCh38
rs751857871 Chr.17: 4900957-4900957 on Build GRCh38
rs745821689 Chr.17: 4900892-4900892 on Build GRCh38
rs768177094 Chr.17: 4900923-4900923 on Build GRCh38
rs746993242 Chr.17: 4901077-4901077 on Build GRCh38
rs921757512 Chr.17: 4901049-4901049 on Build GRCh38
rs766792066 Chr.17: 4901012-4901012 on Build GRCh38
rs199876621 Chr.17: 4900981-4900981 on Build GRCh38
rs777939796 Chr.17: 4901039-4901039 on Build GRCh38
rs570266272 Chr.17: 4900883-4900883 on Build GRCh38
rs746772722 Chr.17: 4900994-4900994 on Build GRCh38
rs1049141147 Chr.17: 4901072-4901072 on Build GRCh38
rs150133823 Chr.17: 4901066-4901066 on Build GRCh38
rs770586668 Chr.17: 4900858-4900858 on Build GRCh38
rs933082734 Chr.17: 4900866-4900866 on Build GRCh38
rs767945817 Chr.17: 4901027-4901027 on Build GRCh38
rs867237170 Chr.17: 4901041-4901041 on Build GRCh38
rs372509633 Chr.17: 4901024-4901024 on Build GRCh38
rs979841930 Chr.17: 4900974-4900974 on Build GRCh38
rs755170993 Chr.17: 4900879-4900879 on Build GRCh38
rs769228953 Chr.17: 4901008-4901008 on Build GRCh38
rs752148595 Chr.17: 4901015-4901015 on Build GRCh38
rs761487611 Chr.17: 4901003-4901003 on Build GRCh38
rs760314762 Chr.17: 4900915-4900915 on Build GRCh38
rs768299579 Chr.17: 4901078-4901078 on Build GRCh38
rs140846401 Chr.17: 4901023-4901023 on Build GRCh38

View Full Product Details