Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 34200807-34201058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf94
Entrez Gene ID
84970
Gene Name
chromosome 1 open reading frame 94
Gene Aliases
-
UniGene
Hs.194610
Gene Chromosome Location
Chr.1: 34166883-34219131 on Build GRCh38
Transcripts
NM_001134734.1 NM_032884.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
268252 c.664C>T p.R222* Chr.1: 34200996-34200996 on Build GRCh38
6126498 c.695T>C p.L232P Chr.1: 34201027-34201027 on Build GRCh38
2079756 c.1152G>A p.K384K Chr.1: 34200914-34200914 on Build GRCh38
6063293 c.1270+1G>A p.? Chr.1: 34201033-34201033 on Build GRCh38
1342057 c.1227G>A p.P409P Chr.1: 34200989-34200989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764072490 Chr.1: 34200915-34200915 on Build GRCh38
rs774050391 Chr.1: 34200835-34200835 on Build GRCh38
rs775448857 Chr.1: 34200996-34200996 on Build GRCh38
rs372183455 Chr.1: 34201017-34201017 on Build GRCh38
rs753248839 Chr.1: 34200949-34200949 on Build GRCh38
rs144742239 Chr.1: 34200911-34200911 on Build GRCh38
rs375613990 Chr.1: 34200932-34200932 on Build GRCh38
rs758569553 Chr.1: 34200866-34200866 on Build GRCh38
rs759779250 Chr.1: 34201021-34201021 on Build GRCh38
rs140030017 Chr.1: 34200872-34200872 on Build GRCh38
rs772043436 Chr.1: 34200989-34200989 on Build GRCh38
rs145466967 Chr.1: 34200980-34200980 on Build GRCh38
rs770499000 Chr.1: 34200905-34200905 on Build GRCh38
rs776573759 Chr.1: 34201013-34201013 on Build GRCh38
rs373115271 Chr.1: 34200859-34200859 on Build GRCh38
rs756648350 Chr.1: 34200959-34200959 on Build GRCh38
rs115310612 Chr.1: 34200944-34200944 on Build GRCh38
rs754025034 Chr.1: 34200955-34200955 on Build GRCh38
rs753066946 Chr.1: 34201025-34201025 on Build GRCh38
rs771988395 Chr.1: 34200909-34200909 on Build GRCh38
rs754760756 Chr.1: 34200937-34200938 on Build GRCh38
rs374427865 Chr.1: 34200997-34200997 on Build GRCh38
rs535230432 Chr.1: 34200957-34200959 on Build GRCh38
rs769331067 Chr.1: 34200890-34200890 on Build GRCh38
rs751789307 Chr.1: 34200933-34200933 on Build GRCh38
rs142985527 Chr.1: 34200853-34200853 on Build GRCh38
rs775610323 Chr.1: 34200910-34200910 on Build GRCh38
rs114118360 Chr.1: 34200988-34200988 on Build GRCh38
rs548017054 Chr.1: 34200938-34200938 on Build GRCh38
rs780152081 Chr.1: 34200891-34200891 on Build GRCh38
rs750560713 Chr.1: 34200922-34200922 on Build GRCh38
rs371680483 Chr.1: 34200908-34200908 on Build GRCh38
rs750724452 Chr.1: 34200863-34200863 on Build GRCh38
rs115878225 Chr.1: 34200907-34200907 on Build GRCh38
rs142411412 Chr.1: 34200981-34200981 on Build GRCh38
rs781502570 Chr.1: 34200942-34200942 on Build GRCh38
rs748765822 Chr.1: 34200970-34200970 on Build GRCh38
rs114203973 Chr.1: 34201022-34201022 on Build GRCh38
rs370854360 Chr.1: 34200851-34200851 on Build GRCh38
rs759312025 Chr.1: 34200850-34200850 on Build GRCh38
rs149791275 Chr.1: 34200878-34200878 on Build GRCh38
rs145958815 Chr.1: 34200852-34200852 on Build GRCh38
rs370151677 Chr.1: 34200961-34200961 on Build GRCh38
rs781704626 Chr.1: 34200879-34200879 on Build GRCh38
rs763023074 Chr.1: 34200930-34200930 on Build GRCh38
rs748793632 Chr.1: 34200894-34200894 on Build GRCh38
rs762997561 Chr.1: 34201015-34201015 on Build GRCh38
rs914064876 Chr.1: 34200963-34200963 on Build GRCh38
rs774474330 Chr.1: 34201020-34201020 on Build GRCh38
rs945525412 Chr.1: 34200965-34200965 on Build GRCh38
rs376238971 Chr.1: 34200877-34200877 on Build GRCh38

View Full Product Details