Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 12698414-12698687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TYRP1
Entrez Gene ID
7306
Gene Name
tyrosinase related protein 1
Gene Aliases
CAS2, CATB, GP75, OCA3, TRP, TRP1, TYRP, b-PROTEIN
UniGene
Hs.270279
Gene Chromosome Location
Chr.9: 12693385-12710266 on Build GRCh38
Transcripts
NM_000550.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3763623 c.729T>C p.S243S Chr.9: 12698471-12698471 on Build GRCh38
1636021 c.903A>G p.T301T Chr.9: 12698645-12698645 on Build GRCh38
6814701 c.913+1G>T p.? Chr.9: 12698656-12698656 on Build GRCh38
1489661 c.858G>A p.W286* Chr.9: 12698600-12698600 on Build GRCh38
6879709 c.760G>A p.G254R Chr.9: 12698502-12698502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200160846 Chr.9: 12698528-12698528 on Build GRCh38
rs748926400 Chr.9: 12698656-12698656 on Build GRCh38
rs757120481 Chr.9: 12698520-12698520 on Build GRCh38
rs769437036 Chr.9: 12698567-12698567 on Build GRCh38
rs202126779 Chr.9: 12698500-12698500 on Build GRCh38
rs139647745 Chr.9: 12698510-12698510 on Build GRCh38
rs770141693 Chr.9: 12698646-12698646 on Build GRCh38
rs377629560 Chr.9: 12698481-12698481 on Build GRCh38
rs762386039 Chr.9: 12698571-12698571 on Build GRCh38
rs748738090 Chr.9: 12698466-12698466 on Build GRCh38
rs749452154 Chr.9: 12698444-12698444 on Build GRCh38
rs767466270 Chr.9: 12698580-12698580 on Build GRCh38
rs187959351 Chr.9: 12698641-12698641 on Build GRCh38
rs779089153 Chr.9: 12698531-12698531 on Build GRCh38
rs752110288 Chr.9: 12698629-12698629 on Build GRCh38
rs747320688 Chr.9: 12698482-12698482 on Build GRCh38
rs777863437 Chr.9: 12698618-12698618 on Build GRCh38
rs960474548 Chr.9: 12698648-12698648 on Build GRCh38
rs765071773 Chr.9: 12698575-12698575 on Build GRCh38
rs370404984 Chr.9: 12698523-12698523 on Build GRCh38
rs182508840 Chr.9: 12698565-12698565 on Build GRCh38
rs368145423 Chr.9: 12698574-12698574 on Build GRCh38
rs766031869 Chr.9: 12698589-12698590 on Build GRCh38
rs201345670 Chr.9: 12698539-12698539 on Build GRCh38
rs999232321 Chr.9: 12698542-12698542 on Build GRCh38
rs61752939 Chr.9: 12698527-12698527 on Build GRCh38
rs758440173 Chr.9: 12698602-12698602 on Build GRCh38
rs774836325 Chr.9: 12698503-12698503 on Build GRCh38
rs1031656874 Chr.9: 12698549-12698549 on Build GRCh38
rs1038628786 Chr.9: 12698505-12698505 on Build GRCh38
rs900003133 Chr.9: 12698513-12698513 on Build GRCh38
rs759390349 Chr.9: 12698646-12698647 on Build GRCh38
rs371227148 Chr.9: 12698501-12698501 on Build GRCh38
rs747475719 Chr.9: 12698559-12698559 on Build GRCh38
rs775126268 Chr.9: 12698568-12698568 on Build GRCh38
rs778806389 Chr.9: 12698455-12698455 on Build GRCh38
rs202189890 Chr.9: 12698552-12698552 on Build GRCh38
rs771307980 Chr.9: 12698562-12698562 on Build GRCh38
rs371562555 Chr.9: 12698595-12698595 on Build GRCh38
rs757733461 Chr.9: 12698638-12698638 on Build GRCh38
rs941234320 Chr.9: 12698470-12698470 on Build GRCh38
rs776296557 Chr.9: 12698492-12698492 on Build GRCh38
rs193035382 Chr.9: 12698649-12698649 on Build GRCh38
rs754993789 Chr.9: 12698573-12698573 on Build GRCh38
rs751453501 Chr.9: 12698516-12698516 on Build GRCh38
rs763016978 Chr.9: 12698507-12698507 on Build GRCh38
rs753419037 Chr.9: 12698601-12698614 on Build GRCh38
rs1045422255 Chr.9: 12698522-12698522 on Build GRCh38
rs750211458 Chr.9: 12698582-12698582 on Build GRCh38
rs778148332 Chr.9: 12698555-12698555 on Build GRCh38
rs757247093 Chr.9: 12698631-12698631 on Build GRCh38
rs748170782 Chr.9: 12698538-12698538 on Build GRCh38
rs746220547 Chr.9: 12698644-12698644 on Build GRCh38
rs760181936 Chr.9: 12698522-12698533 on Build GRCh38
rs755136711 Chr.9: 12698446-12698446 on Build GRCh38
rs35866166 Chr.9: 12698471-12698471 on Build GRCh38
rs374769303 Chr.9: 12698660-12698660 on Build GRCh38
rs772638423 Chr.9: 12698477-12698480 on Build GRCh38
rs767376414 Chr.9: 12698524-12698524 on Build GRCh38
rs771220162 Chr.9: 12698489-12698489 on Build GRCh38
rs200754545 Chr.9: 12698487-12698487 on Build GRCh38

View Full Product Details