Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7893872-7894133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD3
Entrez Gene ID
1107
Gene Name
chromodomain helicase DNA binding protein 3
Gene Aliases
Mi-2a, Mi2-ALPHA, ZFH
UniGene
Hs.25601
Gene Chromosome Location
Chr.17: 7884764-7912757 on Build GRCh38
Transcripts
NM_001005271.2 NM_001005273.2 NM_005852.3 XM_005256427.4 XM_005256428.4 XM_005256429.4 XM_005256431.4 XM_006721423.3 XM_006721424.3 XM_006721428.3 XM_017024061.1 XM_017024062.1 XM_017024063.1 XM_017024064.1 XM_017024065.1 XM_017024066.1 XM_017024067.1 XM_017024068.1 XM_017024069.1 XM_017024070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6315969 c.924+7G>T p.? Chr.17: 7893942-7893942 on Build GRCh38
1248204 c.911A>T p.K304M Chr.17: 7893922-7893922 on Build GRCh38
6347576 c.920G>T p.G307V Chr.17: 7893931-7893931 on Build GRCh38
1166744 c.929T>C p.V310A Chr.17: 7894119-7894119 on Build GRCh38
1580705 c.861G>T p.K287N Chr.17: 7893872-7893872 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375616544 Chr.17: 7894083-7894083 on Build GRCh38
rs772132381 Chr.17: 7893959-7893959 on Build GRCh38
rs963882405 Chr.17: 7893930-7893930 on Build GRCh38
rs549222344 Chr.17: 7893996-7893996 on Build GRCh38
rs761715430 Chr.17: 7894065-7894066 on Build GRCh38
rs1025007835 Chr.17: 7894035-7894035 on Build GRCh38
rs747464671 Chr.17: 7894107-7894107 on Build GRCh38
rs527979074 Chr.17: 7893974-7893974 on Build GRCh38
rs895185094 Chr.17: 7894015-7894015 on Build GRCh38
rs757171450 Chr.17: 7894039-7894039 on Build GRCh38
rs78632606 Chr.17: 7893935-7893935 on Build GRCh38
rs758686232 Chr.17: 7894102-7894102 on Build GRCh38
rs145306324 Chr.17: 7893994-7893995 on Build GRCh38
rs199568516 Chr.17: 7894101-7894101 on Build GRCh38
rs760441335 Chr.17: 7893981-7894009 on Build GRCh38
rs202009423 Chr.17: 7894097-7894097 on Build GRCh38
rs1043190902 Chr.17: 7894063-7894063 on Build GRCh38
rs778894067 Chr.17: 7893951-7893951 on Build GRCh38
rs755101105 Chr.17: 7894100-7894101 on Build GRCh38
rs1001435359 Chr.17: 7893991-7893991 on Build GRCh38
rs781081716 Chr.17: 7893946-7893946 on Build GRCh38
rs780494957 Chr.17: 7893967-7893967 on Build GRCh38
rs757319732 Chr.17: 7893948-7893948 on Build GRCh38
rs573962893 Chr.17: 7894111-7894111 on Build GRCh38
rs747304332 Chr.17: 7893970-7893970 on Build GRCh38
rs376928718 Chr.17: 7894069-7894069 on Build GRCh38
rs780115167 Chr.17: 7894103-7894103 on Build GRCh38
rs996661018 Chr.17: 7893933-7893933 on Build GRCh38
rs534122095 Chr.17: 7893970-7894000 on Build GRCh38
rs377180914 Chr.17: 7893905-7893905 on Build GRCh38
rs759308763 Chr.17: 7894094-7894094 on Build GRCh38
rs531443908 Chr.17: 7894027-7894027 on Build GRCh38
rs762689656 Chr.17: 7894082-7894082 on Build GRCh38
rs567731285 Chr.17: 7894003-7894003 on Build GRCh38
rs987169301 Chr.17: 7894104-7894104 on Build GRCh38
rs751264217 Chr.17: 7893934-7893934 on Build GRCh38
rs374434743 Chr.17: 7894108-7894108 on Build GRCh38
rs535053246 Chr.17: 7894005-7894005 on Build GRCh38
rs781636257 Chr.17: 7894109-7894109 on Build GRCh38
rs772539844 Chr.17: 7894080-7894080 on Build GRCh38
rs377613189 Chr.17: 7894090-7894090 on Build GRCh38
rs764276937 Chr.17: 7894089-7894092 on Build GRCh38
rs376849433 Chr.17: 7893969-7893969 on Build GRCh38
rs750087267 Chr.17: 7893903-7893903 on Build GRCh38
rs777263214 Chr.17: 7894065-7894065 on Build GRCh38
rs776546162 Chr.17: 7893983-7893984 on Build GRCh38
rs764034469 Chr.17: 7893965-7893965 on Build GRCh38
rs200857173 Chr.17: 7893962-7893962 on Build GRCh38
rs371364679 Chr.17: 7894070-7894070 on Build GRCh38
rs775714799 Chr.17: 7893978-7893979 on Build GRCh38
rs75122325 Chr.17: 7893947-7893947 on Build GRCh38
rs904069648 Chr.17: 7893972-7893972 on Build GRCh38
rs766209905 Chr.17: 7893925-7893925 on Build GRCh38
rs568976535 Chr.17: 7894028-7894028 on Build GRCh38
rs763918158 Chr.17: 7893981-7893982 on Build GRCh38
rs201232886 Chr.17: 7893979-7893979 on Build GRCh38
rs1033780112 Chr.17: 7894011-7894011 on Build GRCh38
rs115087532 Chr.17: 7894040-7894040 on Build GRCh38
rs373120724 Chr.17: 7893984-7893984 on Build GRCh38
rs760820477 Chr.17: 7894072-7894073 on Build GRCh38
rs116670269 Chr.17: 7894100-7894100 on Build GRCh38
rs374961455 Chr.17: 7894095-7894095 on Build GRCh38
rs71359656 Chr.17: 7893972-7893973 on Build GRCh38

View Full Product Details