Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7436668-7436939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM102
Entrez Gene ID
284114
Gene Name
transmembrane protein 102
Gene Aliases
CBAP
UniGene
Hs.655662
Gene Chromosome Location
Chr.17: 7435443-7437679 on Build GRCh38
Transcripts
NM_001320444.1 NM_178518.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1229496 c.724G>T p.A242S Chr.17: 7436703-7436703 on Build GRCh38
6243964 c.727G>T p.A243S Chr.17: 7436706-7436706 on Build GRCh38
4837938 c.753G>T p.P251P Chr.17: 7436732-7436732 on Build GRCh38
227133 c.857G>A p.G286D Chr.17: 7436836-7436836 on Build GRCh38
7394057 c.863C>G p.P288R Chr.17: 7436842-7436842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749535709 Chr.17: 7436735-7436735 on Build GRCh38
rs142866620 Chr.17: 7436706-7436706 on Build GRCh38
rs982380928 Chr.17: 7436814-7436814 on Build GRCh38
rs764925303 Chr.17: 7436907-7436917 on Build GRCh38
rs534406142 Chr.17: 7436787-7436787 on Build GRCh38
rs919409619 Chr.17: 7436793-7436793 on Build GRCh38
rs780548070 Chr.17: 7436878-7436878 on Build GRCh38
rs1021553485 Chr.17: 7436916-7436916 on Build GRCh38
rs562274930 Chr.17: 7436714-7436714 on Build GRCh38
rs746348316 Chr.17: 7436766-7436766 on Build GRCh38
rs202087288 Chr.17: 7436908-7436908 on Build GRCh38
rs771774177 Chr.17: 7436783-7436783 on Build GRCh38
rs775889675 Chr.17: 7436771-7436771 on Build GRCh38
rs891634932 Chr.17: 7436843-7436843 on Build GRCh38
rs760753161 Chr.17: 7436836-7436836 on Build GRCh38
rs376235154 Chr.17: 7436895-7436895 on Build GRCh38
rs563100483 Chr.17: 7436799-7436799 on Build GRCh38
rs779491141 Chr.17: 7436763-7436763 on Build GRCh38
rs772881094 Chr.17: 7436918-7436918 on Build GRCh38
rs777987292 Chr.17: 7436732-7436732 on Build GRCh38
rs745454020 Chr.17: 7436731-7436731 on Build GRCh38
rs762108785 Chr.17: 7436888-7436889 on Build GRCh38
rs530508330 Chr.17: 7436863-7436863 on Build GRCh38
rs531917336 Chr.17: 7436774-7436774 on Build GRCh38
rs753938549 Chr.17: 7436885-7436886 on Build GRCh38
rs764186213 Chr.17: 7436755-7436757 on Build GRCh38
rs574111274 Chr.17: 7436769-7436769 on Build GRCh38
rs747775232 Chr.17: 7436901-7436901 on Build GRCh38
rs938285510 Chr.17: 7436855-7436855 on Build GRCh38
rs780719919 Chr.17: 7436896-7436896 on Build GRCh38
rs767471413 Chr.17: 7436711-7436711 on Build GRCh38
rs747525167 Chr.17: 7436882-7436882 on Build GRCh38
rs764138950 Chr.17: 7436851-7436851 on Build GRCh38
rs751456353 Chr.17: 7436832-7436832 on Build GRCh38
rs1055361876 Chr.17: 7436874-7436874 on Build GRCh38
rs766301556 Chr.17: 7436813-7436813 on Build GRCh38
rs754111817 Chr.17: 7436859-7436859 on Build GRCh38
rs997341556 Chr.17: 7436720-7436720 on Build GRCh38
rs762758558 Chr.17: 7436806-7436806 on Build GRCh38
rs564203341 Chr.17: 7436913-7436913 on Build GRCh38
rs779402397 Chr.17: 7436864-7436864 on Build GRCh38
rs750771562 Chr.17: 7436867-7436867 on Build GRCh38
rs926826911 Chr.17: 7436834-7436834 on Build GRCh38
rs773039467 Chr.17: 7436805-7436805 on Build GRCh38
rs544943453 Chr.17: 7436795-7436795 on Build GRCh38
rs750355901 Chr.17: 7436699-7436699 on Build GRCh38
rs758690914 Chr.17: 7436869-7436869 on Build GRCh38
rs752772136 Chr.17: 7436712-7436712 on Build GRCh38
rs761301484 Chr.17: 7436870-7436870 on Build GRCh38
rs776249270 Chr.17: 7436785-7436785 on Build GRCh38
rs1016285320 Chr.17: 7436708-7436708 on Build GRCh38
rs947120003 Chr.17: 7436911-7436911 on Build GRCh38
rs757686737 Chr.17: 7436759-7436759 on Build GRCh38
rs746459982 Chr.17: 7436772-7436772 on Build GRCh38

View Full Product Details