Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196656915-196657174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC150
Entrez Gene ID
284992
Gene Name
coiled-coil domain containing 150
Gene Aliases
-
UniGene
Hs.132519
Gene Chromosome Location
Chr.2: 196639632-196733668 on Build GRCh38
Transcripts
NM_001080539.1 XM_006712438.3 XM_011510985.2 XM_011510988.2 XM_017003861.1 XM_017003862.1 XM_017003863.1 XM_017003864.1 XM_017003865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1482587 c.465C>T p.L155L Chr.2: 196657025-196657025 on Build GRCh38
3838208 c.453G>T p.K151N Chr.2: 196657013-196657013 on Build GRCh38
1014404 c.548C>T p.A183V Chr.2: 196657108-196657108 on Build GRCh38
1014403 c.481C>T p.R161C Chr.2: 196657041-196657041 on Build GRCh38
4089867 c.567G>A p.S189S Chr.2: 196657127-196657127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781772582 Chr.2: 196656958-196656958 on Build GRCh38
rs757646842 Chr.2: 196657033-196657033 on Build GRCh38
rs756750979 Chr.2: 196657052-196657052 on Build GRCh38
rs1053670635 Chr.2: 196657036-196657036 on Build GRCh38
rs747654565 Chr.2: 196656973-196656973 on Build GRCh38
rs765158094 Chr.2: 196657007-196657007 on Build GRCh38
rs766839293 Chr.2: 196656945-196656945 on Build GRCh38
rs146046280 Chr.2: 196657127-196657127 on Build GRCh38
rs768296702 Chr.2: 196656970-196656970 on Build GRCh38
rs909877313 Chr.2: 196656954-196656954 on Build GRCh38
rs746514107 Chr.2: 196657137-196657137 on Build GRCh38
rs561326085 Chr.2: 196657093-196657093 on Build GRCh38
rs775361379 Chr.2: 196657076-196657076 on Build GRCh38
rs34133636 Chr.2: 196657026-196657026 on Build GRCh38
rs750855228 Chr.2: 196657110-196657110 on Build GRCh38
rs368558740 Chr.2: 196656937-196656937 on Build GRCh38
rs369742781 Chr.2: 196657042-196657042 on Build GRCh38
rs779985058 Chr.2: 196657128-196657128 on Build GRCh38
rs572442181 Chr.2: 196657060-196657060 on Build GRCh38
rs758716984 Chr.2: 196656947-196656947 on Build GRCh38
rs781731493 Chr.2: 196657016-196657016 on Build GRCh38
rs770612889 Chr.2: 196657143-196657143 on Build GRCh38
rs757929204 Chr.2: 196657131-196657131 on Build GRCh38
rs371236615 Chr.2: 196657107-196657107 on Build GRCh38
rs192386368 Chr.2: 196656948-196656948 on Build GRCh38
rs745895649 Chr.2: 196657052-196657053 on Build GRCh38
rs746572664 Chr.2: 196657062-196657062 on Build GRCh38
rs780675493 Chr.2: 196657055-196657055 on Build GRCh38
rs781642729 Chr.2: 196657041-196657041 on Build GRCh38
rs771795597 Chr.2: 196656974-196656974 on Build GRCh38
rs541438855 Chr.2: 196657068-196657068 on Build GRCh38
rs763124897 Chr.2: 196657092-196657092 on Build GRCh38
rs762923560 Chr.2: 196657025-196657025 on Build GRCh38
rs936863285 Chr.2: 196657114-196657114 on Build GRCh38
rs990843427 Chr.2: 196657040-196657040 on Build GRCh38
rs142774075 Chr.2: 196657105-196657105 on Build GRCh38
rs746311530 Chr.2: 196656963-196656963 on Build GRCh38
rs777088827 Chr.2: 196657133-196657133 on Build GRCh38
rs770723440 Chr.2: 196657064-196657064 on Build GRCh38
rs751821946 Chr.2: 196657101-196657101 on Build GRCh38
rs373513944 Chr.2: 196657056-196657056 on Build GRCh38
rs368906039 Chr.2: 196656972-196656972 on Build GRCh38
rs200072904 Chr.2: 196657011-196657011 on Build GRCh38
rs760537528 Chr.2: 196656979-196656979 on Build GRCh38
rs769775155 Chr.2: 196657075-196657075 on Build GRCh38
rs770588840 Chr.2: 196656985-196656985 on Build GRCh38
rs755378315 Chr.2: 196656990-196656990 on Build GRCh38
rs950093867 Chr.2: 196656995-196656995 on Build GRCh38
rs372845500 Chr.2: 196657126-196657126 on Build GRCh38
rs372655242 Chr.2: 196657003-196657003 on Build GRCh38
rs376154930 Chr.2: 196656949-196656949 on Build GRCh38
rs776550640 Chr.2: 196657002-196657002 on Build GRCh38

View Full Product Details