Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 122274953-122275224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD16
Entrez Gene ID
118663
Gene Name
BTB domain containing 16
Gene Aliases
C10orf87
UniGene
Hs.422466
Gene Chromosome Location
Chr.10: 122270745-122338180 on Build GRCh38
Transcripts
NM_001318189.1 NM_144587.3 XM_011539239.2 XM_011539240.2 XM_011539241.2 XM_011539242.2 XM_017015636.1 XM_017015637.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2056824 c.11C>T p.S4L Chr.10: 122275092-122275092 on Build GRCh38
1317481 c.17C>T p.T6M Chr.10: 122275098-122275098 on Build GRCh38
6613339 c.18G>A p.T6T Chr.10: 122275099-122275099 on Build GRCh38
7141798 c.12G>T p.S4S Chr.10: 122275093-122275093 on Build GRCh38
3434986 c.3G>A p.M1I Chr.10: 122275084-122275084 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749930849 Chr.10: 122275199-122275199 on Build GRCh38
rs749355195 Chr.10: 122275082-122275082 on Build GRCh38
rs372045833 Chr.10: 122274999-122274999 on Build GRCh38
rs201366692 Chr.10: 122275093-122275093 on Build GRCh38
rs757960672 Chr.10: 122275103-122275103 on Build GRCh38
rs1047076527 Chr.10: 122275195-122275195 on Build GRCh38
rs897981826 Chr.10: 122275059-122275059 on Build GRCh38
rs533560810 Chr.10: 122275024-122275024 on Build GRCh38
rs370307107 Chr.10: 122275056-122275056 on Build GRCh38
rs747568703 Chr.10: 122275062-122275062 on Build GRCh38
rs202065129 Chr.10: 122275098-122275098 on Build GRCh38
rs144192653 Chr.10: 122275087-122275087 on Build GRCh38
rs528281696 Chr.10: 122275026-122275026 on Build GRCh38
rs979306019 Chr.10: 122275167-122275167 on Build GRCh38
rs756474278 Chr.10: 122275136-122275136 on Build GRCh38
rs756908935 Chr.10: 122275099-122275099 on Build GRCh38
rs1050856148 Chr.10: 122275198-122275198 on Build GRCh38
rs1050760235 Chr.10: 122274991-122274991 on Build GRCh38
rs7080859 Chr.10: 122275188-122275188 on Build GRCh38
rs746231038 Chr.10: 122275070-122275070 on Build GRCh38
rs969519112 Chr.10: 122275078-122275078 on Build GRCh38
rs920982980 Chr.10: 122275174-122275174 on Build GRCh38
rs917524336 Chr.10: 122275181-122275181 on Build GRCh38
rs148578895 Chr.10: 122275063-122275063 on Build GRCh38
rs369262041 Chr.10: 122275005-122275005 on Build GRCh38
rs758017882 Chr.10: 122275044-122275044 on Build GRCh38
rs7096744 Chr.10: 122275013-122275013 on Build GRCh38
rs563023839 Chr.10: 122275018-122275018 on Build GRCh38
rs978519397 Chr.10: 122275110-122275110 on Build GRCh38
rs372370160 Chr.10: 122275114-122275114 on Build GRCh38
rs946500906 Chr.10: 122275153-122275153 on Build GRCh38
rs201539893 Chr.10: 122275101-122275101 on Build GRCh38
rs773115186 Chr.10: 122275089-122275089 on Build GRCh38
rs764907336 Chr.10: 122275100-122275100 on Build GRCh38
rs374347900 Chr.10: 122275120-122275120 on Build GRCh38
rs753278526 Chr.10: 122275107-122275107 on Build GRCh38
rs570472899 Chr.10: 122275117-122275117 on Build GRCh38
rs376078771 Chr.10: 122275039-122275039 on Build GRCh38
rs780300597 Chr.10: 122275143-122275143 on Build GRCh38
rs539329146 Chr.10: 122275130-122275130 on Build GRCh38
rs775605643 Chr.10: 122275076-122275076 on Build GRCh38
rs367945813 Chr.10: 122275149-122275149 on Build GRCh38
rs565874548 Chr.10: 122275109-122275109 on Build GRCh38
rs202210822 Chr.10: 122275092-122275092 on Build GRCh38
rs560605264 Chr.10: 122275071-122275071 on Build GRCh38

View Full Product Details