Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 185124737-185124986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT1L
Entrez Gene ID
81627
Gene Name
tRNA methyltransferase 1 like
Gene Aliases
C1orf25, MST070, MSTP070, TRM1L, bG120K12.3
UniGene
Hs.107149
Gene Chromosome Location
Chr.1: 185118085-185157098 on Build GRCh38
Transcripts
NM_001202423.1 NM_030934.4 XM_011510028.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3984547 c.1717A>T p.S573C Chr.1: 185124986-185124986 on Build GRCh38
5597761 c.1727C>T p.S576L Chr.1: 185124976-185124976 on Build GRCh38
3943204 c.1725T>A p.F575L Chr.1: 185124978-185124978 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754637987 Chr.1: 185124808-185124808 on Build GRCh38
rs754234562 Chr.1: 185124930-185124930 on Build GRCh38
rs992359184 Chr.1: 185124785-185124786 on Build GRCh38
rs765658887 Chr.1: 185124939-185124939 on Build GRCh38
rs892642601 Chr.1: 185124853-185124853 on Build GRCh38
rs761220869 Chr.1: 185124900-185124900 on Build GRCh38
rs142906342 Chr.1: 185124952-185124952 on Build GRCh38
rs757875804 Chr.1: 185124931-185124931 on Build GRCh38
rs766807336 Chr.1: 185124926-185124926 on Build GRCh38
rs561844404 Chr.1: 185124821-185124821 on Build GRCh38
rs901657176 Chr.1: 185124839-185124839 on Build GRCh38
rs371190970 Chr.1: 185124912-185124912 on Build GRCh38
rs760407346 Chr.1: 185124924-185124924 on Build GRCh38
rs540888959 Chr.1: 185124936-185124936 on Build GRCh38
rs939629916 Chr.1: 185124776-185124776 on Build GRCh38
rs200485806 Chr.1: 185124948-185124948 on Build GRCh38
rs778559520 Chr.1: 185124850-185124850 on Build GRCh38
rs948075727 Chr.1: 185124791-185124791 on Build GRCh38
rs774382822 Chr.1: 185124899-185124899 on Build GRCh38
rs1050347088 Chr.1: 185124883-185124883 on Build GRCh38
rs767113110 Chr.1: 185124901-185124901 on Build GRCh38
rs573744214 Chr.1: 185124861-185124861 on Build GRCh38
rs543694675 Chr.1: 185124794-185124794 on Build GRCh38
rs556821716 Chr.1: 185124935-185124935 on Build GRCh38
rs752697581 Chr.1: 185124926-185124927 on Build GRCh38
rs903942302 Chr.1: 185124796-185124796 on Build GRCh38

View Full Product Details