Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 58346837-58347110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A1BG
Entrez Gene ID
1
Gene Name
alpha-1-B glycoprotein
Gene Aliases
A1B, ABG, GAB, HYST2477
UniGene
Hs.529161
Gene Chromosome Location
Chr.19: 58346806-58353499 on Build GRCh38
Transcripts
NM_130786.3

SNP Detail

SNP ID Chromosome Location
rs757346192 Chr.19: 58347018-58347018 on Build GRCh38
rs917961071 Chr.19: 58347082-58347082 on Build GRCh38
rs949404683 Chr.19: 58347089-58347089 on Build GRCh38
rs1047461 Chr.19: 58346970-58346970 on Build GRCh38
rs760105773 Chr.19: 58346997-58346997 on Build GRCh38
rs557377182 Chr.19: 58347078-58347078 on Build GRCh38
rs765925878 Chr.19: 58346987-58346987 on Build GRCh38
rs200495425 Chr.19: 58346865-58346865 on Build GRCh38
rs996930699 Chr.19: 58346954-58346954 on Build GRCh38
rs981385982 Chr.19: 58347090-58347090 on Build GRCh38
rs200870181 Chr.19: 58347011-58347011 on Build GRCh38
rs529512912 Chr.19: 58346985-58346985 on Build GRCh38
rs753760765 Chr.19: 58347031-58347031 on Build GRCh38
rs376213403 Chr.19: 58347080-58347080 on Build GRCh38
rs892648783 Chr.19: 58346918-58346918 on Build GRCh38
rs200801249 Chr.19: 58346982-58346982 on Build GRCh38
rs367914795 Chr.19: 58346931-58346931 on Build GRCh38
rs753125623 Chr.19: 58347000-58347000 on Build GRCh38
rs202132547 Chr.19: 58346872-58346872 on Build GRCh38
rs764342582 Chr.19: 58347004-58347004 on Build GRCh38
rs398035147 Chr.19: 58346867-58346867 on Build GRCh38
rs772682513 Chr.19: 58347044-58347045 on Build GRCh38
rs570220357 Chr.19: 58346967-58346967 on Build GRCh38
rs373324898 Chr.19: 58347038-58347038 on Build GRCh38
rs550459885 Chr.19: 58346956-58346956 on Build GRCh38
rs777113256 Chr.19: 58346989-58346989 on Build GRCh38
rs955393076 Chr.19: 58346960-58346960 on Build GRCh38
rs372964699 Chr.19: 58347035-58347035 on Build GRCh38
rs777463133 Chr.19: 58347042-58347042 on Build GRCh38
rs764893172 Chr.19: 58346976-58346976 on Build GRCh38
rs1022566101 Chr.19: 58346948-58346948 on Build GRCh38
rs376165835 Chr.19: 58346974-58346974 on Build GRCh38
rs770425278 Chr.19: 58347062-58347062 on Build GRCh38
rs776125888 Chr.19: 58347066-58347066 on Build GRCh38
rs376202936 Chr.19: 58347039-58347039 on Build GRCh38
rs565471556 Chr.19: 58347029-58347029 on Build GRCh38
rs370242476 Chr.19: 58347049-58347049 on Build GRCh38
rs369421999 Chr.19: 58347001-58347001 on Build GRCh38
rs758148429 Chr.19: 58346903-58346903 on Build GRCh38
rs111251782 Chr.19: 58347032-58347032 on Build GRCh38
rs771803346 Chr.19: 58347041-58347041 on Build GRCh38
rs760248960 Chr.19: 58347066-58347090 on Build GRCh38
rs533320081 Chr.19: 58346952-58346952 on Build GRCh38
rs765524646 Chr.19: 58346999-58346999 on Build GRCh38
rs3214663 Chr.19: 58346866-58346866 on Build GRCh38
rs772558287 Chr.19: 58346981-58346981 on Build GRCh38

View Full Product Details