Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 51965140-51965409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF350
Entrez Gene ID
59348
Gene Name
zinc finger protein 350
Gene Aliases
ZBRK1, ZFQR
UniGene
Hs.407694
Gene Chromosome Location
Chr.19: 51964340-51986876 on Build GRCh38
Transcripts
NM_021632.3 XM_011527196.1 XM_017027094.1 XM_017027095.1 XM_017027096.1 XM_017027097.1 XM_017027098.1 XM_017027099.1 XM_017027100.1 XM_017027101.1 XM_017027102.1 XM_017027103.1 XM_017027104.1 NR_103847.1

Gene Symbol
ZNF350-AS1
Entrez Gene ID
101669766
Gene Name
ZNF350 antisense RNA 1
Gene Aliases
HCCAT3, uc002pyc
UniGene
Hs.637744
Gene Chromosome Location
Chr.19: 51949134-51981367 on Build GRCh38
Transcripts
NM_021632.3 XM_011527196.1 XM_017027094.1 XM_017027095.1 XM_017027096.1 XM_017027097.1 XM_017027098.1 XM_017027099.1 XM_017027100.1 XM_017027101.1 XM_017027102.1 XM_017027103.1 XM_017027104.1 NR_103847.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4459282 c.1119C>T p.P373P Chr.19: 51965334-51965334 on Build GRCh38
4524541 c.1278G>A p.R426R Chr.19: 51965175-51965175 on Build GRCh38
1000264 c.1298A>C p.K433T Chr.19: 51965155-51965155 on Build GRCh38
1000266 c.1153A>G p.T385A Chr.19: 51965300-51965300 on Build GRCh38
1000268 c.1097G>T p.R366I Chr.19: 51965356-51965356 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772535085 Chr.19: 51965256-51965256 on Build GRCh38
rs778853227 Chr.19: 51965245-51965245 on Build GRCh38
rs771994012 Chr.19: 51965224-51965224 on Build GRCh38
rs751677921 Chr.19: 51965235-51965236 on Build GRCh38
rs753015381 Chr.19: 51965164-51965166 on Build GRCh38
rs4988335 Chr.19: 51965274-51965274 on Build GRCh38
rs772739547 Chr.19: 51965257-51965257 on Build GRCh38
rs757355164 Chr.19: 51965240-51965241 on Build GRCh38
rs780033650 Chr.19: 51965345-51965345 on Build GRCh38
rs779901758 Chr.19: 51965233-51965233 on Build GRCh38
rs781065955 Chr.19: 51965359-51965359 on Build GRCh38
rs778067618 Chr.19: 51965168-51965168 on Build GRCh38
rs768047677 Chr.19: 51965269-51965269 on Build GRCh38
rs767942276 Chr.19: 51965179-51965179 on Build GRCh38
rs769439995 Chr.19: 51965369-51965369 on Build GRCh38
rs3764539 Chr.19: 51965237-51965237 on Build GRCh38
rs150778438 Chr.19: 51965248-51965248 on Build GRCh38
rs776408816 Chr.19: 51965243-51965243 on Build GRCh38
rs747985668 Chr.19: 51965368-51965368 on Build GRCh38
rs760020329 Chr.19: 51965176-51965176 on Build GRCh38
rs569458587 Chr.19: 51965308-51965308 on Build GRCh38
rs754073345 Chr.19: 51965166-51965167 on Build GRCh38
rs975049311 Chr.19: 51965192-51965192 on Build GRCh38
rs759806410 Chr.19: 51965238-51965238 on Build GRCh38
rs148753740 Chr.19: 51965208-51965208 on Build GRCh38
rs1046484041 Chr.19: 51965273-51965273 on Build GRCh38
rs754130782 Chr.19: 51965211-51965211 on Build GRCh38
rs781765781 Chr.19: 51965362-51965362 on Build GRCh38
rs776636199 Chr.19: 51965338-51965338 on Build GRCh38
rs571896047 Chr.19: 51965267-51965267 on Build GRCh38
rs753058181 Chr.19: 51965202-51965202 on Build GRCh38
rs4986772 Chr.19: 51965298-51965298 on Build GRCh38
rs566120255 Chr.19: 51965356-51965356 on Build GRCh38
rs761027666 Chr.19: 51965282-51965282 on Build GRCh38
rs3764538 Chr.19: 51965334-51965334 on Build GRCh38
rs371531041 Chr.19: 51965223-51965223 on Build GRCh38
rs779023028 Chr.19: 51965330-51965330 on Build GRCh38
rs1018920159 Chr.19: 51965384-51965384 on Build GRCh38
rs61762979 Chr.19: 51965217-51965217 on Build GRCh38
rs945552113 Chr.19: 51965264-51965264 on Build GRCh38
rs113072853 Chr.19: 51965309-51965309 on Build GRCh38
rs777940934 Chr.19: 51965209-51965209 on Build GRCh38
rs757319872 Chr.19: 51965215-51965215 on Build GRCh38
rs200843965 Chr.19: 51965254-51965254 on Build GRCh38
rs867884219 Chr.19: 51965239-51965240 on Build GRCh38
rs768639393 Chr.19: 51965240-51965240 on Build GRCh38

View Full Product Details