Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1986925-1987186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD2
Entrez Gene ID
55643
Gene Name
BTB domain containing 2
Gene Aliases
-
UniGene
Hs.465543
Gene Chromosome Location
Chr.19: 1985438-2015703 on Build GRCh38
Transcripts
NM_017797.3 XM_005259593.1 XM_011528127.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7095674 c.1276C>G p.H426D Chr.19: 1986970-1986970 on Build GRCh38
3989857 c.1287C>T p.S429S Chr.19: 1986959-1986959 on Build GRCh38
3783211 c.1317C>T p.G439G Chr.19: 1986929-1986929 on Build GRCh38
4444660 c.1313C>T p.T438M Chr.19: 1986933-1986933 on Build GRCh38
5389188 c.1282G>A p.D428N Chr.19: 1986964-1986964 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540696210 Chr.19: 1987012-1987012 on Build GRCh38
rs200226391 Chr.19: 1987159-1987159 on Build GRCh38
rs540076203 Chr.19: 1987154-1987154 on Build GRCh38
rs781276510 Chr.19: 1987020-1987020 on Build GRCh38
rs776479719 Chr.19: 1986981-1986981 on Build GRCh38
rs371639988 Chr.19: 1987029-1987029 on Build GRCh38
rs138467239 Chr.19: 1987124-1987124 on Build GRCh38
rs766978404 Chr.19: 1986970-1986970 on Build GRCh38
rs764728850 Chr.19: 1987004-1987004 on Build GRCh38
rs762131393 Chr.19: 1986962-1986962 on Build GRCh38
rs548147289 Chr.19: 1987125-1987125 on Build GRCh38
rs747145889 Chr.19: 1986955-1986955 on Build GRCh38
rs761223048 Chr.19: 1987129-1987129 on Build GRCh38
rs920007441 Chr.19: 1987032-1987032 on Build GRCh38
rs764995572 Chr.19: 1987138-1987138 on Build GRCh38
rs746913630 Chr.19: 1987013-1987013 on Build GRCh38
rs751310183 Chr.19: 1987155-1987155 on Build GRCh38
rs765313138 Chr.19: 1986965-1986965 on Build GRCh38
rs1044638798 Chr.19: 1987083-1987083 on Build GRCh38
rs748119907 Chr.19: 1987078-1987078 on Build GRCh38
rs370801177 Chr.19: 1987016-1987016 on Build GRCh38
rs370739634 Chr.19: 1987024-1987024 on Build GRCh38
rs778790644 Chr.19: 1986948-1986948 on Build GRCh38
rs1056651196 Chr.19: 1987073-1987073 on Build GRCh38
rs370012025 Chr.19: 1986981-1986981 on Build GRCh38
rs367654379 Chr.19: 1987120-1987120 on Build GRCh38
rs923779439 Chr.19: 1987034-1987034 on Build GRCh38
rs117688255 Chr.19: 1986998-1986998 on Build GRCh38
rs896150712 Chr.19: 1987074-1987074 on Build GRCh38
rs201662565 Chr.19: 1987025-1987025 on Build GRCh38
rs745589915 Chr.19: 1986949-1986949 on Build GRCh38
rs772049204 Chr.19: 1986952-1986952 on Build GRCh38
rs947734938 Chr.19: 1987076-1987076 on Build GRCh38
rs566370769 Chr.19: 1987146-1987146 on Build GRCh38
rs955391242 Chr.19: 1987144-1987144 on Build GRCh38
rs776882173 Chr.19: 1986957-1986957 on Build GRCh38
rs756365423 Chr.19: 1987084-1987084 on Build GRCh38
rs763163030 Chr.19: 1986968-1986968 on Build GRCh38
rs1030197342 Chr.19: 1987128-1987128 on Build GRCh38
rs972813833 Chr.19: 1986982-1986982 on Build GRCh38
rs763728213 Chr.19: 1986994-1986994 on Build GRCh38
rs750295923 Chr.19: 1987007-1987007 on Build GRCh38
rs902710418 Chr.19: 1987098-1987098 on Build GRCh38
rs754925188 Chr.19: 1987158-1987158 on Build GRCh38
rs373856064 Chr.19: 1986996-1986996 on Build GRCh38
rs368441913 Chr.19: 1987143-1987143 on Build GRCh38
rs376970283 Chr.19: 1986964-1986964 on Build GRCh38
rs1017074058 Chr.19: 1987123-1987123 on Build GRCh38
rs931307069 Chr.19: 1987035-1987035 on Build GRCh38
rs199975886 Chr.19: 1986956-1986956 on Build GRCh38
rs753349156 Chr.19: 1986997-1986997 on Build GRCh38
rs377213437 Chr.19: 1987021-1987021 on Build GRCh38
rs768079814 Chr.19: 1987127-1987127 on Build GRCh38
rs760311435 Chr.19: 1986983-1986983 on Build GRCh38
rs759304224 Chr.19: 1987137-1987149 on Build GRCh38
rs201109257 Chr.19: 1987119-1987119 on Build GRCh38
rs770062327 Chr.19: 1987023-1987023 on Build GRCh38
rs935048581 Chr.19: 1987058-1987058 on Build GRCh38
rs540962872 Chr.19: 1986953-1986953 on Build GRCh38
rs916504513 Chr.19: 1986986-1986986 on Build GRCh38
rs758112535 Chr.19: 1987011-1987011 on Build GRCh38
rs1017126915 Chr.19: 1986951-1986951 on Build GRCh38

View Full Product Details