Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131359385-131359656 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC42SE2
Entrez Gene ID
56990
Gene Name
CDC42 small effector 2
Gene Aliases
SPEC2
UniGene
Hs.508829
Gene Chromosome Location
Chr.5: 131264009-131394690 on Build GRCh38
Transcripts
NM_001038702.1 NM_020240.2 XM_017009648.1 XM_017009649.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4628355 c.45G>A p.Q15Q Chr.5: 131359538-131359538 on Build GRCh38
6624441 c.52C>T p.P18S Chr.5: 131359545-131359545 on Build GRCh38
3136664 c.22T>C p.F8L Chr.5: 131359515-131359515 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs557923001 Chr.5: 131359420-131359420 on Build GRCh38
rs376654083 Chr.5: 131359569-131359569 on Build GRCh38
rs1017237571 Chr.5: 131359620-131359620 on Build GRCh38
rs777962585 Chr.5: 131359464-131359464 on Build GRCh38
rs755427236 Chr.5: 131359559-131359559 on Build GRCh38
rs199987880 Chr.5: 131359537-131359537 on Build GRCh38
rs773950759 Chr.5: 131359532-131359532 on Build GRCh38
rs767044363 Chr.5: 131359444-131359444 on Build GRCh38
rs778142261 Chr.5: 131359584-131359584 on Build GRCh38
rs138505481 Chr.5: 131359488-131359488 on Build GRCh38
rs755337482 Chr.5: 131359456-131359456 on Build GRCh38
rs767176791 Chr.5: 131359619-131359619 on Build GRCh38
rs771254022 Chr.5: 131359467-131359467 on Build GRCh38
rs934263022 Chr.5: 131359576-131359576 on Build GRCh38
rs200700138 Chr.5: 131359475-131359475 on Build GRCh38
rs762827850 Chr.5: 131359504-131359504 on Build GRCh38
rs761223026 Chr.5: 131359541-131359541 on Build GRCh38
rs369822334 Chr.5: 131359442-131359442 on Build GRCh38
rs535034160 Chr.5: 131359414-131359414 on Build GRCh38
rs11951274 Chr.5: 131359459-131359459 on Build GRCh38
rs111259308 Chr.5: 131359491-131359491 on Build GRCh38
rs781477382 Chr.5: 131359597-131359597 on Build GRCh38
rs754294772 Chr.5: 131359455-131359455 on Build GRCh38
rs747175072 Chr.5: 131359588-131359588 on Build GRCh38
rs776701914 Chr.5: 131359468-131359468 on Build GRCh38
rs763995890 Chr.5: 131359531-131359531 on Build GRCh38
rs202042579 Chr.5: 131359465-131359465 on Build GRCh38
rs373744699 Chr.5: 131359562-131359562 on Build GRCh38
rs572384835 Chr.5: 131359410-131359410 on Build GRCh38
rs963216768 Chr.5: 131359412-131359412 on Build GRCh38
rs966808253 Chr.5: 131359580-131359580 on Build GRCh38
rs542241478 Chr.5: 131359558-131359558 on Build GRCh38
rs758573670 Chr.5: 131359461-131359461 on Build GRCh38
rs990956526 Chr.5: 131359473-131359473 on Build GRCh38
rs890168773 Chr.5: 131359602-131359602 on Build GRCh38
rs745673297 Chr.5: 131359472-131359472 on Build GRCh38
rs376985674 Chr.5: 131359443-131359443 on Build GRCh38
rs1037528654 Chr.5: 131359417-131359417 on Build GRCh38
rs757223623 Chr.5: 131359596-131359596 on Build GRCh38
rs767213563 Chr.5: 131359554-131359554 on Build GRCh38
rs748627066 Chr.5: 131359460-131359460 on Build GRCh38
rs902657296 Chr.5: 131359571-131359571 on Build GRCh38

View Full Product Details