Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 75597966-75598197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKL2
Entrez Gene ID
8999
Gene Name
cyclin dependent kinase like 2
Gene Aliases
KKIAMRE, P56
UniGene
Hs.591698
Gene Chromosome Location
Chr.4: 75576494-75630537 on Build GRCh38
Transcripts
NM_003948.3 XM_005263335.2 XM_006714406.2 XM_017008809.1 XM_017008810.1 XM_017008811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1683994 c.943delA p.K316fs*15 Chr.4: 75598154-75598154 on Build GRCh38
159915 c.1001G>C p.R334T Chr.4: 75598096-75598096 on Build GRCh38
734682 c.939A>G p.L313L Chr.4: 75598158-75598158 on Build GRCh38
4671200 c.978_979insA p.D327fs*2 Chr.4: 75598118-75598119 on Build GRCh38
346763 c.1009C>A p.L337I Chr.4: 75598088-75598088 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112876166 Chr.4: 75598102-75598102 on Build GRCh38
rs767763721 Chr.4: 75598080-75598080 on Build GRCh38
rs897013910 Chr.4: 75598112-75598112 on Build GRCh38
rs574511380 Chr.4: 75598012-75598012 on Build GRCh38
rs112159909 Chr.4: 75598015-75598015 on Build GRCh38
rs368281660 Chr.4: 75598026-75598026 on Build GRCh38
rs769934187 Chr.4: 75598036-75598036 on Build GRCh38
rs762226449 Chr.4: 75598167-75598167 on Build GRCh38
rs746747634 Chr.4: 75598125-75598125 on Build GRCh38
rs762436003 Chr.4: 75598072-75598072 on Build GRCh38
rs764266900 Chr.4: 75598051-75598051 on Build GRCh38
rs778530783 Chr.4: 75598111-75598111 on Build GRCh38
rs773049339 Chr.4: 75598029-75598029 on Build GRCh38
rs578201781 Chr.4: 75598018-75598018 on Build GRCh38
rs775704693 Chr.4: 75598037-75598037 on Build GRCh38
rs1000643761 Chr.4: 75598149-75598149 on Build GRCh38
rs746084953 Chr.4: 75598030-75598030 on Build GRCh38
rs369392841 Chr.4: 75598140-75598140 on Build GRCh38
rs531033737 Chr.4: 75598166-75598166 on Build GRCh38
rs766077540 Chr.4: 75598090-75598090 on Build GRCh38
rs768596365 Chr.4: 75598160-75598160 on Build GRCh38
rs901849456 Chr.4: 75597995-75597995 on Build GRCh38
rs375761704 Chr.4: 75598083-75598083 on Build GRCh38
rs376042977 Chr.4: 75598123-75598123 on Build GRCh38
rs372845187 Chr.4: 75598041-75598041 on Build GRCh38
rs778563667 Chr.4: 75598029-75598030 on Build GRCh38
rs758426435 Chr.4: 75598118-75598118 on Build GRCh38
rs774689477 Chr.4: 75598069-75598069 on Build GRCh38
rs770737634 Chr.4: 75598135-75598135 on Build GRCh38
rs1008947362 Chr.4: 75598078-75598078 on Build GRCh38
rs200524804 Chr.4: 75598117-75598117 on Build GRCh38
rs750887905 Chr.4: 75598082-75598082 on Build GRCh38
rs749565596 Chr.4: 75598155-75598155 on Build GRCh38
rs563761502 Chr.4: 75598100-75598100 on Build GRCh38
rs1043227369 Chr.4: 75597999-75597999 on Build GRCh38
rs997414477 Chr.4: 75598009-75598009 on Build GRCh38
rs185183992 Chr.4: 75598034-75598034 on Build GRCh38
rs754411698 Chr.4: 75598110-75598110 on Build GRCh38

View Full Product Details