Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 48041199-48041460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf57
Entrez Gene ID
136288
Gene Name
chromosome 7 open reading frame 57
Gene Aliases
-
UniGene
Hs.258357
Gene Chromosome Location
Chr.7: 48035511-48061297 on Build GRCh38
Transcripts
NM_001100159.2 NM_001267865.1 NM_001267866.1 XM_006715649.3 XM_006715650.2 XM_011515121.1 XM_011515122.2 XM_011515123.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3264003 c.83G>A p.R28H Chr.7: 48041361-48041361 on Build GRCh38
5429390 c.153C>T p.H51H Chr.7: 48041431-48041431 on Build GRCh38
3721087 c.123C>T p.I41I Chr.7: 48041401-48041401 on Build GRCh38
1090210 c.170G>T p.G57V Chr.7: 48041448-48041448 on Build GRCh38
1090208 c.113C>T p.A38V Chr.7: 48041391-48041391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374151689 Chr.7: 48041291-48041291 on Build GRCh38
rs763237753 Chr.7: 48041411-48041414 on Build GRCh38
rs986320809 Chr.7: 48041336-48041336 on Build GRCh38
rs746538863 Chr.7: 48041413-48041413 on Build GRCh38
rs781493956 Chr.7: 48041379-48041379 on Build GRCh38
rs543935129 Chr.7: 48041316-48041316 on Build GRCh38
rs754948160 Chr.7: 48041377-48041377 on Build GRCh38
rs750211328 Chr.7: 48041369-48041369 on Build GRCh38
rs758457812 Chr.7: 48041371-48041371 on Build GRCh38
rs528667180 Chr.7: 48041414-48041414 on Build GRCh38
rs1026929084 Chr.7: 48041286-48041286 on Build GRCh38
rs761722347 Chr.7: 48041366-48041366 on Build GRCh38
rs376817019 Chr.7: 48041352-48041352 on Build GRCh38
rs367644498 Chr.7: 48041360-48041360 on Build GRCh38
rs761595651 Chr.7: 48041436-48041436 on Build GRCh38
rs776085868 Chr.7: 48041362-48041362 on Build GRCh38
rs769546871 Chr.7: 48041441-48041441 on Build GRCh38
rs779217694 Chr.7: 48041405-48041405 on Build GRCh38
rs778154007 Chr.7: 48041387-48041387 on Build GRCh38
rs368589497 Chr.7: 48041319-48041319 on Build GRCh38
rs200433005 Chr.7: 48041361-48041361 on Build GRCh38
rs374929652 Chr.7: 48041435-48041435 on Build GRCh38
rs748686416 Chr.7: 48041245-48041245 on Build GRCh38
rs756505268 Chr.7: 48041382-48041382 on Build GRCh38
rs749636603 Chr.7: 48041391-48041391 on Build GRCh38
rs202135709 Chr.7: 48041375-48041375 on Build GRCh38
rs755286795 Chr.7: 48041292-48041292 on Build GRCh38
rs748325661 Chr.7: 48041381-48041381 on Build GRCh38
rs770090055 Chr.7: 48041317-48041317 on Build GRCh38
rs773734741 Chr.7: 48041330-48041330 on Build GRCh38
rs868549959 Chr.7: 48041357-48041357 on Build GRCh38
rs750336180 Chr.7: 48041288-48041288 on Build GRCh38
rs771533681 Chr.7: 48041392-48041392 on Build GRCh38
rs530145864 Chr.7: 48041305-48041305 on Build GRCh38
rs766215119 Chr.7: 48041374-48041374 on Build GRCh38
rs955009936 Chr.7: 48041321-48041321 on Build GRCh38
rs1044918186 Chr.7: 48041417-48041417 on Build GRCh38
rs758581366 Chr.7: 48041290-48041290 on Build GRCh38
rs937938608 Chr.7: 48041284-48041284 on Build GRCh38
rs17132050 Chr.7: 48041347-48041347 on Build GRCh38
rs775121954 Chr.7: 48041356-48041356 on Build GRCh38

View Full Product Details