Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 93075811-93076040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUSD3
Entrez Gene ID
203328
Gene Name
sushi domain containing 3
Gene Aliases
-
UniGene
Hs.88417
Gene Chromosome Location
Chr.9: 93058688-93085138 on Build GRCh38
Transcripts
NM_001287005.1 NM_001287006.1 NM_001287007.1 NM_001287008.1 NM_145006.3 XM_011518358.1 XM_011518359.1 XM_011518361.2 XM_011518362.1 XM_017014448.1 XM_017014449.1 XM_017014450.1 XM_017014451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4730973 c.277C>T p.L93L Chr.9: 93075972-93075972 on Build GRCh38
6793399 c.85A>G p.T29A Chr.9: 93075819-93075819 on Build GRCh38
72855 c.125C>A p.T42N Chr.9: 93075820-93075820 on Build GRCh38
4817474 c.247G>C p.E83Q Chr.9: 93075942-93075942 on Build GRCh38
6793400 c.77C>T p.P26L Chr.9: 93075811-93075811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746959146 Chr.9: 93075866-93075866 on Build GRCh38
rs770573104 Chr.9: 93076014-93076014 on Build GRCh38
rs200999861 Chr.9: 93075886-93075886 on Build GRCh38
rs141762113 Chr.9: 93075973-93075973 on Build GRCh38
rs762564066 Chr.9: 93076013-93076013 on Build GRCh38
rs557818939 Chr.9: 93075851-93075851 on Build GRCh38
rs201944177 Chr.9: 93075874-93075874 on Build GRCh38
rs753066803 Chr.9: 93075949-93075949 on Build GRCh38
rs200817329 Chr.9: 93075985-93075985 on Build GRCh38
rs531952596 Chr.9: 93075834-93075834 on Build GRCh38
rs774484881 Chr.9: 93075860-93075860 on Build GRCh38
rs775992481 Chr.9: 93075982-93075982 on Build GRCh38
rs756362131 Chr.9: 93075852-93075852 on Build GRCh38
rs769767174 Chr.9: 93075999-93075999 on Build GRCh38
rs768029547 Chr.9: 93075939-93075939 on Build GRCh38
rs377039386 Chr.9: 93075965-93075965 on Build GRCh38
rs901289492 Chr.9: 93075996-93075996 on Build GRCh38
rs966981605 Chr.9: 93075904-93075904 on Build GRCh38
rs369622588 Chr.9: 93075938-93075938 on Build GRCh38
rs768614143 Chr.9: 93075868-93075868 on Build GRCh38
rs765851118 Chr.9: 93075888-93075888 on Build GRCh38
rs773113490 Chr.9: 93076005-93076005 on Build GRCh38
rs752076564 Chr.9: 93075835-93075835 on Build GRCh38
rs771727595 Chr.9: 93075972-93075973 on Build GRCh38
rs776311331 Chr.9: 93075871-93075871 on Build GRCh38
rs370065626 Chr.9: 93075972-93075972 on Build GRCh38
rs11555624 Chr.9: 93075833-93075833 on Build GRCh38
rs1018555193 Chr.9: 93075850-93075850 on Build GRCh38
rs749384937 Chr.9: 93075858-93075858 on Build GRCh38
rs368292441 Chr.9: 93075873-93075873 on Build GRCh38
rs770975398 Chr.9: 93075859-93075859 on Build GRCh38
rs997302832 Chr.9: 93076010-93076010 on Build GRCh38
rs751061029 Chr.9: 93075932-93075932 on Build GRCh38
rs12352602 Chr.9: 93075893-93075893 on Build GRCh38
rs868317847 Chr.9: 93075955-93075955 on Build GRCh38
rs777907895 Chr.9: 93075857-93075857 on Build GRCh38
rs781461989 Chr.9: 93075841-93075841 on Build GRCh38
rs746326390 Chr.9: 93075923-93075923 on Build GRCh38
rs201623385 Chr.9: 93075971-93075971 on Build GRCh38

View Full Product Details