Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 159035849-159036106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAGAP
Entrez Gene ID
117289
Gene Name
T-cell activation RhoGTPase activating protein
Gene Aliases
ARHGAP47, FKSG15, IDDM21, TAGAP1
UniGene
Hs.529984
Gene Chromosome Location
Chr.6: 159034468-159045152 on Build GRCh38
Transcripts
NM_001278733.1 NM_054114.4 NM_152133.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
168678 c.2131C>T p.R711* Chr.6: 159035892-159035892 on Build GRCh38
397321 c.2068G>C p.E690Q Chr.6: 159035955-159035955 on Build GRCh38
6105747 c.2074C>A p.L692I Chr.6: 159035949-159035949 on Build GRCh38
5187847 c.2059G>T p.G687W Chr.6: 159035964-159035964 on Build GRCh38
6796486 c.2127C>T p.L709L Chr.6: 159035896-159035896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762062599 Chr.6: 159036061-159036061 on Build GRCh38
rs767628374 Chr.6: 159035982-159035982 on Build GRCh38
rs776501479 Chr.6: 159035938-159035938 on Build GRCh38
rs200804434 Chr.6: 159036016-159036016 on Build GRCh38
rs779996502 Chr.6: 159036036-159036036 on Build GRCh38
rs769810341 Chr.6: 159036027-159036027 on Build GRCh38
rs745819016 Chr.6: 159036020-159036020 on Build GRCh38
rs556216423 Chr.6: 159036048-159036048 on Build GRCh38
rs148570036 Chr.6: 159036002-159036002 on Build GRCh38
rs201292158 Chr.6: 159035929-159035929 on Build GRCh38
rs770707646 Chr.6: 159035886-159035886 on Build GRCh38
rs746999036 Chr.6: 159035884-159035884 on Build GRCh38
rs762848425 Chr.6: 159036030-159036030 on Build GRCh38
rs751645528 Chr.6: 159035895-159035895 on Build GRCh38
rs772686747 Chr.6: 159035881-159035881 on Build GRCh38
rs759308422 Chr.6: 159035888-159035888 on Build GRCh38
rs781618609 Chr.6: 159035896-159035897 on Build GRCh38
rs746278309 Chr.6: 159036077-159036078 on Build GRCh38
rs770463047 Chr.6: 159035930-159035930 on Build GRCh38
rs775380993 Chr.6: 159035950-159035950 on Build GRCh38
rs559073180 Chr.6: 159035917-159035917 on Build GRCh38
rs764341891 Chr.6: 159035963-159035963 on Build GRCh38
rs774501392 Chr.6: 159036057-159036057 on Build GRCh38
rs775898458 Chr.6: 159035891-159035891 on Build GRCh38
rs147907532 Chr.6: 159035944-159035944 on Build GRCh38
rs771608749 Chr.6: 159035878-159035878 on Build GRCh38
rs764287043 Chr.6: 159035893-159035893 on Build GRCh38
rs756620977 Chr.6: 159036001-159036001 on Build GRCh38
rs201728690 Chr.6: 159035897-159035897 on Build GRCh38
rs774498911 Chr.6: 159035971-159035971 on Build GRCh38
rs780364584 Chr.6: 159035914-159035914 on Build GRCh38
rs530252270 Chr.6: 159036009-159036009 on Build GRCh38
rs941713504 Chr.6: 159035889-159035889 on Build GRCh38
rs758060464 Chr.6: 159036008-159036008 on Build GRCh38
rs377118382 Chr.6: 159036081-159036081 on Build GRCh38
rs895068807 Chr.6: 159035935-159035935 on Build GRCh38
rs757784048 Chr.6: 159035901-159035901 on Build GRCh38
rs761854410 Chr.6: 159035974-159035974 on Build GRCh38
rs144830124 Chr.6: 159035906-159035906 on Build GRCh38
rs756500896 Chr.6: 159035907-159035907 on Build GRCh38
rs747793651 Chr.6: 159035876-159035876 on Build GRCh38
rs756759950 Chr.6: 159036014-159036014 on Build GRCh38
rs188045239 Chr.6: 159036005-159036005 on Build GRCh38
rs866358245 Chr.6: 159036080-159036080 on Build GRCh38
rs767852044 Chr.6: 159035904-159035904 on Build GRCh38
rs768555725 Chr.6: 159036054-159036054 on Build GRCh38
rs747847015 Chr.6: 159035918-159035918 on Build GRCh38
rs769763460 Chr.6: 159035947-159035947 on Build GRCh38
rs777270446 Chr.6: 159035928-159035928 on Build GRCh38
rs763294731 Chr.6: 159035960-159035960 on Build GRCh38
rs750932465 Chr.6: 159035986-159035986 on Build GRCh38
rs922983390 Chr.6: 159035879-159035879 on Build GRCh38
rs758007058 Chr.6: 159035922-159035922 on Build GRCh38
rs999490089 Chr.6: 159036073-159036073 on Build GRCh38
rs561705660 Chr.6: 159036028-159036028 on Build GRCh38

View Full Product Details