Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 47238867-47239138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMYND8
Entrez Gene ID
23613
Gene Name
zinc finger MYND-type containing 8
Gene Aliases
PRKCBP1, PRO2893, RACK7
UniGene
Hs.446240
Gene Chromosome Location
Chr.20: 47209214-47357780 on Build GRCh38
Transcripts
NM_001281771.2 NM_001281772.2 NM_001281773.2 NM_001281774.2 NM_001281775.2 NM_001281776.2 NM_001281777.2 NM_001281778.2 NM_001281779.2 NM_001281780.2 NM_001281781.2 NM_001281782.2 NM_001281783.2 NM_001281784.2 NM_012408.5 NM_183047.3 NM_183048.3 XM_005260356.4 XM_005260358.2 XM_005260360.4 XM_005260362.2 XM_005260364.2 XM_005260366.2 XM_006723760.3 XM_006723762.3 XM_006723763.1 XM_006723766.1 XM_011528749.2 XM_011528751.1 XM_011528752.1 XM_011528753.1 XM_011528754.1 XM_017027760.1 XM_017027761.1 XM_017027762.1 XM_017027763.1 XM_017027764.1 XM_017027765.1 XM_017027766.1 XM_017027767.1 XM_017027768.1 XM_017027769.1 XM_017027770.1 XM_017027771.1 XM_017027772.1 XM_017027773.1 XM_017027774.1 XM_017027775.1 XM_017027776.1 XM_017027777.1 XM_017027778.1 XM_017027779.1 XM_017027780.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5568867 c.2546C>G p.S849* Chr.20: 47238898-47238898 on Build GRCh38
6323729 c.2576G>A p.W859* Chr.20: 47238868-47238868 on Build GRCh38
3911514 c.2299C>T p.P767S Chr.20: 47239124-47239124 on Build GRCh38
7081120 c.2515C>A p.P839T Chr.20: 47238929-47238929 on Build GRCh38
5577219 c.2450C>T p.P817L Chr.20: 47238994-47238994 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778692984 Chr.20: 47238897-47238899 on Build GRCh38
rs748539408 Chr.20: 47238987-47238988 on Build GRCh38
rs767148826 Chr.20: 47239111-47239111 on Build GRCh38
rs141632187 Chr.20: 47239002-47239002 on Build GRCh38
rs747422979 Chr.20: 47238936-47238936 on Build GRCh38
rs756560198 Chr.20: 47238999-47238999 on Build GRCh38
rs145926250 Chr.20: 47238906-47238906 on Build GRCh38
rs138857366 Chr.20: 47238924-47238924 on Build GRCh38
rs769574932 Chr.20: 47239094-47239094 on Build GRCh38
rs761740673 Chr.20: 47239024-47239024 on Build GRCh38
rs761801914 Chr.20: 47238954-47238954 on Build GRCh38
rs764492353 Chr.20: 47239071-47239071 on Build GRCh38
rs771098315 Chr.20: 47239005-47239005 on Build GRCh38
rs777669089 Chr.20: 47238919-47238919 on Build GRCh38
rs552048788 Chr.20: 47239081-47239081 on Build GRCh38
rs777043855 Chr.20: 47239048-47239048 on Build GRCh38
rs776519050 Chr.20: 47239020-47239020 on Build GRCh38
rs1006081568 Chr.20: 47239114-47239114 on Build GRCh38
rs768512649 Chr.20: 47238951-47238951 on Build GRCh38
rs961716825 Chr.20: 47238942-47238942 on Build GRCh38
rs779095508 Chr.20: 47239083-47239083 on Build GRCh38
rs770799455 Chr.20: 47239106-47239106 on Build GRCh38
rs781498589 Chr.20: 47238997-47238997 on Build GRCh38
rs754633888 Chr.20: 47238994-47238994 on Build GRCh38
rs1036519346 Chr.20: 47238940-47238940 on Build GRCh38
rs143086919 Chr.20: 47239110-47239110 on Build GRCh38
rs749211117 Chr.20: 47239105-47239105 on Build GRCh38
rs751156664 Chr.20: 47238991-47238991 on Build GRCh38
rs373180222 Chr.20: 47239018-47239018 on Build GRCh38
rs755395313 Chr.20: 47238910-47238910 on Build GRCh38
rs1030122395 Chr.20: 47238968-47238968 on Build GRCh38
rs765241161 Chr.20: 47238984-47238984 on Build GRCh38
rs2275801 Chr.20: 47239108-47239108 on Build GRCh38
rs773885950 Chr.20: 47239051-47239051 on Build GRCh38
rs753087305 Chr.20: 47238998-47238998 on Build GRCh38
rs538294214 Chr.20: 47238952-47238952 on Build GRCh38
rs769182077 Chr.20: 47238983-47238983 on Build GRCh38
rs774699036 Chr.20: 47238933-47238933 on Build GRCh38
rs765936429 Chr.20: 47238989-47238989 on Build GRCh38
rs911440102 Chr.20: 47238993-47238993 on Build GRCh38
rs770713974 Chr.20: 47239089-47239089 on Build GRCh38
rs377498368 Chr.20: 47239010-47239010 on Build GRCh38
rs377591219 Chr.20: 47239077-47239077 on Build GRCh38
rs146961945 Chr.20: 47238935-47238935 on Build GRCh38
rs772889505 Chr.20: 47239100-47239100 on Build GRCh38
rs376126311 Chr.20: 47238963-47238963 on Build GRCh38
rs773261004 Chr.20: 47239039-47239039 on Build GRCh38
rs376495893 Chr.20: 47238988-47238988 on Build GRCh38
rs762813377 Chr.20: 47239041-47239041 on Build GRCh38
rs746018284 Chr.20: 47239015-47239015 on Build GRCh38
rs779796771 Chr.20: 47238899-47238899 on Build GRCh38
rs550094860 Chr.20: 47239113-47239113 on Build GRCh38
rs749871413 Chr.20: 47238987-47238987 on Build GRCh38
rs769614808 Chr.20: 47239025-47239025 on Build GRCh38
rs377213693 Chr.20: 47239053-47239053 on Build GRCh38
rs756565514 Chr.20: 47239070-47239070 on Build GRCh38
rs369116353 Chr.20: 47239012-47239012 on Build GRCh38
rs778381613 Chr.20: 47239000-47239000 on Build GRCh38
rs913215860 Chr.20: 47239021-47239021 on Build GRCh38
rs201696430 Chr.20: 47239079-47239079 on Build GRCh38
rs1005821395 Chr.20: 47238932-47238932 on Build GRCh38
rs747882482 Chr.20: 47239092-47239092 on Build GRCh38
rs772050242 Chr.20: 47239023-47239023 on Build GRCh38
rs767210378 Chr.20: 47239060-47239060 on Build GRCh38
rs760531197 Chr.20: 47239109-47239109 on Build GRCh38
rs781553445 Chr.20: 47238911-47238911 on Build GRCh38
rs779642631 Chr.20: 47238912-47238912 on Build GRCh38
rs150522803 Chr.20: 47239011-47239011 on Build GRCh38
rs772501682 Chr.20: 47239050-47239050 on Build GRCh38
rs745967382 Chr.20: 47239085-47239085 on Build GRCh38
rs770320871 Chr.20: 47238918-47238918 on Build GRCh38
rs780491504 Chr.20: 47239090-47239090 on Build GRCh38
rs771024245 Chr.20: 47238929-47238929 on Build GRCh38
rs753025764 Chr.20: 47239068-47239068 on Build GRCh38

View Full Product Details