Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94160096-94160339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH17
Entrez Gene ID
1015
Gene Name
cadherin 17
Gene Aliases
CDH16, HPT-1, HPT1
UniGene
Hs.591853
Gene Chromosome Location
Chr.8: 94127162-94217303 on Build GRCh38
Transcripts
NM_001144663.1 NM_004063.3 XM_011516790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5442220 c.1407C>G p.I469M Chr.8: 94160115-94160115 on Build GRCh38
6296462 c.1379C>A p.A460D Chr.8: 94160143-94160143 on Build GRCh38
5988296 c.1378G>T p.A460S Chr.8: 94160144-94160144 on Build GRCh38
454955 c.1371G>C p.L457L Chr.8: 94160151-94160151 on Build GRCh38
6260324 c.1404C>A p.T468T Chr.8: 94160118-94160118 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs574816167 Chr.8: 94160188-94160188 on Build GRCh38
rs543726654 Chr.8: 94160238-94160238 on Build GRCh38
rs918400221 Chr.8: 94160298-94160298 on Build GRCh38
rs759620990 Chr.8: 94160216-94160216 on Build GRCh38
rs1043843928 Chr.8: 94160132-94160132 on Build GRCh38
rs1029709896 Chr.8: 94160230-94160230 on Build GRCh38
rs147833454 Chr.8: 94160155-94160155 on Build GRCh38
rs963168357 Chr.8: 94160240-94160240 on Build GRCh38
rs570967968 Chr.8: 94160289-94160289 on Build GRCh38
rs767887356 Chr.8: 94160168-94160168 on Build GRCh38
rs955517252 Chr.8: 94160234-94160234 on Build GRCh38
rs974269562 Chr.8: 94160245-94160245 on Build GRCh38
rs772372298 Chr.8: 94160210-94160210 on Build GRCh38
rs764863610 Chr.8: 94160129-94160129 on Build GRCh38
rs11785909 Chr.8: 94160259-94160259 on Build GRCh38
rs779047718 Chr.8: 94160203-94160203 on Build GRCh38
rs1007299224 Chr.8: 94160258-94160258 on Build GRCh38
rs962935283 Chr.8: 94160260-94160260 on Build GRCh38
rs370779407 Chr.8: 94160199-94160199 on Build GRCh38
rs11786634 Chr.8: 94160175-94160175 on Build GRCh38
rs749388288 Chr.8: 94160197-94160197 on Build GRCh38
rs1004680057 Chr.8: 94160214-94160214 on Build GRCh38
rs532549673 Chr.8: 94160269-94160269 on Build GRCh38
rs746082345 Chr.8: 94160207-94160207 on Build GRCh38
rs546132791 Chr.8: 94160302-94160302 on Build GRCh38
rs753015799 Chr.8: 94160170-94160170 on Build GRCh38
rs755346799 Chr.8: 94160159-94160159 on Build GRCh38
rs753573389 Chr.8: 94160272-94160272 on Build GRCh38
rs371627823 Chr.8: 94160281-94160281 on Build GRCh38

View Full Product Details