Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27599490-27599761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF512
Entrez Gene ID
84450
Gene Name
zinc finger protein 512
Gene Aliases
-
UniGene
Hs.529178
Gene Chromosome Location
Chr.2: 27582969-27623215 on Build GRCh38
Transcripts
NM_001271286.1 NM_001271287.1 NM_001271288.1 NM_001271289.1 NM_001271318.1 NM_032434.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4804612 c.336C>A p.D112E Chr.2: 27599641-27599641 on Build GRCh38
6091411 c.361C>A p.H121N Chr.2: 27599666-27599666 on Build GRCh38
5481746 c.301G>A p.G101R Chr.2: 27599606-27599606 on Build GRCh38
2940544 c.343G>T p.E115* Chr.2: 27599648-27599648 on Build GRCh38
5479449 c.326A>C p.E109A Chr.2: 27599631-27599631 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773007585 Chr.2: 27599564-27599565 on Build GRCh38
rs776942493 Chr.2: 27599708-27599708 on Build GRCh38
rs746412236 Chr.2: 27599550-27599550 on Build GRCh38
rs765102105 Chr.2: 27599574-27599574 on Build GRCh38
rs772005672 Chr.2: 27599551-27599551 on Build GRCh38
rs969960890 Chr.2: 27599680-27599680 on Build GRCh38
rs760887927 Chr.2: 27599559-27599559 on Build GRCh38
rs958604957 Chr.2: 27599618-27599618 on Build GRCh38
rs771335392 Chr.2: 27599548-27599548 on Build GRCh38
rs780877720 Chr.2: 27599544-27599544 on Build GRCh38
rs370685691 Chr.2: 27599626-27599626 on Build GRCh38
rs374541949 Chr.2: 27599663-27599663 on Build GRCh38
rs775681456 Chr.2: 27599552-27599552 on Build GRCh38
rs1032833056 Chr.2: 27599519-27599519 on Build GRCh38
rs11539165 Chr.2: 27599672-27599672 on Build GRCh38
rs144084742 Chr.2: 27599571-27599571 on Build GRCh38
rs1046811280 Chr.2: 27599694-27599694 on Build GRCh38
rs751052657 Chr.2: 27599598-27599598 on Build GRCh38
rs552997319 Chr.2: 27599527-27599527 on Build GRCh38
rs768987268 Chr.2: 27599569-27599569 on Build GRCh38
rs746559600 Chr.2: 27599538-27599538 on Build GRCh38
rs369461593 Chr.2: 27599596-27599596 on Build GRCh38
rs1022891496 Chr.2: 27599624-27599624 on Build GRCh38
rs780765712 Chr.2: 27599679-27599679 on Build GRCh38
rs768868497 Chr.2: 27599705-27599705 on Build GRCh38
rs373922991 Chr.2: 27599698-27599698 on Build GRCh38
rs749646519 Chr.2: 27599545-27599545 on Build GRCh38
rs370178587 Chr.2: 27599715-27599715 on Build GRCh38
rs1038605026 Chr.2: 27599523-27599523 on Build GRCh38
rs201516895 Chr.2: 27599656-27599656 on Build GRCh38
rs772610605 Chr.2: 27599670-27599670 on Build GRCh38
rs769821161 Chr.2: 27599535-27599535 on Build GRCh38
rs912361260 Chr.2: 27599712-27599712 on Build GRCh38
rs754599261 Chr.2: 27599540-27599540 on Build GRCh38
rs1002650978 Chr.2: 27599734-27599734 on Build GRCh38
rs11539164 Chr.2: 27599587-27599587 on Build GRCh38
rs762587432 Chr.2: 27599658-27599660 on Build GRCh38
rs750286253 Chr.2: 27599577-27599577 on Build GRCh38
rs568111381 Chr.2: 27599529-27599529 on Build GRCh38
rs754546470 Chr.2: 27599608-27599608 on Build GRCh38
rs757518564 Chr.2: 27599645-27599645 on Build GRCh38
rs112688190 Chr.2: 27599623-27599623 on Build GRCh38

View Full Product Details