Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16503238-16503469 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf44
Entrez Gene ID
84167
Gene Name
chromosome 19 open reading frame 44
Gene Aliases
-
UniGene
Hs.631627
Gene Chromosome Location
Chr.19: 16496311-16521369 on Build GRCh38
Transcripts
NM_001288834.1 NM_032207.3 XM_006722920.3 XM_011528354.2 XM_017027350.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4131356 c.1034T>G p.V345G Chr.19: 16503339-16503339 on Build GRCh38
5038442 c.1006T>C p.S336P Chr.19: 16503311-16503311 on Build GRCh38
1283755 c.1038C>G p.D346E Chr.19: 16503343-16503343 on Build GRCh38
992471 c.933G>A p.T311T Chr.19: 16503238-16503238 on Build GRCh38
1304134 c.976T>G p.L326V Chr.19: 16503281-16503281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142233029 Chr.19: 16503377-16503377 on Build GRCh38
rs766766510 Chr.19: 16503399-16503399 on Build GRCh38
rs758075001 Chr.19: 16503274-16503275 on Build GRCh38
rs755096258 Chr.19: 16503341-16503341 on Build GRCh38
rs759035957 Chr.19: 16503271-16503271 on Build GRCh38
rs200689896 Chr.19: 16503395-16503395 on Build GRCh38
rs755744269 Chr.19: 16503339-16503340 on Build GRCh38
rs759693575 Chr.19: 16503427-16503427 on Build GRCh38
rs12461313 Chr.19: 16503394-16503394 on Build GRCh38
rs767011257 Chr.19: 16503335-16503336 on Build GRCh38
rs147838234 Chr.19: 16503380-16503380 on Build GRCh38
rs149899766 Chr.19: 16503275-16503275 on Build GRCh38
rs761747982 Chr.19: 16503379-16503379 on Build GRCh38
rs868053526 Chr.19: 16503306-16503306 on Build GRCh38
rs779542281 Chr.19: 16503330-16503330 on Build GRCh38
rs761576423 Chr.19: 16503336-16503336 on Build GRCh38
rs779536966 Chr.19: 16503446-16503446 on Build GRCh38
rs148603243 Chr.19: 16503315-16503315 on Build GRCh38
rs756149126 Chr.19: 16503325-16503325 on Build GRCh38
rs766481334 Chr.19: 16503339-16503339 on Build GRCh38
rs754110043 Chr.19: 16503418-16503418 on Build GRCh38
rs867293445 Chr.19: 16503347-16503347 on Build GRCh38
rs748851278 Chr.19: 16503376-16503376 on Build GRCh38
rs774820419 Chr.19: 16503259-16503259 on Build GRCh38
rs376391840 Chr.19: 16503344-16503344 on Build GRCh38
rs767687964 Chr.19: 16503318-16503318 on Build GRCh38
rs757021909 Chr.19: 16503316-16503316 on Build GRCh38
rs887340085 Chr.19: 16503432-16503432 on Build GRCh38
rs143897261 Chr.19: 16503304-16503304 on Build GRCh38
rs931464844 Chr.19: 16503387-16503387 on Build GRCh38
rs756739412 Chr.19: 16503280-16503281 on Build GRCh38
rs769209472 Chr.19: 16503332-16503332 on Build GRCh38
rs749847642 Chr.19: 16503329-16503329 on Build GRCh38
rs774939611 Chr.19: 16503333-16503333 on Build GRCh38
rs1001610668 Chr.19: 16503360-16503360 on Build GRCh38
rs760915024 Chr.19: 16503337-16503337 on Build GRCh38
rs372399570 Chr.19: 16503320-16503320 on Build GRCh38
rs770540187 Chr.19: 16503261-16503261 on Build GRCh38
rs534852339 Chr.19: 16503331-16503331 on Build GRCh38
rs762418021 Chr.19: 16503335-16503335 on Build GRCh38
rs1018537495 Chr.19: 16503450-16503450 on Build GRCh38
rs776334884 Chr.19: 16503269-16503269 on Build GRCh38
rs565938711 Chr.19: 16503313-16503313 on Build GRCh38
rs779883633 Chr.19: 16503343-16503344 on Build GRCh38
rs202132782 Chr.19: 16503343-16503343 on Build GRCh38
rs780137209 Chr.19: 16503327-16503327 on Build GRCh38
rs750081527 Chr.19: 16503337-16503338 on Build GRCh38
rs779632289 Chr.19: 16503342-16503342 on Build GRCh38
rs573681066 Chr.19: 16503392-16503392 on Build GRCh38

View Full Product Details