Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 81961725-81961980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA2D1
Entrez Gene ID
781
Gene Name
calcium voltage-gated channel auxiliary subunit alpha2delta 1
Gene Aliases
CACNA2, CACNL2A, CCHL2A, LINC01112, lncRNA-N3
UniGene
Hs.282151
Gene Chromosome Location
Chr.7: 81946444-82443921 on Build GRCh38
Transcripts
NM_000722.3 XM_005250570.2 XM_005250572.2 XM_005250573.2 XM_005250574.2 XM_006716118.2 XM_006716119.3 XM_006716120.3 XM_006716121.3 XM_011516570.2 XM_011516571.2 XM_011516572.2 XM_017012588.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6246508 c.2949C>A p.F983L Chr.7: 81961947-81961947 on Build GRCh38
4787145 c.2941T>C p.Y981H Chr.7: 81961955-81961955 on Build GRCh38
5419145 c.2910C>T p.F970F Chr.7: 81961950-81961950 on Build GRCh38
4778472 c.2937C>T p.T979T Chr.7: 81961959-81961959 on Build GRCh38
383496 c.2925T>C p.S975S Chr.7: 81961935-81961935 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377593515 Chr.7: 81961835-81961835 on Build GRCh38
rs911083218 Chr.7: 81961758-81961761 on Build GRCh38
rs929648340 Chr.7: 81961873-81961875 on Build GRCh38
rs1055682951 Chr.7: 81961956-81961956 on Build GRCh38
rs753645659 Chr.7: 81961854-81961855 on Build GRCh38
rs374073497 Chr.7: 81961941-81961941 on Build GRCh38
rs774285483 Chr.7: 81961940-81961940 on Build GRCh38
rs768408345 Chr.7: 81961942-81961942 on Build GRCh38
rs771237530 Chr.7: 81961929-81961929 on Build GRCh38
rs749485291 Chr.7: 81961856-81961856 on Build GRCh38
rs1025767914 Chr.7: 81961754-81961754 on Build GRCh38
rs368967221 Chr.7: 81961756-81961756 on Build GRCh38
rs35112512 Chr.7: 81961834-81961862 on Build GRCh38
rs202002627 Chr.7: 81961877-81961877 on Build GRCh38
rs754772847 Chr.7: 81961879-81961882 on Build GRCh38
rs878889643 Chr.7: 81961827-81961827 on Build GRCh38
rs952910720 Chr.7: 81961790-81961790 on Build GRCh38
rs776720930 Chr.7: 81961946-81961946 on Build GRCh38
rs534412371 Chr.7: 81961938-81961938 on Build GRCh38
rs199938654 Chr.7: 81961889-81961889 on Build GRCh38
rs541068286 Chr.7: 81961867-81961867 on Build GRCh38
rs374759811 Chr.7: 81961834-81961834 on Build GRCh38
rs941254514 Chr.7: 81961791-81961791 on Build GRCh38
rs777932597 Chr.7: 81961845-81961845 on Build GRCh38
rs72063149 Chr.7: 81961799-81961827 on Build GRCh38
rs918220897 Chr.7: 81961852-81961852 on Build GRCh38
rs767769534 Chr.7: 81961849-81961849 on Build GRCh38
rs372480982 Chr.7: 81961904-81961904 on Build GRCh38
rs761648569 Chr.7: 81961947-81961947 on Build GRCh38
rs572038424 Chr.7: 81961798-81961798 on Build GRCh38
rs879110905 Chr.7: 81961827-81961828 on Build GRCh38

View Full Product Details