Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 105296291-105296546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TGFBRAP1
Entrez Gene ID
9392
Gene Name
transforming growth factor beta receptor associated protein 1
Gene Aliases
TRAP-1, TRAP1, VPS3
UniGene
Hs.446350
Gene Chromosome Location
Chr.2: 105249404-105329714 on Build GRCh38
Transcripts
NM_001142621.2 NM_004257.5 XM_011512193.2 XM_017005339.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5859807 c.910G>A p.G304R Chr.2: 105296484-105296484 on Build GRCh38
4582558 c.970C>T p.R324C Chr.2: 105296424-105296424 on Build GRCh38
4759831 c.889G>C p.V297L Chr.2: 105296505-105296505 on Build GRCh38
6884616 c.895G>A p.V299I Chr.2: 105296499-105296499 on Build GRCh38
1751961 c.979G>A p.E327K Chr.2: 105296415-105296415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372720283 Chr.2: 105296423-105296423 on Build GRCh38
rs1006251544 Chr.2: 105296449-105296449 on Build GRCh38
rs757846181 Chr.2: 105296332-105296332 on Build GRCh38
rs748250358 Chr.2: 105296507-105296507 on Build GRCh38
rs768778929 Chr.2: 105296384-105296384 on Build GRCh38
rs754566078 Chr.2: 105296323-105296323 on Build GRCh38
rs956318914 Chr.2: 105296503-105296503 on Build GRCh38
rs369956167 Chr.2: 105296348-105296348 on Build GRCh38
rs151031175 Chr.2: 105296499-105296499 on Build GRCh38
rs752180337 Chr.2: 105296328-105296328 on Build GRCh38
rs780368094 Chr.2: 105296373-105296373 on Build GRCh38
rs772009140 Chr.2: 105296387-105296387 on Build GRCh38
rs199581714 Chr.2: 105296386-105296386 on Build GRCh38
rs372467313 Chr.2: 105296361-105296361 on Build GRCh38
rs375638406 Chr.2: 105296378-105296378 on Build GRCh38
rs762357954 Chr.2: 105296460-105296460 on Build GRCh38
rs764685549 Chr.2: 105296325-105296325 on Build GRCh38
rs752266174 Chr.2: 105296454-105296454 on Build GRCh38
rs756555392 Chr.2: 105296464-105296464 on Build GRCh38
rs149456678 Chr.2: 105296385-105296385 on Build GRCh38
rs764771412 Chr.2: 105296433-105296433 on Build GRCh38
rs148371972 Chr.2: 105296481-105296481 on Build GRCh38
rs781644089 Chr.2: 105296340-105296340 on Build GRCh38
rs758964139 Chr.2: 105296424-105296424 on Build GRCh38
rs78999474 Chr.2: 105296492-105296492 on Build GRCh38
rs772941262 Chr.2: 105296388-105296388 on Build GRCh38
rs746269191 Chr.2: 105296357-105296357 on Build GRCh38
rs765926055 Chr.2: 105296415-105296415 on Build GRCh38
rs923483752 Chr.2: 105296500-105296500 on Build GRCh38
rs748534171 Chr.2: 105296395-105296395 on Build GRCh38
rs750717995 Chr.2: 105296463-105296463 on Build GRCh38
rs760550026 Chr.2: 105296391-105296391 on Build GRCh38

View Full Product Details