Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 17497304-17497577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CECR2
Entrez Gene ID
27443
Gene Name
CECR2, histone acetyl-lysine reader
Gene Aliases
-
UniGene
Hs.231895
Gene Chromosome Location
Chr.22: 17359949-17558155 on Build GRCh38
Transcripts
NM_001290046.1 NM_001290047.1 XM_011546128.2 XM_011546129.2 XM_011546132.2 XM_017028785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4448271 c.252G>A p.Q84Q Chr.22: 17497499-17497499 on Build GRCh38
4977223 c.232C>T p.R78* Chr.22: 17497536-17497536 on Build GRCh38
3551963 c.208C>T p.R70C Chr.22: 17497512-17497512 on Build GRCh38
7129605 c.174G>C p.L58L Chr.22: 17497481-17497481 on Build GRCh38
4102402 c.157C>T p.P53S Chr.22: 17497404-17497404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756577418 Chr.22: 17497394-17497394 on Build GRCh38
rs780640857 Chr.22: 17497427-17497427 on Build GRCh38
rs749804824 Chr.22: 17497532-17497532 on Build GRCh38
rs1000953634 Chr.22: 17497536-17497536 on Build GRCh38
rs1016326737 Chr.22: 17497474-17497474 on Build GRCh38
rs560359613 Chr.22: 17497347-17497347 on Build GRCh38
rs774719391 Chr.22: 17497374-17497375 on Build GRCh38
rs983154308 Chr.22: 17497409-17497409 on Build GRCh38
rs371059170 Chr.22: 17497503-17497503 on Build GRCh38
rs781596070 Chr.22: 17497378-17497378 on Build GRCh38
rs746168371 Chr.22: 17497385-17497385 on Build GRCh38
rs1009969146 Chr.22: 17497520-17497520 on Build GRCh38
rs759290476 Chr.22: 17497470-17497470 on Build GRCh38
rs779029996 Chr.22: 17497553-17497553 on Build GRCh38
rs374467554 Chr.22: 17497509-17497509 on Build GRCh38
rs756489464 Chr.22: 17497514-17497514 on Build GRCh38
rs1019246940 Chr.22: 17497351-17497351 on Build GRCh38
rs1051316815 Chr.22: 17497499-17497499 on Build GRCh38
rs1042373950 Chr.22: 17497519-17497519 on Build GRCh38
rs577083118 Chr.22: 17497492-17497492 on Build GRCh38
rs772234587 Chr.22: 17497457-17497457 on Build GRCh38
rs748483742 Chr.22: 17497463-17497464 on Build GRCh38
rs755579495 Chr.22: 17497537-17497537 on Build GRCh38
rs762936835 Chr.22: 17497479-17497479 on Build GRCh38
rs965434338 Chr.22: 17497405-17497405 on Build GRCh38
rs756929292 Chr.22: 17497368-17497368 on Build GRCh38
rs750897323 Chr.22: 17497342-17497344 on Build GRCh38
rs751212399 Chr.22: 17497357-17497357 on Build GRCh38
rs774359092 Chr.22: 17497446-17497446 on Build GRCh38
rs756812082 Chr.22: 17497501-17497501 on Build GRCh38
rs755295727 Chr.22: 17497388-17497388 on Build GRCh38
rs775349031 Chr.22: 17497478-17497478 on Build GRCh38
rs950356762 Chr.22: 17497390-17497390 on Build GRCh38
rs748368295 Chr.22: 17497447-17497447 on Build GRCh38
rs200642767 Chr.22: 17497430-17497430 on Build GRCh38
rs781664783 Chr.22: 17497545-17497545 on Build GRCh38
rs921431915 Chr.22: 17497488-17497488 on Build GRCh38
rs368407320 Chr.22: 17497526-17497526 on Build GRCh38
rs763591044 Chr.22: 17497483-17497483 on Build GRCh38
rs768839952 Chr.22: 17497441-17497441 on Build GRCh38
rs765025297 Chr.22: 17497471-17497471 on Build GRCh38

View Full Product Details