Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29330636-29330871 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP1B1
Entrez Gene ID
162
Gene Name
adaptor related protein complex 1 beta 1 subunit
Gene Aliases
ADTB1, AP105A, BAM22, CLAPB2
UniGene
Hs.368794
Gene Chromosome Location
Chr.22: 29327680-29388583 on Build GRCh38
Transcripts
NM_001127.3 NM_001166019.1 NM_145730.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4481621 c.2506C>T p.R836W Chr.22: 29330707-29330707 on Build GRCh38
2936391 c.2578C>T p.Q860* Chr.22: 29330656-29330656 on Build GRCh38
1484140 c.2566G>A p.E856K Chr.22: 29330668-29330668 on Build GRCh38
4481620 c.2527C>T p.R843W Chr.22: 29330707-29330707 on Build GRCh38
2936390 c.2591G>C p.R864T Chr.22: 29330643-29330643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770087578 Chr.22: 29330742-29330742 on Build GRCh38
rs576322881 Chr.22: 29330794-29330794 on Build GRCh38
rs776990644 Chr.22: 29330734-29330734 on Build GRCh38
rs1008967556 Chr.22: 29330816-29330816 on Build GRCh38
rs905260689 Chr.22: 29330712-29330712 on Build GRCh38
rs906320206 Chr.22: 29330728-29330728 on Build GRCh38
rs543460834 Chr.22: 29330808-29330808 on Build GRCh38
rs1010535846 Chr.22: 29330819-29330819 on Build GRCh38
rs779005670 Chr.22: 29330787-29330787 on Build GRCh38
rs748320683 Chr.22: 29330707-29330707 on Build GRCh38
rs755965250 Chr.22: 29330687-29330687 on Build GRCh38
rs148036031 Chr.22: 29330697-29330697 on Build GRCh38
rs773948769 Chr.22: 29330666-29330666 on Build GRCh38
rs564890756 Chr.22: 29330843-29330843 on Build GRCh38
rs1017379362 Chr.22: 29330837-29330837 on Build GRCh38
rs762554974 Chr.22: 29330752-29330752 on Build GRCh38
rs374288161 Chr.22: 29330680-29330680 on Build GRCh38
rs542971487 Chr.22: 29330691-29330691 on Build GRCh38
rs771517508 Chr.22: 29330733-29330733 on Build GRCh38
rs369034576 Chr.22: 29330716-29330716 on Build GRCh38
rs555268901 Chr.22: 29330776-29330776 on Build GRCh38
rs759899475 Chr.22: 29330740-29330740 on Build GRCh38
rs777867797 Chr.22: 29330714-29330714 on Build GRCh38
rs775148176 Chr.22: 29330743-29330743 on Build GRCh38
rs890133861 Chr.22: 29330800-29330800 on Build GRCh38
rs780974385 Chr.22: 29330673-29330673 on Build GRCh38
rs377256748 Chr.22: 29330713-29330713 on Build GRCh38
rs751221758 Chr.22: 29330679-29330682 on Build GRCh38

View Full Product Details