Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161513738-161514011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR2A
Entrez Gene ID
2212
Gene Name
Fc fragment of IgG receptor IIa
Gene Aliases
CD32, CD32A, CDw32, FCG2, FCGR2, FCGR2A1, FcGR, IGFR2
UniGene
Hs.352642
Gene Chromosome Location
Chr.1: 161505415-161524048 on Build GRCh38
Transcripts
NM_001136219.1 NM_021642.3 XM_011509287.2 XM_011509290.2 XM_011509291.1 XM_017000663.1 XM_017000664.1 XM_017000665.1 XM_017000666.1 XM_017000667.1 XM_017000668.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5004090 c.771A>G p.Q257Q Chr.1: 161513926-161513926 on Build GRCh38
1601146 c.777+5G>T p.? Chr.1: 161513937-161513937 on Build GRCh38
898560 c.757G>A p.V253M Chr.1: 161513912-161513912 on Build GRCh38
5166205 c.740-1G>A p.? Chr.1: 161513894-161513894 on Build GRCh38
3740782 c.777+1G>A p.? Chr.1: 161513933-161513933 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751734756 Chr.1: 161513892-161513892 on Build GRCh38
rs545571774 Chr.1: 161513905-161513905 on Build GRCh38
rs374658515 Chr.1: 161513908-161513908 on Build GRCh38
rs780263997 Chr.1: 161513889-161513889 on Build GRCh38
rs409763 Chr.1: 161513933-161513933 on Build GRCh38
rs755168729 Chr.1: 161513949-161513949 on Build GRCh38
rs770971960 Chr.1: 161513977-161513977 on Build GRCh38
rs771912362 Chr.1: 161513931-161513931 on Build GRCh38
rs887396700 Chr.1: 161513783-161513783 on Build GRCh38
rs781233917 Chr.1: 161513901-161513901 on Build GRCh38
rs765896497 Chr.1: 161513870-161513870 on Build GRCh38
rs750916273 Chr.1: 161513943-161513943 on Build GRCh38
rs376496166 Chr.1: 161513847-161513847 on Build GRCh38
rs576577725 Chr.1: 161513846-161513846 on Build GRCh38
rs368278311 Chr.1: 161513966-161513966 on Build GRCh38
rs758721700 Chr.1: 161513881-161513881 on Build GRCh38
rs1057161029 Chr.1: 161513799-161513799 on Build GRCh38
rs395463 Chr.1: 161513960-161513960 on Build GRCh38
rs371392191 Chr.1: 161513824-161513824 on Build GRCh38
rs767143639 Chr.1: 161513863-161513863 on Build GRCh38
rs374120720 Chr.1: 161513958-161513958 on Build GRCh38
rs150021274 Chr.1: 161513896-161513896 on Build GRCh38
rs376207146 Chr.1: 161513946-161513946 on Build GRCh38
rs182370196 Chr.1: 161513837-161513837 on Build GRCh38
rs56157533 Chr.1: 161513866-161513866 on Build GRCh38
rs749190976 Chr.1: 161513971-161513971 on Build GRCh38
rs403988 Chr.1: 161513876-161513876 on Build GRCh38
rs773893338 Chr.1: 161513857-161513857 on Build GRCh38
rs770648578 Chr.1: 161513917-161513917 on Build GRCh38
rs759109384 Chr.1: 161513928-161513928 on Build GRCh38
rs373636398 Chr.1: 161513913-161513913 on Build GRCh38
rs375706777 Chr.1: 161513891-161513891 on Build GRCh38
rs371086260 Chr.1: 161513860-161513860 on Build GRCh38
rs575925747 Chr.1: 161513954-161513954 on Build GRCh38
rs572323543 Chr.1: 161513831-161513831 on Build GRCh38
rs534850522 Chr.1: 161513836-161513836 on Build GRCh38
rs149146966 Chr.1: 161513910-161513910 on Build GRCh38
rs773991787 Chr.1: 161513921-161513921 on Build GRCh38
rs765799102 Chr.1: 161513936-161513936 on Build GRCh38
rs199957022 Chr.1: 161513947-161513947 on Build GRCh38

View Full Product Details